Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176526659-176527162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF44
Entrez Gene ID
22838
Gene Name
ring finger protein 44
Gene Aliases
-
UniGene
Hs.434888
Gene Chromosome Location
Chr.5: 176526699-176543456 on Build GRCh38
Transcripts
NM_014901.4 XM_005265840.2 XM_005265841.4 XM_005265842.4 XM_005265843.2 XM_005265844.3 XM_005265845.4 XM_006714831.2 XM_006714832.2 XM_011534466.2 XM_011534467.2

SNP Detail

SNP ID Chromosome Location
rs935019974 Chr.5: 176527036-176527036 on Build GRCh38
rs921512617 Chr.5: 176527070-176527070 on Build GRCh38
rs557168750 Chr.5: 176526822-176526822 on Build GRCh38
rs776355644 Chr.5: 176527094-176527094 on Build GRCh38
rs991624367 Chr.5: 176526737-176526737 on Build GRCh38
rs887246659 Chr.5: 176526768-176526768 on Build GRCh38
rs548104353 Chr.5: 176526767-176526768 on Build GRCh38
rs996909242 Chr.5: 176526809-176526809 on Build GRCh38
rs942470633 Chr.5: 176526727-176526727 on Build GRCh38
rs1020818807 Chr.5: 176526918-176526918 on Build GRCh38
rs575629358 Chr.5: 176526867-176526867 on Build GRCh38
rs986312733 Chr.5: 176526719-176526719 on Build GRCh38
rs1042127708 Chr.5: 176527119-176527119 on Build GRCh38
rs1011198435 Chr.5: 176526776-176526776 on Build GRCh38
rs916770958 Chr.5: 176527051-176527062 on Build GRCh38
rs982804614 Chr.5: 176527004-176527004 on Build GRCh38
rs946663607 Chr.5: 176527085-176527085 on Build GRCh38
rs922331867 Chr.5: 176526990-176526990 on Build GRCh38
rs535686438 Chr.5: 176527080-176527080 on Build GRCh38
rs567220965 Chr.5: 176526700-176526700 on Build GRCh38
rs971465471 Chr.5: 176526978-176526978 on Build GRCh38
rs917358390 Chr.5: 176526974-176526977 on Build GRCh38
rs1052327144 Chr.5: 176526683-176526683 on Build GRCh38
rs1027899813 Chr.5: 176526908-176526908 on Build GRCh38
rs1034009102 Chr.5: 176526954-176526954 on Build GRCh38
rs1038986531 Chr.5: 176526782-176526782 on Build GRCh38
rs966584161 Chr.5: 176526961-176526961 on Build GRCh38
rs904991055 Chr.5: 176526818-176526818 on Build GRCh38
rs1044909986 Chr.5: 176526799-176526799 on Build GRCh38
rs748094126 Chr.5: 176526744-176526744 on Build GRCh38
rs568502767 Chr.5: 176526812-176526812 on Build GRCh38
rs952332115 Chr.5: 176526917-176526917 on Build GRCh38
rs979736295 Chr.5: 176526722-176526723 on Build GRCh38
rs182944891 Chr.5: 176526728-176526728 on Build GRCh38
rs192714079 Chr.5: 176526935-176526935 on Build GRCh38
rs952345519 Chr.5: 176526716-176526716 on Build GRCh38
rs1009293237 Chr.5: 176526916-176526916 on Build GRCh38
rs537183096 Chr.5: 176526682-176526682 on Build GRCh38
rs969499353 Chr.5: 176526695-176526695 on Build GRCh38
rs562712276 Chr.5: 176527029-176527030 on Build GRCh38
rs770862014 Chr.5: 176526971-176526971 on Build GRCh38
rs972308027 Chr.5: 176526707-176526707 on Build GRCh38
rs771941954 Chr.5: 176526891-176526891 on Build GRCh38
rs78282406 Chr.5: 176526712-176526712 on Build GRCh38
rs893716727 Chr.5: 176526765-176526765 on Build GRCh38
rs958775850 Chr.5: 176526960-176526960 on Build GRCh38
rs759319674 Chr.5: 176527112-176527112 on Build GRCh38
rs370259202 Chr.5: 176527122-176527122 on Build GRCh38
rs140405441 Chr.5: 176526706-176526706 on Build GRCh38
rs890773666 Chr.5: 176527097-176527097 on Build GRCh38
rs908717401 Chr.5: 176527010-176527010 on Build GRCh38
rs78596722 Chr.5: 176526948-176526948 on Build GRCh38
rs1024175821 Chr.5: 176526690-176526690 on Build GRCh38
rs893666177 Chr.5: 176526684-176526684 on Build GRCh38
rs1038649621 Chr.5: 176527054-176527055 on Build GRCh38
rs1052099566 Chr.5: 176527056-176527056 on Build GRCh38
rs1045356800 Chr.5: 176527087-176527087 on Build GRCh38
rs934963804 Chr.5: 176526745-176526745 on Build GRCh38
rs1035045673 Chr.5: 176526865-176526865 on Build GRCh38
rs534028162 Chr.5: 176526919-176526919 on Build GRCh38
rs545191229 Chr.5: 176526784-176526784 on Build GRCh38
rs910671710 Chr.5: 176526723-176526723 on Build GRCh38
rs1009324391 Chr.5: 176527118-176527118 on Build GRCh38
rs899504803 Chr.5: 176526806-176526806 on Build GRCh38
rs555040920 Chr.5: 176526686-176526686 on Build GRCh38
rs186868593 Chr.5: 176526898-176526898 on Build GRCh38
rs538009347 Chr.5: 176526756-176526756 on Build GRCh38
rs75813813 Chr.5: 176526701-176526701 on Build GRCh38
rs780052569 Chr.5: 176526743-176526743 on Build GRCh38
rs757240274 Chr.5: 176526870-176526870 on Build GRCh38

View Full Product Details