Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 218331972-218332468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRP15
Entrez Gene ID
51018
Gene Name
ribosomal RNA processing 15 homolog
Gene Aliases
CGI-115, KIAA0507
UniGene
Hs.660109
Gene Chromosome Location
Chr.1: 218285270-218337983 on Build GRCh38
Transcripts
NM_016052.3

SNP Detail

SNP ID Chromosome Location
rs981527907 Chr.1: 218332212-218332212 on Build GRCh38
rs898292018 Chr.1: 218332408-218332408 on Build GRCh38
rs978710708 Chr.1: 218332120-218332120 on Build GRCh38
rs150282295 Chr.1: 218332114-218332115 on Build GRCh38
rs1053337987 Chr.1: 218332290-218332290 on Build GRCh38
rs969726803 Chr.1: 218332209-218332209 on Build GRCh38
rs781699379 Chr.1: 218332386-218332386 on Build GRCh38
rs189407178 Chr.1: 218332434-218332434 on Build GRCh38
rs934892799 Chr.1: 218332284-218332284 on Build GRCh38
rs556548195 Chr.1: 218332095-218332095 on Build GRCh38
rs928392629 Chr.1: 218332046-218332046 on Build GRCh38
rs572310586 Chr.1: 218332231-218332231 on Build GRCh38
rs184711483 Chr.1: 218332082-218332082 on Build GRCh38
rs925275446 Chr.1: 218332157-218332157 on Build GRCh38
rs982513320 Chr.1: 218332016-218332016 on Build GRCh38
rs567355178 Chr.1: 218332116-218332116 on Build GRCh38
rs138653980 Chr.1: 218332369-218332369 on Build GRCh38
rs577980857 Chr.1: 218332336-218332336 on Build GRCh38
rs778901332 Chr.1: 218332325-218332326 on Build GRCh38
rs796314953 Chr.1: 218332345-218332345 on Build GRCh38
rs538517597 Chr.1: 218332292-218332292 on Build GRCh38
rs192254326 Chr.1: 218332206-218332206 on Build GRCh38
rs900050845 Chr.1: 218332208-218332208 on Build GRCh38
rs959757341 Chr.1: 218332412-218332412 on Build GRCh38
rs542097871 Chr.1: 218332151-218332151 on Build GRCh38
rs1045942030 Chr.1: 218332064-218332064 on Build GRCh38
rs974269811 Chr.1: 218332021-218332021 on Build GRCh38
rs1039095917 Chr.1: 218332330-218332330 on Build GRCh38
rs1039538329 Chr.1: 218332107-218332107 on Build GRCh38
rs948397930 Chr.1: 218332054-218332054 on Build GRCh38
rs995680366 Chr.1: 218332248-218332248 on Build GRCh38
rs375856278 Chr.1: 218332115-218332116 on Build GRCh38
rs187977901 Chr.1: 218332177-218332177 on Build GRCh38
rs900463791 Chr.1: 218332110-218332110 on Build GRCh38
rs1011612682 Chr.1: 218332106-218332106 on Build GRCh38
rs541275635 Chr.1: 218332372-218332372 on Build GRCh38
rs1015435644 Chr.1: 218332187-218332187 on Build GRCh38
rs991305382 Chr.1: 218332093-218332093 on Build GRCh38
rs1035388689 Chr.1: 218332377-218332377 on Build GRCh38
rs145982615 Chr.1: 218332307-218332307 on Build GRCh38
rs759431288 Chr.1: 218332118-218332118 on Build GRCh38
rs12059750 Chr.1: 218332176-218332176 on Build GRCh38
rs183504170 Chr.1: 218332362-218332362 on Build GRCh38
rs944155017 Chr.1: 218332203-218332203 on Build GRCh38
rs373150181 Chr.1: 218332399-218332399 on Build GRCh38
rs973814624 Chr.1: 218332205-218332205 on Build GRCh38
rs569922669 Chr.1: 218332105-218332105 on Build GRCh38
rs962571368 Chr.1: 218332196-218332196 on Build GRCh38
rs915572641 Chr.1: 218332047-218332047 on Build GRCh38
rs935334604 Chr.1: 218332172-218332172 on Build GRCh38

View Full Product Details