Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42510535-42511033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRP7A
Entrez Gene ID
27341
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
BK126B4.3, CGI-96
UniGene
Hs.745006
Gene Chromosome Location
Chr.22: 42508335-42519823 on Build GRCh38
Transcripts
NM_015703.4 NR_027786.1

Gene Symbol
SERHL
Entrez Gene ID
94009
Gene Name
serine hydrolase-like (pseudogene)
Gene Aliases
BK126B4.1, BK126B4.2, HS126B42, dJ222E13.1
UniGene
Hs.360940
Gene Chromosome Location
Chr.22: 42500579-42512560 on Build GRCh38
Transcripts
NM_015703.4 NR_027786.1

SNP Detail

SNP ID Chromosome Location
rs149533716 Chr.22: 42510688-42510688 on Build GRCh38
rs367610336 Chr.22: 42510725-42510725 on Build GRCh38
rs55883011 Chr.22: 42510807-42510807 on Build GRCh38
rs879146443 Chr.22: 42510653-42510653 on Build GRCh38
rs112254859 Chr.22: 42510928-42510928 on Build GRCh38
rs984768187 Chr.22: 42510638-42510638 on Build GRCh38
rs538822796 Chr.22: 42510703-42510703 on Build GRCh38
rs964747555 Chr.22: 42510992-42510992 on Build GRCh38
rs138516959 Chr.22: 42510879-42510879 on Build GRCh38
rs56076734 Chr.22: 42510944-42510944 on Build GRCh38
rs553707508 Chr.22: 42510803-42510803 on Build GRCh38
rs2003672 Chr.22: 42510832-42510832 on Build GRCh38
rs936409811 Chr.22: 42510875-42510875 on Build GRCh38
rs535297168 Chr.22: 42510768-42510768 on Build GRCh38
rs1024459968 Chr.22: 42510586-42510586 on Build GRCh38
rs2281103 Chr.22: 42510855-42510855 on Build GRCh38
rs972135799 Chr.22: 42510612-42510612 on Build GRCh38
rs1054689103 Chr.22: 42510702-42510702 on Build GRCh38
rs9623612 Chr.22: 42510924-42510924 on Build GRCh38
rs563755723 Chr.22: 42510842-42510842 on Build GRCh38
rs1017584695 Chr.22: 42510828-42510828 on Build GRCh38
rs1013167378 Chr.22: 42510722-42510722 on Build GRCh38
rs952031248 Chr.22: 42510626-42510626 on Build GRCh38
rs56211179 Chr.22: 42510673-42510673 on Build GRCh38
rs976015904 Chr.22: 42510846-42510846 on Build GRCh38
rs867038871 Chr.22: 42510904-42510904 on Build GRCh38
rs952082214 Chr.22: 42510775-42510775 on Build GRCh38
rs2003673 Chr.22: 42510947-42510947 on Build GRCh38
rs578262073 Chr.22: 42510830-42510830 on Build GRCh38
rs1026368375 Chr.22: 42510616-42510616 on Build GRCh38
rs55803321 Chr.22: 42510908-42510908 on Build GRCh38
rs368492230 Chr.22: 42510732-42510732 on Build GRCh38
rs560965992 Chr.22: 42510665-42510665 on Build GRCh38
rs976342435 Chr.22: 42510655-42510655 on Build GRCh38
rs765203094 Chr.22: 42510654-42510654 on Build GRCh38
rs568919240 Chr.22: 42510728-42510728 on Build GRCh38
rs188762316 Chr.22: 42510757-42510757 on Build GRCh38
rs879886935 Chr.22: 42510704-42510704 on Build GRCh38
rs971929143 Chr.22: 42510610-42510610 on Build GRCh38
rs907377200 Chr.22: 42510730-42510730 on Build GRCh38
rs1064877 Chr.22: 42510952-42510952 on Build GRCh38
rs566430379 Chr.22: 42510978-42510978 on Build GRCh38
rs200792519 Chr.22: 42510687-42510687 on Build GRCh38
rs554577456 Chr.22: 42510740-42510740 on Build GRCh38
rs73173063 Chr.22: 42510917-42510917 on Build GRCh38
rs528469974 Chr.22: 42510888-42510888 on Build GRCh38
rs549335444 Chr.22: 42510621-42510621 on Build GRCh38
rs997884493 Chr.22: 42510993-42510993 on Build GRCh38
rs878972849 Chr.22: 42510659-42510659 on Build GRCh38
rs371548479 Chr.22: 42510764-42510764 on Build GRCh38
rs1047913846 Chr.22: 42510941-42510941 on Build GRCh38
rs769798137 Chr.22: 42510650-42510650 on Build GRCh38
rs116229100 Chr.22: 42510871-42510871 on Build GRCh38
rs910487068 Chr.22: 42510641-42510641 on Build GRCh38
rs548459160 Chr.22: 42510677-42510677 on Build GRCh38
rs766411200 Chr.22: 42510718-42510718 on Build GRCh38
rs1018054291 Chr.22: 42510981-42510981 on Build GRCh38

View Full Product Details