Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6746112-6746610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIP10
Entrez Gene ID
9322
Gene Name
thyroid hormone receptor interactor 10
Gene Aliases
CIP4, HSTP, STOT, STP, TRIP-10
UniGene
Hs.515094
Gene Chromosome Location
Chr.19: 6739682-6751526 on Build GRCh38
Transcripts
NM_001288962.1 NM_001288963.1 NM_004240.3 NR_110231.1 XM_006722940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4973897 c.1045C>A p.Q349K Chr.19: 6746512-6746512 on Build GRCh38
4082634 c.989T>C p.V330A Chr.19: 6746456-6746456 on Build GRCh38
4804426 c.1217A>T p.Q406L Chr.19: 6746516-6746516 on Build GRCh38
4896157 c.1001A>T p.D334V Chr.19: 6746468-6746468 on Build GRCh38
5857036 c.1205G>A p.R402Q Chr.19: 6746504-6746504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754293339 Chr.19: 6746192-6746192 on Build GRCh38
rs760605986 Chr.19: 6746232-6746232 on Build GRCh38
rs763860073 Chr.19: 6746454-6746454 on Build GRCh38
rs994438071 Chr.19: 6746250-6746250 on Build GRCh38
rs201977475 Chr.19: 6746402-6746402 on Build GRCh38
rs915187512 Chr.19: 6746343-6746343 on Build GRCh38
rs756396774 Chr.19: 6746504-6746504 on Build GRCh38
rs762690761 Chr.19: 6746554-6746554 on Build GRCh38
rs770946008 Chr.19: 6746433-6746433 on Build GRCh38
rs528617196 Chr.19: 6746249-6746249 on Build GRCh38
rs748379269 Chr.19: 6746412-6746412 on Build GRCh38
rs1002905579 Chr.19: 6746133-6746133 on Build GRCh38
rs746975148 Chr.19: 6746527-6746527 on Build GRCh38
rs567473375 Chr.19: 6746380-6746380 on Build GRCh38
rs976875574 Chr.19: 6746290-6746290 on Build GRCh38
rs947936207 Chr.19: 6746224-6746224 on Build GRCh38
rs773003286 Chr.19: 6746544-6746544 on Build GRCh38
rs769833837 Chr.19: 6746146-6746146 on Build GRCh38
rs761419356 Chr.19: 6746458-6746458 on Build GRCh38
rs551451664 Chr.19: 6746271-6746271 on Build GRCh38
rs74349758 Chr.19: 6746439-6746439 on Build GRCh38
rs551250043 Chr.19: 6746327-6746327 on Build GRCh38
rs767414808 Chr.19: 6746157-6746157 on Build GRCh38
rs952831762 Chr.19: 6746173-6746173 on Build GRCh38
rs572578299 Chr.19: 6746369-6746369 on Build GRCh38
rs775416433 Chr.19: 6746434-6746434 on Build GRCh38
rs57513281 Chr.19: 6746582-6746582 on Build GRCh38
rs755480065 Chr.19: 6746575-6746575 on Build GRCh38
rs368978767 Chr.19: 6746253-6746253 on Build GRCh38
rs776732807 Chr.19: 6746144-6746144 on Build GRCh38
rs201134453 Chr.19: 6746528-6746528 on Build GRCh38
rs938726014 Chr.19: 6746246-6746246 on Build GRCh38
rs529912590 Chr.19: 6746169-6746169 on Build GRCh38
rs879061761 Chr.19: 6746270-6746270 on Build GRCh38
rs965491953 Chr.19: 6746182-6746182 on Build GRCh38
rs770999367 Chr.19: 6746521-6746521 on Build GRCh38
rs372729740 Chr.19: 6746578-6746578 on Build GRCh38
rs897223402 Chr.19: 6746415-6746415 on Build GRCh38
rs774406788 Chr.19: 6746523-6746523 on Build GRCh38
rs777997852 Chr.19: 6746505-6746505 on Build GRCh38
rs1028418634 Chr.19: 6746382-6746382 on Build GRCh38
rs760405696 Chr.19: 6746190-6746190 on Build GRCh38
rs199832105 Chr.19: 6746515-6746515 on Build GRCh38
rs989140890 Chr.19: 6746536-6746536 on Build GRCh38
rs756926704 Chr.19: 6746210-6746210 on Build GRCh38
rs553180479 Chr.19: 6746344-6746344 on Build GRCh38
rs911001175 Chr.19: 6746178-6746178 on Build GRCh38
rs766046716 Chr.19: 6746565-6746565 on Build GRCh38
rs976486042 Chr.19: 6746187-6746187 on Build GRCh38
rs773529727 Chr.19: 6746546-6746548 on Build GRCh38
rs201496765 Chr.19: 6746411-6746411 on Build GRCh38
rs965162001 Chr.19: 6746281-6746281 on Build GRCh38
rs1027261785 Chr.19: 6746252-6746252 on Build GRCh38
rs1045288012 Chr.19: 6746228-6746228 on Build GRCh38
rs927752797 Chr.19: 6746370-6746370 on Build GRCh38
rs571624171 Chr.19: 6746273-6746273 on Build GRCh38
rs1037604217 Chr.19: 6746251-6746251 on Build GRCh38
rs901311261 Chr.19: 6746513-6746513 on Build GRCh38
rs765075809 Chr.19: 6746542-6746542 on Build GRCh38
rs182836308 Chr.19: 6746283-6746283 on Build GRCh38
rs371917765 Chr.19: 6746463-6746463 on Build GRCh38
rs780737107 Chr.19: 6746393-6746393 on Build GRCh38
rs559872795 Chr.19: 6746214-6746214 on Build GRCh38
rs1021876399 Chr.19: 6746541-6746541 on Build GRCh38
rs747460405 Chr.19: 6746394-6746394 on Build GRCh38
rs935777427 Chr.19: 6746572-6746572 on Build GRCh38
rs574820524 Chr.19: 6746479-6746479 on Build GRCh38
rs768553936 Chr.19: 6746164-6746173 on Build GRCh38
rs537296174 Chr.19: 6746280-6746280 on Build GRCh38
rs981067258 Chr.19: 6746569-6746569 on Build GRCh38
rs1007091016 Chr.19: 6746259-6746259 on Build GRCh38
rs368066896 Chr.19: 6746457-6746457 on Build GRCh38
rs1010193855 Chr.19: 6746340-6746340 on Build GRCh38
rs546858335 Chr.19: 6746211-6746211 on Build GRCh38
rs954010549 Chr.19: 6746396-6746396 on Build GRCh38
rs556173810 Chr.19: 6746579-6746579 on Build GRCh38
rs929986823 Chr.19: 6746422-6746422 on Build GRCh38
rs927032918 Chr.19: 6746552-6746552 on Build GRCh38
rs1044171008 Chr.19: 6746262-6746262 on Build GRCh38
rs200373674 Chr.19: 6746413-6746413 on Build GRCh38
rs749575315 Chr.19: 6746584-6746584 on Build GRCh38
rs752074519 Chr.19: 6746481-6746481 on Build GRCh38
rs781461983 Chr.19: 6746503-6746503 on Build GRCh38
rs558228842 Chr.19: 6746389-6746389 on Build GRCh38
rs749929332 Chr.19: 6746388-6746388 on Build GRCh38
rs370411799 Chr.19: 6746418-6746418 on Build GRCh38
rs1032943268 Chr.19: 6746159-6746159 on Build GRCh38
rs1036097879 Chr.19: 6746140-6746140 on Build GRCh38
rs768938918 Chr.19: 6746257-6746257 on Build GRCh38
rs1057160416 Chr.19: 6746247-6746247 on Build GRCh38
rs167973 Chr.19: 6746341-6746341 on Build GRCh38
rs796807990 Chr.19: 6746293-6746293 on Build GRCh38
rs560872636 Chr.19: 6746148-6746148 on Build GRCh38
rs766802760 Chr.19: 6746191-6746191 on Build GRCh38
rs759521484 Chr.19: 6746156-6746156 on Build GRCh38
rs776329646 Chr.19: 6746533-6746533 on Build GRCh38
rs1030639790 Chr.19: 6746294-6746294 on Build GRCh38
rs142688804 Chr.19: 6746464-6746464 on Build GRCh38
rs79256079 Chr.19: 6746331-6746331 on Build GRCh38
rs573741547 Chr.19: 6746256-6746256 on Build GRCh38
rs986012413 Chr.19: 6746174-6746174 on Build GRCh38
rs755499266 Chr.19: 6746497-6746497 on Build GRCh38
rs749405574 Chr.19: 6746421-6746421 on Build GRCh38
rs751060964 Chr.19: 6746574-6746574 on Build GRCh38
rs200472405 Chr.19: 6746534-6746534 on Build GRCh38

View Full Product Details