Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 23999726-24000215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBXN2A
Entrez Gene ID
165324
Gene Name
UBX domain protein 2A
Gene Aliases
UBXD4
UniGene
Hs.591576
Gene Chromosome Location
Chr.2: 23927201-24001000 on Build GRCh38
Transcripts
NM_181713.3 XM_005264166.4 XM_005264167.4 XM_005264168.4 XM_005264169.4 XM_011532633.2 XM_017003517.1 XM_017003518.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
267722 c.742_743insA p.T250fs*5 Chr.2: 23999829-23999830 on Build GRCh38
4646685 c.751G>A p.A251T Chr.2: 23999838-23999838 on Build GRCh38
4133723 c.706G>T p.A236S Chr.2: 23999793-23999793 on Build GRCh38
7396613 c.772G>C p.E258Q Chr.2: 23999859-23999859 on Build GRCh38
4503499 c.640C>T p.P214S Chr.2: 23999727-23999727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140758090 Chr.2: 24000015-24000015 on Build GRCh38
rs753336862 Chr.2: 23999790-23999790 on Build GRCh38
rs1024258583 Chr.2: 24000133-24000133 on Build GRCh38
rs772682222 Chr.2: 23999749-23999749 on Build GRCh38
rs1041744278 Chr.2: 24000134-24000134 on Build GRCh38
rs1005080641 Chr.2: 23999984-23999984 on Build GRCh38
rs772354852 Chr.2: 23999861-23999861 on Build GRCh38
rs763697656 Chr.2: 23999771-23999771 on Build GRCh38
rs761612072 Chr.2: 23999791-23999791 on Build GRCh38
rs577603833 Chr.2: 24000115-24000115 on Build GRCh38
rs768756879 Chr.2: 23999875-23999883 on Build GRCh38
rs145330406 Chr.2: 23999831-23999831 on Build GRCh38
rs371072453 Chr.2: 23999868-23999868 on Build GRCh38
rs878946800 Chr.2: 23999965-23999965 on Build GRCh38
rs766100627 Chr.2: 23999916-23999916 on Build GRCh38
rs145877496 Chr.2: 24000064-24000064 on Build GRCh38
rs543592686 Chr.2: 23999982-23999982 on Build GRCh38
rs373217233 Chr.2: 23999807-23999807 on Build GRCh38
rs746053681 Chr.2: 23999851-23999851 on Build GRCh38
rs150980335 Chr.2: 23999864-23999864 on Build GRCh38
rs772774991 Chr.2: 23999903-23999903 on Build GRCh38
rs911438707 Chr.2: 24000053-24000053 on Build GRCh38
rs3088337 Chr.2: 24000072-24000072 on Build GRCh38
rs72851398 Chr.2: 24000159-24000159 on Build GRCh38
rs781412631 Chr.2: 23999911-23999912 on Build GRCh38
rs762902922 Chr.2: 23999974-23999974 on Build GRCh38
rs757766899 Chr.2: 24000032-24000032 on Build GRCh38
rs369003723 Chr.2: 24000093-24000094 on Build GRCh38
rs764790310 Chr.2: 23999803-23999803 on Build GRCh38
rs1039362292 Chr.2: 23999840-23999840 on Build GRCh38
rs751342826 Chr.2: 23999919-23999919 on Build GRCh38
rs779862238 Chr.2: 23999813-23999813 on Build GRCh38
rs201777835 Chr.2: 23999783-23999783 on Build GRCh38
rs747095744 Chr.2: 23999868-23999868 on Build GRCh38
rs762603745 Chr.2: 23999911-23999911 on Build GRCh38
rs890695301 Chr.2: 23999902-23999902 on Build GRCh38
rs780605417 Chr.2: 23999829-23999830 on Build GRCh38
rs10186915 Chr.2: 23999957-23999957 on Build GRCh38
rs761223439 Chr.2: 23999766-23999766 on Build GRCh38
rs764758491 Chr.2: 23999892-23999892 on Build GRCh38
rs376193605 Chr.2: 23999838-23999838 on Build GRCh38
rs761387092 Chr.2: 23999887-23999887 on Build GRCh38
rs148039467 Chr.2: 23999842-23999842 on Build GRCh38
rs778980014 Chr.2: 23999843-23999843 on Build GRCh38
rs970459625 Chr.2: 23999855-23999855 on Build GRCh38
rs749368535 Chr.2: 23999841-23999841 on Build GRCh38
rs942904875 Chr.2: 24000059-24000059 on Build GRCh38
rs754852674 Chr.2: 23999834-23999834 on Build GRCh38
rs750283614 Chr.2: 23999805-23999805 on Build GRCh38
rs149482799 Chr.2: 24000092-24000093 on Build GRCh38
rs775883351 Chr.2: 23999765-23999765 on Build GRCh38
rs776345312 Chr.2: 23999873-23999873 on Build GRCh38

View Full Product Details