Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35716035-35716538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB32
Entrez Gene ID
27033
Gene Name
zinc finger and BTB domain containing 32
Gene Aliases
FAXF, FAZF, Rog, TZFP, ZNF538
UniGene
Hs.99430
Gene Chromosome Location
Chr.19: 35704481-35717048 on Build GRCh38
Transcripts
NM_001316902.1 NM_001316903.1 NM_014383.2 XM_011526718.2 XM_017026588.1 XM_017026589.1 XM_017026590.1 XM_017026591.1 XM_017026592.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5030583 c.1249A>G p.T417A Chr.19: 35716537-35716537 on Build GRCh38
3388886 c.1242G>A p.S414S Chr.19: 35716530-35716530 on Build GRCh38
4919025 c.1160A>T p.Q387L Chr.19: 35716268-35716268 on Build GRCh38
1612091 c.1243G>C p.A415P Chr.19: 35716531-35716531 on Build GRCh38
7086708 c.1223A>T p.Q408L Chr.19: 35716511-35716511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778656736 Chr.19: 35716242-35716242 on Build GRCh38
rs756898471 Chr.19: 35716116-35716116 on Build GRCh38
rs747888035 Chr.19: 35716245-35716245 on Build GRCh38
rs754805810 Chr.19: 35716233-35716233 on Build GRCh38
rs993926752 Chr.19: 35716362-35716362 on Build GRCh38
rs758129693 Chr.19: 35716494-35716494 on Build GRCh38
rs776003721 Chr.19: 35716193-35716193 on Build GRCh38
rs2234371 Chr.19: 35716084-35716084 on Build GRCh38
rs769195780 Chr.19: 35716338-35716338 on Build GRCh38
rs751248060 Chr.19: 35716110-35716110 on Build GRCh38
rs754929684 Chr.19: 35716455-35716455 on Build GRCh38
rs965205426 Chr.19: 35716064-35716064 on Build GRCh38
rs1011700160 Chr.19: 35716230-35716230 on Build GRCh38
rs972552298 Chr.19: 35716384-35716384 on Build GRCh38
rs765417152 Chr.19: 35716378-35716378 on Build GRCh38
rs867346854 Chr.19: 35716199-35716199 on Build GRCh38
rs2234374 Chr.19: 35716200-35716200 on Build GRCh38
rs752490307 Chr.19: 35716472-35716472 on Build GRCh38
rs1053692319 Chr.19: 35716518-35716518 on Build GRCh38
rs1041039385 Chr.19: 35716080-35716080 on Build GRCh38
rs1025479988 Chr.19: 35716369-35716369 on Build GRCh38
rs201608304 Chr.19: 35716346-35716346 on Build GRCh38
rs753768462 Chr.19: 35716436-35716436 on Build GRCh38
rs2234373 Chr.19: 35716175-35716175 on Build GRCh38
rs769900860 Chr.19: 35716327-35716327 on Build GRCh38
rs367974286 Chr.19: 35716115-35716115 on Build GRCh38
rs747199049 Chr.19: 35716170-35716170 on Build GRCh38
rs766137981 Chr.19: 35716277-35716277 on Build GRCh38
rs775930692 Chr.19: 35716328-35716328 on Build GRCh38
rs745945734 Chr.19: 35716320-35716320 on Build GRCh38
rs762229059 Chr.19: 35716348-35716348 on Build GRCh38
rs930706032 Chr.19: 35716411-35716411 on Build GRCh38
rs549898834 Chr.19: 35716188-35716188 on Build GRCh38
rs771046426 Chr.19: 35716174-35716174 on Build GRCh38
rs959309395 Chr.19: 35716284-35716284 on Build GRCh38
rs750424655 Chr.19: 35716134-35716134 on Build GRCh38
rs928844453 Chr.19: 35716440-35716440 on Build GRCh38
rs371476322 Chr.19: 35716217-35716217 on Build GRCh38
rs774879839 Chr.19: 35716345-35716345 on Build GRCh38
rs919797152 Chr.19: 35716399-35716399 on Build GRCh38
rs780820898 Chr.19: 35716127-35716127 on Build GRCh38
rs538151412 Chr.19: 35716355-35716355 on Build GRCh38
rs768425181 Chr.19: 35716156-35716156 on Build GRCh38
rs368121024 Chr.19: 35716057-35716057 on Build GRCh38
rs200892981 Chr.19: 35716135-35716135 on Build GRCh38
rs749041875 Chr.19: 35716154-35716154 on Build GRCh38
rs767231160 Chr.19: 35716209-35716209 on Build GRCh38
rs143646755 Chr.19: 35716173-35716173 on Build GRCh38
rs772864287 Chr.19: 35716433-35716433 on Build GRCh38
rs781404007 Chr.19: 35716319-35716319 on Build GRCh38
rs1043442202 Chr.19: 35716283-35716283 on Build GRCh38
rs549110178 Chr.19: 35716205-35716205 on Build GRCh38
rs983921139 Chr.19: 35716392-35716392 on Build GRCh38
rs2234375 Chr.19: 35716434-35716434 on Build GRCh38
rs187428056 Chr.19: 35716349-35716349 on Build GRCh38
rs765115444 Chr.19: 35716465-35716465 on Build GRCh38
rs943600262 Chr.19: 35716061-35716061 on Build GRCh38
rs201490110 Chr.19: 35716085-35716085 on Build GRCh38
rs763306439 Chr.19: 35716090-35716090 on Build GRCh38
rs566021481 Chr.19: 35716224-35716224 on Build GRCh38
rs2234372 Chr.19: 35716140-35716140 on Build GRCh38
rs199720480 Chr.19: 35716329-35716329 on Build GRCh38
rs867536394 Chr.19: 35716192-35716192 on Build GRCh38
rs753807569 Chr.19: 35716226-35716226 on Build GRCh38
rs761538387 Chr.19: 35716207-35716207 on Build GRCh38
rs563278405 Chr.19: 35716306-35716310 on Build GRCh38
rs758527029 Chr.19: 35716471-35716471 on Build GRCh38
rs368774016 Chr.19: 35716138-35716138 on Build GRCh38
rs763849354 Chr.19: 35716105-35716105 on Build GRCh38
rs938915755 Chr.19: 35716467-35716467 on Build GRCh38
rs549832682 Chr.19: 35716213-35716213 on Build GRCh38
rs570044531 Chr.19: 35716191-35716191 on Build GRCh38
rs568164296 Chr.19: 35716466-35716466 on Build GRCh38
rs992064628 Chr.19: 35716290-35716290 on Build GRCh38
rs376730289 Chr.19: 35716311-35716311 on Build GRCh38
rs780001367 Chr.19: 35716152-35716152 on Build GRCh38
rs778035492 Chr.19: 35716162-35716162 on Build GRCh38
rs779112324 Chr.19: 35716451-35716451 on Build GRCh38
rs764552810 Chr.19: 35716201-35716201 on Build GRCh38

View Full Product Details