Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57807228-57807751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF552
Entrez Gene ID
79818
Gene Name
zinc finger protein 552
Gene Aliases
-
UniGene
Hs.560727
Gene Chromosome Location
Chr.19: 57807082-57814913 on Build GRCh38
Transcripts
NM_024762.3 XM_005259267.3

SNP Detail

SNP ID Chromosome Location
rs1010842504 Chr.19: 57807536-57807536 on Build GRCh38
rs147134439 Chr.19: 57807542-57807542 on Build GRCh38
rs949120459 Chr.19: 57807715-57807715 on Build GRCh38
rs1033497644 Chr.19: 57807512-57807512 on Build GRCh38
rs561890158 Chr.19: 57807330-57807330 on Build GRCh38
rs966245361 Chr.19: 57807553-57807558 on Build GRCh38
rs879035406 Chr.19: 57807359-57807359 on Build GRCh38
rs56276301 Chr.19: 57807338-57807338 on Build GRCh38
rs975132252 Chr.19: 57807723-57807724 on Build GRCh38
rs549953961 Chr.19: 57807708-57807708 on Build GRCh38
rs982269135 Chr.19: 57807563-57807563 on Build GRCh38
rs746267438 Chr.19: 57807688-57807688 on Build GRCh38
rs921132671 Chr.19: 57807724-57807739 on Build GRCh38
rs927766732 Chr.19: 57807597-57807597 on Build GRCh38
rs779203155 Chr.19: 57807551-57807551 on Build GRCh38
rs976740357 Chr.19: 57807562-57807562 on Build GRCh38
rs757715716 Chr.19: 57807534-57807534 on Build GRCh38
rs57000025 Chr.19: 57807519-57807519 on Build GRCh38
rs547156538 Chr.19: 57807454-57807454 on Build GRCh38
rs950936544 Chr.19: 57807646-57807646 on Build GRCh38
rs1040463651 Chr.19: 57807438-57807438 on Build GRCh38
rs1000739033 Chr.19: 57807507-57807507 on Build GRCh38
rs751218964 Chr.19: 57807297-57807297 on Build GRCh38
rs1050361700 Chr.19: 57807385-57807385 on Build GRCh38
rs60267176 Chr.19: 57807489-57807489 on Build GRCh38
rs929532717 Chr.19: 57807312-57807312 on Build GRCh38
rs949309857 Chr.19: 57807422-57807422 on Build GRCh38
rs930625276 Chr.19: 57807355-57807355 on Build GRCh38
rs112468548 Chr.19: 57807296-57807296 on Build GRCh38
rs562648189 Chr.19: 57807264-57807264 on Build GRCh38
rs756188466 Chr.19: 57807598-57807598 on Build GRCh38
rs920872635 Chr.19: 57807341-57807341 on Build GRCh38
rs1049280475 Chr.19: 57807317-57807317 on Build GRCh38
rs185966429 Chr.19: 57807468-57807468 on Build GRCh38
rs972226323 Chr.19: 57807657-57807657 on Build GRCh38
rs1050643787 Chr.19: 57807691-57807691 on Build GRCh38
rs1030906543 Chr.19: 57807479-57807479 on Build GRCh38
rs766147496 Chr.19: 57807314-57807315 on Build GRCh38
rs976243626 Chr.19: 57807280-57807280 on Build GRCh38
rs189811423 Chr.19: 57807675-57807675 on Build GRCh38
rs1042184282 Chr.19: 57807450-57807450 on Build GRCh38
rs551609149 Chr.19: 57807327-57807327 on Build GRCh38
rs962506455 Chr.19: 57807637-57807637 on Build GRCh38
rs908940997 Chr.19: 57807667-57807667 on Build GRCh38
rs754300943 Chr.19: 57807416-57807416 on Build GRCh38
rs752711318 Chr.19: 57807587-57807587 on Build GRCh38
rs1026603916 Chr.19: 57807599-57807599 on Build GRCh38
rs201638186 Chr.19: 57807337-57807337 on Build GRCh38
rs143547809 Chr.19: 57807541-57807541 on Build GRCh38
rs900994032 Chr.19: 57807469-57807469 on Build GRCh38
rs780631056 Chr.19: 57807387-57807387 on Build GRCh38
rs999388684 Chr.19: 57807473-57807473 on Build GRCh38
rs140334337 Chr.19: 57807689-57807689 on Build GRCh38
rs61117701 Chr.19: 57807491-57807491 on Build GRCh38
rs896376651 Chr.19: 57807398-57807398 on Build GRCh38
rs115106669 Chr.19: 57807684-57807684 on Build GRCh38

View Full Product Details