Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26934301-26934807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00202-1
Entrez Gene ID
387644
Gene Name
long intergenic non-protein coding RNA 202-1
Gene Aliases
C10orf51, LINC00202, NCRNA00202, bB27G4.1
UniGene
Hs.529965
Gene Chromosome Location
Chr.10: 26931206-26942001 on Build GRCh38
Transcripts
NR_026795.1

SNP Detail

SNP ID Chromosome Location
rs1057358577 Chr.10: 26934778-26934778 on Build GRCh38
rs397846831 Chr.10: 26934649-26934650 on Build GRCh38
rs544896611 Chr.10: 26934487-26934487 on Build GRCh38
rs79873121 Chr.10: 26934338-26934338 on Build GRCh38
rs911816414 Chr.10: 26934675-26934675 on Build GRCh38
rs555986112 Chr.10: 26934524-26934524 on Build GRCh38
rs907649463 Chr.10: 26934412-26934412 on Build GRCh38
rs747582203 Chr.10: 26934410-26934410 on Build GRCh38
rs189051709 Chr.10: 26934533-26934533 on Build GRCh38
rs931915767 Chr.10: 26934429-26934429 on Build GRCh38
rs570510479 Chr.10: 26934498-26934498 on Build GRCh38
rs531585472 Chr.10: 26934354-26934354 on Build GRCh38
rs980803586 Chr.10: 26934380-26934380 on Build GRCh38
rs891897461 Chr.10: 26934512-26934512 on Build GRCh38
rs192960539 Chr.10: 26934717-26934717 on Build GRCh38
rs371842538 Chr.10: 26934389-26934389 on Build GRCh38
rs73596331 Chr.10: 26934373-26934373 on Build GRCh38
rs138092914 Chr.10: 26934417-26934417 on Build GRCh38
rs576038245 Chr.10: 26934472-26934472 on Build GRCh38
rs935163384 Chr.10: 26934504-26934504 on Build GRCh38
rs141556674 Chr.10: 26934332-26934332 on Build GRCh38
rs1011692737 Chr.10: 26934521-26934521 on Build GRCh38
rs915118533 Chr.10: 26934735-26934735 on Build GRCh38
rs572094603 Chr.10: 26934699-26934699 on Build GRCh38
rs947445027 Chr.10: 26934738-26934738 on Build GRCh38
rs577356924 Chr.10: 26934748-26934748 on Build GRCh38
rs935149458 Chr.10: 26934698-26934698 on Build GRCh38
rs112468009 Chr.10: 26934416-26934416 on Build GRCh38
rs567507046 Chr.10: 26934405-26934405 on Build GRCh38
rs987366527 Chr.10: 26934646-26934647 on Build GRCh38
rs766927097 Chr.10: 26934488-26934488 on Build GRCh38
rs1021665935 Chr.10: 26934375-26934375 on Build GRCh38
rs201675934 Chr.10: 26934345-26934345 on Build GRCh38
rs535429396 Chr.10: 26934613-26934613 on Build GRCh38
rs1005477285 Chr.10: 26934591-26934591 on Build GRCh38
rs370471716 Chr.10: 26934644-26934645 on Build GRCh38
rs963078163 Chr.10: 26934604-26934604 on Build GRCh38
rs553651263 Chr.10: 26934428-26934428 on Build GRCh38
rs112469050 Chr.10: 26934523-26934523 on Build GRCh38
rs960213463 Chr.10: 26934588-26934588 on Build GRCh38
rs1001962034 Chr.10: 26934766-26934766 on Build GRCh38
rs1015076422 Chr.10: 26934601-26934601 on Build GRCh38
rs35120720 Chr.10: 26934390-26934390 on Build GRCh38
rs564292566 Chr.10: 26934643-26934644 on Build GRCh38
rs780786906 Chr.10: 26934390-26934390 on Build GRCh38
rs953167879 Chr.10: 26934628-26934628 on Build GRCh38
rs562256244 Chr.10: 26934363-26934363 on Build GRCh38
rs752966853 Chr.10: 26934724-26934724 on Build GRCh38
rs555369571 Chr.10: 26934625-26934625 on Build GRCh38
rs185227363 Chr.10: 26934749-26934749 on Build GRCh38
rs11015388 Chr.10: 26934713-26934713 on Build GRCh38
rs117573983 Chr.10: 26934352-26934352 on Build GRCh38
rs1053170034 Chr.10: 26934510-26934510 on Build GRCh38
rs35951564 Chr.10: 26934650-26934651 on Build GRCh38

View Full Product Details