Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 44959416-44959913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASSF4
Entrez Gene ID
83937
Gene Name
Ras association domain family member 4
Gene Aliases
AD037
UniGene
Hs.522895
Gene Chromosome Location
Chr.10: 44959753-44994724 on Build GRCh38
Transcripts
NM_032023.3 XM_005271833.4 XM_005271835.4 XM_006718013.3 XM_006718014.3 XM_011540255.2 XM_017016754.1 XM_017016755.1 XM_017016756.1 XM_017016757.1 XM_017016758.1 NR_033842.1

Gene Symbol
TMEM72-AS1
Entrez Gene ID
220980
Gene Name
TMEM72 antisense RNA 1
Gene Aliases
-
UniGene
Hs.568801
Gene Chromosome Location
Chr.10: 44811024-44959689 on Build GRCh38
Transcripts
NM_032023.3 XM_005271833.4 XM_005271835.4 XM_006718013.3 XM_006718014.3 XM_011540255.2 XM_017016754.1 XM_017016755.1 XM_017016756.1 XM_017016757.1 XM_017016758.1 NR_033842.1

SNP Detail

SNP ID Chromosome Location
rs866456137 Chr.10: 44959757-44959757 on Build GRCh38
rs867423589 Chr.10: 44959756-44959756 on Build GRCh38
rs998476036 Chr.10: 44959740-44959740 on Build GRCh38
rs539252182 Chr.10: 44959844-44959844 on Build GRCh38
rs1030262834 Chr.10: 44959444-44959444 on Build GRCh38
rs898467037 Chr.10: 44959434-44959434 on Build GRCh38
rs751017958 Chr.10: 44959812-44959812 on Build GRCh38
rs1009290224 Chr.10: 44959572-44959572 on Build GRCh38
rs903643062 Chr.10: 44959765-44959765 on Build GRCh38
rs955495538 Chr.10: 44959446-44959446 on Build GRCh38
rs920029679 Chr.10: 44959694-44959694 on Build GRCh38
rs546788550 Chr.10: 44959467-44959467 on Build GRCh38
rs891323800 Chr.10: 44959721-44959722 on Build GRCh38
rs1041331309 Chr.10: 44959746-44959746 on Build GRCh38
rs890015728 Chr.10: 44959716-44959716 on Build GRCh38
rs977903811 Chr.10: 44959819-44959819 on Build GRCh38
rs890077469 Chr.10: 44959512-44959512 on Build GRCh38
rs897318289 Chr.10: 44959714-44959714 on Build GRCh38
rs555738785 Chr.10: 44959835-44959835 on Build GRCh38
rs569355314 Chr.10: 44959690-44959690 on Build GRCh38
rs959058202 Chr.10: 44959788-44959788 on Build GRCh38
rs974136324 Chr.10: 44959601-44959601 on Build GRCh38
rs926273378 Chr.10: 44959602-44959602 on Build GRCh38
rs1008396765 Chr.10: 44959526-44959526 on Build GRCh38
rs920002271 Chr.10: 44959826-44959826 on Build GRCh38
rs571552022 Chr.10: 44959492-44959492 on Build GRCh38
rs917832849 Chr.10: 44959801-44959801 on Build GRCh38
rs925078973 Chr.10: 44959831-44959831 on Build GRCh38
rs755674308 Chr.10: 44959838-44959838 on Build GRCh38
rs961161222 Chr.10: 44959795-44959795 on Build GRCh38
rs191514124 Chr.10: 44959882-44959882 on Build GRCh38
rs1041825790 Chr.10: 44959657-44959657 on Build GRCh38
rs1051338813 Chr.10: 44959702-44959702 on Build GRCh38
rs901578673 Chr.10: 44959558-44959558 on Build GRCh38
rs1029899795 Chr.10: 44959747-44959747 on Build GRCh38
rs1010410300 Chr.10: 44959763-44959763 on Build GRCh38
rs532626054 Chr.10: 44959546-44959546 on Build GRCh38
rs1004381291 Chr.10: 44959738-44959738 on Build GRCh38
rs972753725 Chr.10: 44959693-44959693 on Build GRCh38
rs917721765 Chr.10: 44959614-44959614 on Build GRCh38
rs1049790140 Chr.10: 44959701-44959701 on Build GRCh38
rs536219397 Chr.10: 44959776-44959776 on Build GRCh38
rs931298034 Chr.10: 44959699-44959699 on Build GRCh38
rs931438073 Chr.10: 44959833-44959833 on Build GRCh38
rs968086593 Chr.10: 44959589-44959589 on Build GRCh38
rs1034012300 Chr.10: 44959603-44959603 on Build GRCh38
rs1000921387 Chr.10: 44959769-44959769 on Build GRCh38
rs972574024 Chr.10: 44959798-44959798 on Build GRCh38
rs1024851611 Chr.10: 44959783-44959783 on Build GRCh38
rs959045693 Chr.10: 44959777-44959777 on Build GRCh38
rs945196157 Chr.10: 44959638-44959638 on Build GRCh38
rs891943332 Chr.10: 44959762-44959762 on Build GRCh38
rs11593887 Chr.10: 44959499-44959499 on Build GRCh38
rs991941490 Chr.10: 44959610-44959610 on Build GRCh38
rs12773699 Chr.10: 44959779-44959779 on Build GRCh38
rs924691468 Chr.10: 44959682-44959682 on Build GRCh38
rs187129897 Chr.10: 44959583-44959583 on Build GRCh38
rs368670323 Chr.10: 44959648-44959648 on Build GRCh38
rs117957541 Chr.10: 44959607-44959607 on Build GRCh38
rs1021116984 Chr.10: 44959764-44959764 on Build GRCh38
rs979157209 Chr.10: 44959596-44959596 on Build GRCh38

View Full Product Details