Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45722197-45722704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507384
Entrez Gene ID
100507384
Gene Name
uncharacterized LOC100507384
Gene Aliases
-
UniGene
Hs.259426
Gene Chromosome Location
Chr.11: 45722308-45724555 on Build GRCh38
Transcripts
NR_047569.3

SNP Detail

SNP ID Chromosome Location
rs964796233 Chr.11: 45722294-45722294 on Build GRCh38
rs1042833352 Chr.11: 45722642-45722642 on Build GRCh38
rs182792182 Chr.11: 45722549-45722549 on Build GRCh38
rs2593731 Chr.11: 45722639-45722639 on Build GRCh38
rs955652354 Chr.11: 45722390-45722390 on Build GRCh38
rs35881131 Chr.11: 45722487-45722487 on Build GRCh38
rs998550791 Chr.11: 45722668-45722668 on Build GRCh38
rs769783501 Chr.11: 45722593-45722593 on Build GRCh38
rs563503284 Chr.11: 45722345-45722345 on Build GRCh38
rs188252968 Chr.11: 45722429-45722429 on Build GRCh38
rs1055666368 Chr.11: 45722268-45722268 on Build GRCh38
rs1021760816 Chr.11: 45722319-45722319 on Build GRCh38
rs563909056 Chr.11: 45722405-45722405 on Build GRCh38
rs745967213 Chr.11: 45722386-45722386 on Build GRCh38
rs919978164 Chr.11: 45722440-45722440 on Build GRCh38
rs1018481649 Chr.11: 45722422-45722422 on Build GRCh38
rs114550454 Chr.11: 45722571-45722571 on Build GRCh38
rs1056078953 Chr.11: 45722622-45722622 on Build GRCh38
rs528737339 Chr.11: 45722523-45722523 on Build GRCh38
rs748549490 Chr.11: 45722614-45722614 on Build GRCh38
rs997050805 Chr.11: 45722327-45722327 on Build GRCh38
rs938457177 Chr.11: 45722477-45722477 on Build GRCh38
rs948796362 Chr.11: 45722436-45722436 on Build GRCh38
rs768429419 Chr.11: 45722274-45722288 on Build GRCh38
rs539334420 Chr.11: 45722255-45722255 on Build GRCh38
rs982805175 Chr.11: 45722505-45722505 on Build GRCh38
rs955540010 Chr.11: 45722664-45722664 on Build GRCh38
rs974191094 Chr.11: 45722435-45722435 on Build GRCh38
rs529289472 Chr.11: 45722419-45722419 on Build GRCh38
rs998455684 Chr.11: 45722348-45722348 on Build GRCh38
rs990526914 Chr.11: 45722427-45722427 on Build GRCh38
rs116548716 Chr.11: 45722480-45722480 on Build GRCh38
rs184436211 Chr.11: 45722296-45722296 on Build GRCh38
rs938562170 Chr.11: 45722243-45722243 on Build GRCh38
rs1011329085 Chr.11: 45722299-45722299 on Build GRCh38
rs902966797 Chr.11: 45722656-45722656 on Build GRCh38
rs757398846 Chr.11: 45722306-45722306 on Build GRCh38
rs894298146 Chr.11: 45722270-45722270 on Build GRCh38
rs955653708 Chr.11: 45722377-45722377 on Build GRCh38
rs748087560 Chr.11: 45722466-45722466 on Build GRCh38
rs114396759 Chr.11: 45722637-45722637 on Build GRCh38
rs1044287090 Chr.11: 45722450-45722450 on Build GRCh38

View Full Product Details