Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86957827-86958340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506368
Entrez Gene ID
100506368
Gene Name
uncharacterized LOC100506368
Gene Aliases
-
UniGene
Hs.559920
Gene Chromosome Location
Chr.11: 86955621-87000950 on Build GRCh38
Transcripts
NR_038905.1

SNP Detail

SNP ID Chromosome Location
rs145713226 Chr.11: 86957856-86957856 on Build GRCh38
rs1023604824 Chr.11: 86958092-86958092 on Build GRCh38
rs888669832 Chr.11: 86958295-86958295 on Build GRCh38
rs543218032 Chr.11: 86958106-86958106 on Build GRCh38
rs542453461 Chr.11: 86958019-86958019 on Build GRCh38
rs897304003 Chr.11: 86958012-86958012 on Build GRCh38
rs188203568 Chr.11: 86958065-86958065 on Build GRCh38
rs985764847 Chr.11: 86958216-86958216 on Build GRCh38
rs938174455 Chr.11: 86958178-86958184 on Build GRCh38
rs970760982 Chr.11: 86958098-86958098 on Build GRCh38
rs903088368 Chr.11: 86958034-86958034 on Build GRCh38
rs888700937 Chr.11: 86958064-86958064 on Build GRCh38
rs776039986 Chr.11: 86957927-86957927 on Build GRCh38
rs144872786 Chr.11: 86958145-86958145 on Build GRCh38
rs1041587200 Chr.11: 86958006-86958006 on Build GRCh38
rs1026373488 Chr.11: 86958123-86958123 on Build GRCh38
rs985410714 Chr.11: 86958162-86958162 on Build GRCh38
rs796175615 Chr.11: 86957873-86957873 on Build GRCh38
rs915123817 Chr.11: 86958171-86958171 on Build GRCh38
rs944378920 Chr.11: 86958235-86958235 on Build GRCh38
rs994773912 Chr.11: 86958028-86958028 on Build GRCh38
rs965808049 Chr.11: 86958115-86958115 on Build GRCh38
rs1049047233 Chr.11: 86958282-86958282 on Build GRCh38
rs532328413 Chr.11: 86958315-86958315 on Build GRCh38
rs1007379525 Chr.11: 86958308-86958308 on Build GRCh38
rs72963453 Chr.11: 86958033-86958033 on Build GRCh38
rs926771494 Chr.11: 86957938-86957938 on Build GRCh38
rs930120319 Chr.11: 86958257-86958257 on Build GRCh38
rs74649650 Chr.11: 86958007-86958007 on Build GRCh38
rs911578947 Chr.11: 86958220-86958220 on Build GRCh38
rs919039026 Chr.11: 86958251-86958251 on Build GRCh38
rs769252380 Chr.11: 86957999-86957999 on Build GRCh38
rs1054773250 Chr.11: 86958283-86958283 on Build GRCh38
rs938187407 Chr.11: 86957974-86957974 on Build GRCh38
rs1032930039 Chr.11: 86958067-86958067 on Build GRCh38
rs1007430545 Chr.11: 86958112-86958112 on Build GRCh38
rs1012167626 Chr.11: 86958087-86958087 on Build GRCh38
rs1009521238 Chr.11: 86958029-86958029 on Build GRCh38
rs1049333559 Chr.11: 86957903-86957903 on Build GRCh38
rs879555431 Chr.11: 86958297-86958297 on Build GRCh38
rs1042495082 Chr.11: 86958244-86958244 on Build GRCh38
rs148285107 Chr.11: 86958205-86958205 on Build GRCh38
rs78483160 Chr.11: 86958008-86958008 on Build GRCh38
rs4943965 Chr.11: 86958236-86958236 on Build GRCh38
rs141017019 Chr.11: 86957970-86957970 on Build GRCh38
rs137971742 Chr.11: 86957894-86957894 on Build GRCh38
rs956557733 Chr.11: 86958138-86958138 on Build GRCh38
rs915178767 Chr.11: 86957865-86957865 on Build GRCh38
rs926887653 Chr.11: 86958169-86958169 on Build GRCh38
rs889074052 Chr.11: 86958301-86958301 on Build GRCh38
rs553109106 Chr.11: 86957896-86957896 on Build GRCh38
rs767168983 Chr.11: 86958265-86958265 on Build GRCh38
rs563278198 Chr.11: 86958117-86958117 on Build GRCh38
rs1000497308 Chr.11: 86958066-86958066 on Build GRCh38
rs944451939 Chr.11: 86957976-86957976 on Build GRCh38
rs763647932 Chr.11: 86958113-86958113 on Build GRCh38

View Full Product Details