Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53045393-53045900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC283335
Entrez Gene ID
283335
Gene Name
uncharacterized LOC283335
Gene Aliases
-
UniGene
Hs.657060
Gene Chromosome Location
Chr.12: 53043189-53054438 on Build GRCh38
Transcripts
NR_033854.1

SNP Detail

SNP ID Chromosome Location
rs180860554 Chr.12: 53045784-53045784 on Build GRCh38
rs547055759 Chr.12: 53045422-53045422 on Build GRCh38
rs565703597 Chr.12: 53045494-53045494 on Build GRCh38
rs1015844987 Chr.12: 53045582-53045582 on Build GRCh38
rs906726234 Chr.12: 53045588-53045588 on Build GRCh38
rs556514970 Chr.12: 53045530-53045530 on Build GRCh38
rs977339055 Chr.12: 53045717-53045717 on Build GRCh38
rs796286812 Chr.12: 53045580-53045580 on Build GRCh38
rs917526007 Chr.12: 53045745-53045745 on Build GRCh38
rs776724259 Chr.12: 53045738-53045738 on Build GRCh38
rs552325108 Chr.12: 53045865-53045865 on Build GRCh38
rs1014680159 Chr.12: 53045443-53045443 on Build GRCh38
rs918232649 Chr.12: 53045659-53045659 on Build GRCh38
rs989935062 Chr.12: 53045696-53045696 on Build GRCh38
rs115939966 Chr.12: 53045750-53045750 on Build GRCh38
rs1003694686 Chr.12: 53045595-53045595 on Build GRCh38
rs939653683 Chr.12: 53045875-53045875 on Build GRCh38
rs903309137 Chr.12: 53045471-53045471 on Build GRCh38
rs960100 Chr.12: 53045753-53045753 on Build GRCh38
rs188004633 Chr.12: 53045686-53045686 on Build GRCh38
rs537201748 Chr.12: 53045563-53045563 on Build GRCh38
rs1048615886 Chr.12: 53045827-53045827 on Build GRCh38
rs5798233 Chr.12: 53045573-53045573 on Build GRCh38
rs1041615611 Chr.12: 53045457-53045457 on Build GRCh38
rs1055936539 Chr.12: 53045738-53045738 on Build GRCh38
rs924483275 Chr.12: 53045480-53045480 on Build GRCh38
rs896033125 Chr.12: 53045416-53045416 on Build GRCh38
rs965316409 Chr.12: 53045614-53045614 on Build GRCh38
rs777262606 Chr.12: 53045515-53045515 on Build GRCh38
rs888146361 Chr.12: 53045828-53045828 on Build GRCh38
rs1022294577 Chr.12: 53045715-53045715 on Build GRCh38
rs768051657 Chr.12: 53045716-53045716 on Build GRCh38
rs10747657 Chr.12: 53045825-53045825 on Build GRCh38
rs1020277941 Chr.12: 53045451-53045451 on Build GRCh38
rs1004506873 Chr.12: 53045579-53045579 on Build GRCh38
rs577033191 Chr.12: 53045596-53045596 on Build GRCh38
rs754081418 Chr.12: 53045816-53045817 on Build GRCh38
rs12822916 Chr.12: 53045610-53045610 on Build GRCh38
rs576207109 Chr.12: 53045549-53045549 on Build GRCh38
rs545146517 Chr.12: 53045638-53045638 on Build GRCh38
rs976862556 Chr.12: 53045619-53045619 on Build GRCh38
rs558798595 Chr.12: 53045571-53045571 on Build GRCh38
rs563403822 Chr.12: 53045806-53045806 on Build GRCh38
rs1034237288 Chr.12: 53045653-53045653 on Build GRCh38
rs932364380 Chr.12: 53045554-53045554 on Build GRCh38
rs34890457 Chr.12: 53045552-53045552 on Build GRCh38
rs957873813 Chr.12: 53045564-53045564 on Build GRCh38
rs778970617 Chr.12: 53045658-53045658 on Build GRCh38
rs574483661 Chr.12: 53045547-53045547 on Build GRCh38
rs951164955 Chr.12: 53045680-53045680 on Build GRCh38
rs1402396 Chr.12: 53045756-53045756 on Build GRCh38

View Full Product Details