Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86490747-86491257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FENDRR
Entrez Gene ID
400550
Gene Name
FOXF1 adjacent non-coding developmental regulatory RNA
Gene Aliases
FOXF1-AS1, TCONS_00024240, lincFOXF1, onco-lncRNA-21
UniGene
Hs.448825
Gene Chromosome Location
Chr.16: 86474525-86508860 on Build GRCh38
Transcripts
NR_033925.1 NR_036444.1

SNP Detail

SNP ID Chromosome Location
rs561949168 Chr.16: 86490776-86490776 on Build GRCh38
rs868268927 Chr.16: 86491068-86491068 on Build GRCh38
rs534461691 Chr.16: 86491084-86491084 on Build GRCh38
rs907081763 Chr.16: 86491151-86491151 on Build GRCh38
rs1051333297 Chr.16: 86490853-86490853 on Build GRCh38
rs983909694 Chr.16: 86491110-86491110 on Build GRCh38
rs552016972 Chr.16: 86491010-86491010 on Build GRCh38
rs556956676 Chr.16: 86490794-86490794 on Build GRCh38
rs918701717 Chr.16: 86491035-86491035 on Build GRCh38
rs193174226 Chr.16: 86490874-86490874 on Build GRCh38
rs561181616 Chr.16: 86490929-86490929 on Build GRCh38
rs547372721 Chr.16: 86491144-86491144 on Build GRCh38
rs148675716 Chr.16: 86491135-86491135 on Build GRCh38
rs1048548131 Chr.16: 86490938-86490938 on Build GRCh38
rs947737526 Chr.16: 86491161-86491161 on Build GRCh38
rs918761465 Chr.16: 86490823-86490823 on Build GRCh38
rs1022737967 Chr.16: 86491027-86491027 on Build GRCh38
rs868412389 Chr.16: 86490917-86490917 on Build GRCh38
rs149485246 Chr.16: 86490924-86490924 on Build GRCh38
rs889140428 Chr.16: 86491029-86491029 on Build GRCh38
rs968620858 Chr.16: 86490818-86490818 on Build GRCh38
rs1022263964 Chr.16: 86490778-86490778 on Build GRCh38
rs536639034 Chr.16: 86490791-86490791 on Build GRCh38
rs1007506431 Chr.16: 86491032-86491032 on Build GRCh38
rs985322647 Chr.16: 86490824-86490824 on Build GRCh38
rs773377226 Chr.16: 86491130-86491130 on Build GRCh38
rs39524 Chr.16: 86490942-86490942 on Build GRCh38
rs1045509407 Chr.16: 86491145-86491145 on Build GRCh38
rs906777181 Chr.16: 86490956-86490956 on Build GRCh38
rs891523903 Chr.16: 86491107-86491107 on Build GRCh38
rs1054141950 Chr.16: 86491090-86491090 on Build GRCh38
rs372042523 Chr.16: 86491082-86491082 on Build GRCh38
rs143923722 Chr.16: 86491085-86491085 on Build GRCh38
rs1053049163 Chr.16: 86491185-86491186 on Build GRCh38
rs941422041 Chr.16: 86490804-86490804 on Build GRCh38
rs1045371266 Chr.16: 86490952-86490952 on Build GRCh38
rs771826649 Chr.16: 86491220-86491220 on Build GRCh38
rs1034704746 Chr.16: 86491179-86491179 on Build GRCh38
rs951222556 Chr.16: 86491100-86491100 on Build GRCh38
rs1010964712 Chr.16: 86491022-86491022 on Build GRCh38
rs537183164 Chr.16: 86491152-86491152 on Build GRCh38
rs1009996580 Chr.16: 86490945-86490945 on Build GRCh38
rs748588891 Chr.16: 86491083-86491083 on Build GRCh38
rs541654754 Chr.16: 86490916-86490916 on Build GRCh38
rs769269985 Chr.16: 86490957-86490957 on Build GRCh38
rs921610754 Chr.16: 86491086-86491086 on Build GRCh38
rs974166316 Chr.16: 86491019-86491019 on Build GRCh38
rs930150264 Chr.16: 86490831-86490831 on Build GRCh38
rs184593343 Chr.16: 86491088-86491088 on Build GRCh38
rs956810186 Chr.16: 86490774-86490774 on Build GRCh38
rs962728300 Chr.16: 86491191-86491191 on Build GRCh38
rs951453535 Chr.16: 86491053-86491053 on Build GRCh38
rs563738682 Chr.16: 86490997-86490997 on Build GRCh38
rs189040188 Chr.16: 86490859-86490859 on Build GRCh38
rs986940760 Chr.16: 86491078-86491078 on Build GRCh38
rs572642967 Chr.16: 86490905-86490905 on Build GRCh38
rs1001868774 Chr.16: 86490992-86490992 on Build GRCh38
rs1038658651 Chr.16: 86490913-86490913 on Build GRCh38
rs184272041 Chr.16: 86490830-86490830 on Build GRCh38
rs1003753128 Chr.16: 86490964-86490964 on Build GRCh38
rs549899450 Chr.16: 86490971-86490971 on Build GRCh38
rs939587862 Chr.16: 86491185-86491185 on Build GRCh38
rs995882118 Chr.16: 86491227-86491227 on Build GRCh38
rs9925614 Chr.16: 86491005-86491005 on Build GRCh38
rs974598526 Chr.16: 86490805-86490805 on Build GRCh38
rs892613778 Chr.16: 86491217-86491217 on Build GRCh38
rs576645636 Chr.16: 86490816-86490816 on Build GRCh38
rs980431149 Chr.16: 86491165-86491165 on Build GRCh38
rs965216746 Chr.16: 86490894-86490894 on Build GRCh38

View Full Product Details