Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55988804-55989329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105374690
Entrez Gene ID
105374690
Gene Name
uncharacterized LOC105374690
Gene Aliases
-
UniGene
Hs.711066
Gene Chromosome Location
Chr.2: 55945824-56177554 on Build GRCh38
Transcripts
NR_029629.1 NR_126406.1

Gene Symbol
MIR216A
Entrez Gene ID
406998
Gene Name
microRNA 216a
Gene Aliases
MIR216, MIRN216, MIRN216A, miRNA216, mir-216, mir-216a
UniGene
Gene Chromosome Location
Chr.2: 55988950-55989059 on Build GRCh38
Transcripts
NR_029629.1 NR_126406.1

Gene Symbol
MIR217HG
Entrez Gene ID
104355290
Gene Name
MIR217 host gene
Gene Aliases
MIR216AHG, MIR216BHG
UniGene
Hs.580331
Gene Chromosome Location
Chr.2: 55963406-56047326 on Build GRCh38
Transcripts
NR_029629.1 NR_126406.1

SNP Detail

SNP ID Chromosome Location
rs746116536 Chr.2: 55988952-55988952 on Build GRCh38
rs572237125 Chr.2: 55989132-55989132 on Build GRCh38
rs754938364 Chr.2: 55988970-55988971 on Build GRCh38
rs995427902 Chr.2: 55988835-55988835 on Build GRCh38
rs764076724 Chr.2: 55988968-55988968 on Build GRCh38
rs982659904 Chr.2: 55988889-55988889 on Build GRCh38
rs757094946 Chr.2: 55989040-55989040 on Build GRCh38
rs982309451 Chr.2: 55989210-55989210 on Build GRCh38
rs756437766 Chr.2: 55989005-55989005 on Build GRCh38
rs1052694334 Chr.2: 55988930-55988930 on Build GRCh38
rs781164963 Chr.2: 55989043-55989043 on Build GRCh38
rs770767645 Chr.2: 55988946-55988946 on Build GRCh38
rs570183235 Chr.2: 55988943-55988943 on Build GRCh38
rs988822429 Chr.2: 55989299-55989299 on Build GRCh38
rs11903947 Chr.2: 55989185-55989185 on Build GRCh38
rs750993938 Chr.2: 55989097-55989097 on Build GRCh38
rs948231821 Chr.2: 55988887-55988887 on Build GRCh38
rs190353342 Chr.2: 55988882-55988882 on Build GRCh38
rs560092808 Chr.2: 55989007-55989007 on Build GRCh38
rs370221981 Chr.2: 55989021-55989021 on Build GRCh38
rs763682473 Chr.2: 55989095-55989095 on Build GRCh38
rs746980700 Chr.2: 55989034-55989034 on Build GRCh38
rs72813873 Chr.2: 55989231-55989231 on Build GRCh38
rs752336989 Chr.2: 55989011-55989011 on Build GRCh38
rs916650420 Chr.2: 55988885-55988885 on Build GRCh38
rs545368179 Chr.2: 55989068-55989068 on Build GRCh38
rs769696880 Chr.2: 55989044-55989044 on Build GRCh38
rs1006759519 Chr.2: 55988855-55988855 on Build GRCh38
rs896825223 Chr.2: 55989154-55989154 on Build GRCh38
rs890388794 Chr.2: 55989136-55989136 on Build GRCh38
rs10865292 Chr.2: 55988917-55988917 on Build GRCh38
rs565181293 Chr.2: 55989100-55989100 on Build GRCh38
rs144312289 Chr.2: 55989164-55989164 on Build GRCh38
rs541243229 Chr.2: 55989157-55989157 on Build GRCh38
rs756798139 Chr.2: 55989098-55989098 on Build GRCh38
rs973524508 Chr.2: 55988883-55988883 on Build GRCh38
rs983476238 Chr.2: 55988879-55988879 on Build GRCh38
rs754572592 Chr.2: 55988905-55988905 on Build GRCh38
rs386646346 Chr.2: 55989184-55989185 on Build GRCh38
rs750380916 Chr.2: 55989101-55989101 on Build GRCh38
rs752402165 Chr.2: 55989080-55989080 on Build GRCh38
rs1021496070 Chr.2: 55989275-55989275 on Build GRCh38
rs897043839 Chr.2: 55988833-55988833 on Build GRCh38
rs13431228 Chr.2: 55989184-55989184 on Build GRCh38
rs1015307723 Chr.2: 55988881-55988881 on Build GRCh38
rs1036716080 Chr.2: 55988830-55988834 on Build GRCh38
rs1030537668 Chr.2: 55989123-55989123 on Build GRCh38
rs41291179 Chr.2: 55988955-55988955 on Build GRCh38
rs371648331 Chr.2: 55989099-55989099 on Build GRCh38
rs531603794 Chr.2: 55989226-55989226 on Build GRCh38
rs763149314 Chr.2: 55988967-55988967 on Build GRCh38
rs373410727 Chr.2: 55989058-55989058 on Build GRCh38
rs573700043 Chr.2: 55988957-55988957 on Build GRCh38
rs879173809 Chr.2: 55988869-55988869 on Build GRCh38
rs757996659 Chr.2: 55989017-55989017 on Build GRCh38
rs752522105 Chr.2: 55989059-55989060 on Build GRCh38
rs774925849 Chr.2: 55988980-55988980 on Build GRCh38
rs762165228 Chr.2: 55988995-55988995 on Build GRCh38
rs751357434 Chr.2: 55988967-55988967 on Build GRCh38
rs781196304 Chr.2: 55988975-55988975 on Build GRCh38
rs1018088430 Chr.2: 55988880-55988880 on Build GRCh38
rs915478947 Chr.2: 55989301-55989301 on Build GRCh38
rs1037068110 Chr.2: 55989048-55989048 on Build GRCh38
rs569193818 Chr.2: 55988859-55988859 on Build GRCh38
rs1005245076 Chr.2: 55989149-55989149 on Build GRCh38
rs776511988 Chr.2: 55988951-55988951 on Build GRCh38
rs377721752 Chr.2: 55989088-55989088 on Build GRCh38
rs771812420 Chr.2: 55988854-55988854 on Build GRCh38
rs187624573 Chr.2: 55988832-55988832 on Build GRCh38
rs534411351 Chr.2: 55988926-55988926 on Build GRCh38
rs900366098 Chr.2: 55988830-55988830 on Build GRCh38
rs538189728 Chr.2: 55988875-55988875 on Build GRCh38
rs956891155 Chr.2: 55989240-55989240 on Build GRCh38
rs749402624 Chr.2: 55989045-55989045 on Build GRCh38
rs757828256 Chr.2: 55988925-55988925 on Build GRCh38
rs928256585 Chr.2: 55988906-55988906 on Build GRCh38
rs556227257 Chr.2: 55989052-55989052 on Build GRCh38
rs746920214 Chr.2: 55988928-55988928 on Build GRCh38
rs778423695 Chr.2: 55988908-55988908 on Build GRCh38
rs770065893 Chr.2: 55988844-55988844 on Build GRCh38
rs549386947 Chr.2: 55988852-55988852 on Build GRCh38
rs772604872 Chr.2: 55989060-55989060 on Build GRCh38
rs576393375 Chr.2: 55989067-55989067 on Build GRCh38
rs182185349 Chr.2: 55989163-55989163 on Build GRCh38
rs767887445 Chr.2: 55989004-55989004 on Build GRCh38
rs766326012 Chr.2: 55988943-55988944 on Build GRCh38

View Full Product Details