Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219293849-219294340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR153-1
Entrez Gene ID
406944
Gene Name
microRNA 153-1
Gene Aliases
MIRN153-1, mir-153-1
UniGene
Gene Chromosome Location
Chr.2: 219294111-219294200 on Build GRCh38
Transcripts
NR_029688.1 NM_001199763.1 NM_001199764.1 NM_002846.3 XM_011511566.1 XM_017004609.1 XM_017004610.1 XM_017004611.1 XM_017004612.1

Gene Symbol
PTPRN
Entrez Gene ID
5798
Gene Name
protein tyrosine phosphatase, receptor type N
Gene Aliases
IA-2, IA-2/PTP, IA2, ICA512, R-PTP-N
UniGene
Hs.89655
Gene Chromosome Location
Chr.2: 219289623-219309573 on Build GRCh38
Transcripts
NR_029688.1 NM_001199763.1 NM_001199764.1 NM_002846.3 XM_011511566.1 XM_017004609.1 XM_017004610.1 XM_017004611.1 XM_017004612.1

SNP Detail

SNP ID Chromosome Location
rs750226581 Chr.2: 219294228-219294228 on Build GRCh38
rs893227338 Chr.2: 219293982-219293982 on Build GRCh38
rs773943788 Chr.2: 219294124-219294124 on Build GRCh38
rs968985024 Chr.2: 219294235-219294235 on Build GRCh38
rs778765983 Chr.2: 219294316-219294316 on Build GRCh38
rs779822632 Chr.2: 219294090-219294090 on Build GRCh38
rs556414742 Chr.2: 219293890-219293890 on Build GRCh38
rs895143371 Chr.2: 219294149-219294149 on Build GRCh38
rs1046384022 Chr.2: 219293917-219293917 on Build GRCh38
rs376216262 Chr.2: 219294128-219294128 on Build GRCh38
rs72955491 Chr.2: 219293986-219293986 on Build GRCh38
rs1016028869 Chr.2: 219294064-219294064 on Build GRCh38
rs766650380 Chr.2: 219294186-219294186 on Build GRCh38
rs765303493 Chr.2: 219294202-219294202 on Build GRCh38
rs772305631 Chr.2: 219294058-219294058 on Build GRCh38
rs757164435 Chr.2: 219294208-219294208 on Build GRCh38
rs1023256548 Chr.2: 219293871-219293871 on Build GRCh38
rs529123538 Chr.2: 219294070-219294070 on Build GRCh38
rs914714720 Chr.2: 219294266-219294266 on Build GRCh38
rs560356020 Chr.2: 219294024-219294024 on Build GRCh38
rs932357733 Chr.2: 219293997-219293997 on Build GRCh38
rs779912650 Chr.2: 219294243-219294243 on Build GRCh38
rs917603942 Chr.2: 219294123-219294123 on Build GRCh38
rs979112652 Chr.2: 219293900-219293900 on Build GRCh38
rs576705384 Chr.2: 219293902-219293902 on Build GRCh38
rs557782621 Chr.2: 219293944-219293944 on Build GRCh38
rs950356606 Chr.2: 219293872-219293872 on Build GRCh38
rs1041753149 Chr.2: 219293993-219293993 on Build GRCh38
rs577620707 Chr.2: 219293978-219293978 on Build GRCh38
rs768175814 Chr.2: 219294202-219294203 on Build GRCh38
rs776224848 Chr.2: 219294211-219294212 on Build GRCh38
rs922295682 Chr.2: 219294315-219294315 on Build GRCh38
rs991347326 Chr.2: 219293869-219293869 on Build GRCh38
rs769382937 Chr.2: 219294089-219294089 on Build GRCh38
rs748876466 Chr.2: 219294103-219294103 on Build GRCh38
rs190595141 Chr.2: 219293916-219293916 on Build GRCh38
rs970794675 Chr.2: 219294187-219294187 on Build GRCh38
rs145546871 Chr.2: 219294312-219294312 on Build GRCh38
rs968580848 Chr.2: 219293899-219293899 on Build GRCh38
rs777742264 Chr.2: 219294068-219294068 on Build GRCh38
rs762232906 Chr.2: 219294147-219294147 on Build GRCh38
rs373271364 Chr.2: 219294081-219294081 on Build GRCh38
rs773474391 Chr.2: 219294164-219294164 on Build GRCh38
rs746255454 Chr.2: 219294020-219294020 on Build GRCh38
rs777525063 Chr.2: 219293938-219293938 on Build GRCh38
rs937496602 Chr.2: 219293963-219293963 on Build GRCh38
rs901823356 Chr.2: 219294010-219294010 on Build GRCh38
rs1034944138 Chr.2: 219294145-219294145 on Build GRCh38
rs1031613754 Chr.2: 219294262-219294262 on Build GRCh38
rs531678968 Chr.2: 219294302-219294302 on Build GRCh38
rs976393505 Chr.2: 219293948-219293948 on Build GRCh38
rs562760902 Chr.2: 219294247-219294247 on Build GRCh38
rs767466844 Chr.2: 219294219-219294219 on Build GRCh38
rs752731776 Chr.2: 219294203-219294203 on Build GRCh38
rs542872066 Chr.2: 219294078-219294078 on Build GRCh38
rs761066898 Chr.2: 219294172-219294172 on Build GRCh38
rs370258207 Chr.2: 219294232-219294232 on Build GRCh38
rs981749809 Chr.2: 219294230-219294230 on Build GRCh38
rs201703216 Chr.2: 219294116-219294116 on Build GRCh38
rs35641664 Chr.2: 219294210-219294211 on Build GRCh38
rs745910267 Chr.2: 219293964-219293965 on Build GRCh38
rs772596083 Chr.2: 219294163-219294163 on Build GRCh38
rs776153428 Chr.2: 219294207-219294207 on Build GRCh38
rs754044701 Chr.2: 219294189-219294189 on Build GRCh38
rs761529158 Chr.2: 219294213-219294214 on Build GRCh38
rs953288274 Chr.2: 219294027-219294027 on Build GRCh38
rs759693204 Chr.2: 219294190-219294190 on Build GRCh38
rs768241140 Chr.2: 219294040-219294040 on Build GRCh38

View Full Product Details