Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231455657-231456204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCL
Entrez Gene ID
4691
Gene Name
nucleolin
Gene Aliases
C23
UniGene
Hs.79110
Gene Chromosome Location
Chr.2: 231454748-231464494 on Build GRCh38
Transcripts
NM_005381.2 NR_002921.1

Gene Symbol
SNORA75
Entrez Gene ID
654321
Gene Name
small nucleolar RNA, H/ACA box 75
Gene Aliases
SNORA75A, U23
UniGene
Hs.674388
Gene Chromosome Location
Chr.2: 231455800-231455936 on Build GRCh38
Transcripts
NM_005381.2 NR_002921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3909824 c.1713C>T p.S571S Chr.2: 231456129-231456129 on Build GRCh38
5973652 c.1786G>A p.V596I Chr.2: 231456056-231456056 on Build GRCh38
3579021 c.1712C>T p.S571F Chr.2: 231456130-231456130 on Build GRCh38
1482857 c.1789C>T p.R597W Chr.2: 231456053-231456053 on Build GRCh38
327970 c.1706-10_1706-9insT p.? Chr.2: 231456145-231456146 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759439274 Chr.2: 231455827-231455828 on Build GRCh38
rs771632006 Chr.2: 231455887-231455887 on Build GRCh38
rs752108662 Chr.2: 231455763-231455763 on Build GRCh38
rs764203421 Chr.2: 231455870-231455873 on Build GRCh38
rs752984476 Chr.2: 231455753-231455753 on Build GRCh38
rs142532822 Chr.2: 231456108-231456108 on Build GRCh38
rs751632928 Chr.2: 231455825-231455825 on Build GRCh38
rs915949999 Chr.2: 231455760-231455760 on Build GRCh38
rs763333783 Chr.2: 231455822-231455822 on Build GRCh38
rs758751983 Chr.2: 231455905-231455906 on Build GRCh38
rs896859104 Chr.2: 231455955-231455955 on Build GRCh38
rs779527019 Chr.2: 231455788-231455788 on Build GRCh38
rs766300858 Chr.2: 231455910-231455913 on Build GRCh38
rs747845005 Chr.2: 231455971-231455971 on Build GRCh38
rs746154150 Chr.2: 231455779-231455779 on Build GRCh38
rs761322233 Chr.2: 231455804-231455804 on Build GRCh38
rs1015228303 Chr.2: 231455732-231455732 on Build GRCh38
rs368724845 Chr.2: 231456159-231456159 on Build GRCh38
rs919219827 Chr.2: 231456141-231456141 on Build GRCh38
rs546001499 Chr.2: 231456000-231456000 on Build GRCh38
rs764346257 Chr.2: 231455907-231455907 on Build GRCh38
rs938941781 Chr.2: 231455714-231455714 on Build GRCh38
rs765628776 Chr.2: 231456075-231456075 on Build GRCh38
rs189865329 Chr.2: 231456063-231456063 on Build GRCh38
rs766823824 Chr.2: 231456158-231456158 on Build GRCh38
rs566862419 Chr.2: 231455728-231455728 on Build GRCh38
rs1012663136 Chr.2: 231455707-231455707 on Build GRCh38
rs757814072 Chr.2: 231455972-231455972 on Build GRCh38
rs773333006 Chr.2: 231456168-231456168 on Build GRCh38
rs1056040169 Chr.2: 231455740-231455740 on Build GRCh38
rs915275552 Chr.2: 231455815-231455815 on Build GRCh38
rs913641534 Chr.2: 231455687-231455687 on Build GRCh38
rs767494004 Chr.2: 231456004-231456004 on Build GRCh38
rs112840566 Chr.2: 231456137-231456137 on Build GRCh38
rs777407354 Chr.2: 231455975-231455975 on Build GRCh38
rs77122043 Chr.2: 231455708-231455708 on Build GRCh38
rs768608724 Chr.2: 231455929-231455929 on Build GRCh38
rs769812342 Chr.2: 231455792-231455792 on Build GRCh38
rs762680715 Chr.2: 231455751-231455752 on Build GRCh38
rs760336473 Chr.2: 231455812-231455812 on Build GRCh38
rs941990760 Chr.2: 231455765-231455765 on Build GRCh38
rs769684387 Chr.2: 231456155-231456155 on Build GRCh38
rs752089107 Chr.2: 231455914-231455917 on Build GRCh38
rs374299439 Chr.2: 231456035-231456035 on Build GRCh38
rs767687708 Chr.2: 231455926-231455926 on Build GRCh38
rs59377947 Chr.2: 231456154-231456155 on Build GRCh38
rs537965181 Chr.2: 231455877-231455877 on Build GRCh38
rs774633476 Chr.2: 231455981-231455981 on Build GRCh38
rs34531744 Chr.2: 231455762-231455763 on Build GRCh38
rs766828166 Chr.2: 231456056-231456056 on Build GRCh38
rs530369670 Chr.2: 231455704-231455704 on Build GRCh38
rs16828079 Chr.2: 231456087-231456087 on Build GRCh38
rs201768710 Chr.2: 231455766-231455766 on Build GRCh38
rs2595518 Chr.2: 231456171-231456171 on Build GRCh38
rs746907631 Chr.2: 231455978-231455978 on Build GRCh38
rs769308610 Chr.2: 231456146-231456146 on Build GRCh38
rs760778155 Chr.2: 231456124-231456124 on Build GRCh38
rs755477918 Chr.2: 231455994-231455994 on Build GRCh38
rs149759354 Chr.2: 231456078-231456078 on Build GRCh38
rs113883398 Chr.2: 231456083-231456083 on Build GRCh38
rs868722182 Chr.2: 231456058-231456058 on Build GRCh38
rs570584382 Chr.2: 231455875-231455875 on Build GRCh38
rs372020177 Chr.2: 231455939-231455939 on Build GRCh38
rs771835751 Chr.2: 231455810-231455810 on Build GRCh38
rs775495867 Chr.2: 231456156-231456156 on Build GRCh38
rs774294773 Chr.2: 231455794-231455795 on Build GRCh38
rs200830899 Chr.2: 231455884-231455884 on Build GRCh38
rs888206414 Chr.2: 231455711-231455711 on Build GRCh38
rs748992346 Chr.2: 231455790-231455790 on Build GRCh38
rs137866827 Chr.2: 231455703-231455703 on Build GRCh38
rs776625710 Chr.2: 231455892-231455892 on Build GRCh38
rs558008297 Chr.2: 231456025-231456025 on Build GRCh38
rs978473237 Chr.2: 231455860-231455860 on Build GRCh38
rs769736556 Chr.2: 231455994-231455994 on Build GRCh38
rs939572111 Chr.2: 231455878-231455878 on Build GRCh38
rs752936387 Chr.2: 231455817-231455817 on Build GRCh38
rs778039098 Chr.2: 231455754-231455754 on Build GRCh38
rs781513272 Chr.2: 231455831-231455831 on Build GRCh38
rs200390674 Chr.2: 231456144-231456144 on Build GRCh38
rs770626102 Chr.2: 231455891-231455891 on Build GRCh38
rs954053435 Chr.2: 231455786-231455786 on Build GRCh38
rs765495403 Chr.2: 231455881-231455883 on Build GRCh38
rs572406712 Chr.2: 231455961-231455961 on Build GRCh38
rs747614452 Chr.2: 231455885-231455885 on Build GRCh38
rs756414993 Chr.2: 231455871-231455871 on Build GRCh38
rs770227954 Chr.2: 231455781-231455781 on Build GRCh38
rs966866747 Chr.2: 231455950-231455950 on Build GRCh38
rs748310037 Chr.2: 231456157-231456157 on Build GRCh38
rs553773439 Chr.2: 231455946-231455946 on Build GRCh38
rs757364315 Chr.2: 231455827-231455827 on Build GRCh38
rs756859655 Chr.2: 231456130-231456130 on Build GRCh38
rs554450852 Chr.2: 231455876-231455877 on Build GRCh38
rs185290422 Chr.2: 231455909-231455909 on Build GRCh38
rs761752440 Chr.2: 231455923-231455923 on Build GRCh38
rs768128437 Chr.2: 231455795-231455795 on Build GRCh38
rs773904559 Chr.2: 231455800-231455800 on Build GRCh38
rs745852480 Chr.2: 231456149-231456149 on Build GRCh38
rs1005321165 Chr.2: 231455927-231455927 on Build GRCh38
rs13019380 Chr.2: 231455870-231455870 on Build GRCh38
rs766488719 Chr.2: 231456180-231456180 on Build GRCh38
rs545919038 Chr.2: 231455874-231455876 on Build GRCh38
rs16828077 Chr.2: 231455699-231455699 on Build GRCh38
rs768236698 Chr.2: 231455883-231455883 on Build GRCh38
rs370933084 Chr.2: 231455985-231455985 on Build GRCh38
rs770660707 Chr.2: 231455782-231455783 on Build GRCh38
rs759419449 Chr.2: 231455893-231455893 on Build GRCh38
rs760771676 Chr.2: 231456179-231456179 on Build GRCh38
rs770844868 Chr.2: 231455980-231455980 on Build GRCh38
rs527862159 Chr.2: 231455814-231455814 on Build GRCh38
rs923932492 Chr.2: 231456161-231456161 on Build GRCh38
rs756715384 Chr.2: 231455932-231455932 on Build GRCh38
rs960229822 Chr.2: 231455990-231455990 on Build GRCh38
rs750681418 Chr.2: 231455897-231455898 on Build GRCh38
rs375337610 Chr.2: 231456143-231456143 on Build GRCh38
rs369902237 Chr.2: 231456145-231456145 on Build GRCh38
rs760671544 Chr.2: 231456033-231456033 on Build GRCh38
rs751237457 Chr.2: 231455692-231455692 on Build GRCh38
rs780508751 Chr.2: 231455725-231455725 on Build GRCh38
rs1029443173 Chr.2: 231455777-231455777 on Build GRCh38
rs777303754 Chr.2: 231456101-231456101 on Build GRCh38
rs764256235 Chr.2: 231455824-231455824 on Build GRCh38
rs961257991 Chr.2: 231455741-231455741 on Build GRCh38
rs755364611 Chr.2: 231455953-231455953 on Build GRCh38
rs560441285 Chr.2: 231455695-231455695 on Build GRCh38
rs985667401 Chr.2: 231455995-231455995 on Build GRCh38
rs150234462 Chr.2: 231456145-231456146 on Build GRCh38
rs774695825 Chr.2: 231455996-231455996 on Build GRCh38
rs567099750 Chr.2: 231455888-231455888 on Build GRCh38
rs139786193 Chr.2: 231456057-231456057 on Build GRCh38

View Full Product Details