Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18380787-18381334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00851
Entrez Gene ID
440757
Gene Name
long intergenic non-protein coding RNA 851
Gene Aliases
DZANK1-AS1
UniGene
Hs.120399
Gene Chromosome Location
Chr.20: 18379049-18381484 on Build GRCh38
Transcripts
NR_034167.1

SNP Detail

SNP ID Chromosome Location
rs923365942 Chr.20: 18380834-18380834 on Build GRCh38
rs968127811 Chr.20: 18380974-18380974 on Build GRCh38
rs1009568126 Chr.20: 18380960-18380960 on Build GRCh38
rs750751192 Chr.20: 18381171-18381171 on Build GRCh38
rs1024703713 Chr.20: 18381059-18381060 on Build GRCh38
rs769942234 Chr.20: 18380950-18380950 on Build GRCh38
rs1051384171 Chr.20: 18381228-18381228 on Build GRCh38
rs941093290 Chr.20: 18381170-18381170 on Build GRCh38
rs892097097 Chr.20: 18380992-18380992 on Build GRCh38
rs958629932 Chr.20: 18381037-18381037 on Build GRCh38
rs765899955 Chr.20: 18381172-18381172 on Build GRCh38
rs1006140113 Chr.20: 18380838-18380838 on Build GRCh38
rs917147114 Chr.20: 18381105-18381105 on Build GRCh38
rs1013673037 Chr.20: 18381007-18381007 on Build GRCh38
rs777555864 Chr.20: 18381292-18381292 on Build GRCh38
rs778467570 Chr.20: 18381176-18381176 on Build GRCh38
rs552197684 Chr.20: 18381090-18381090 on Build GRCh38
rs1022257499 Chr.20: 18381239-18381239 on Build GRCh38
rs892714392 Chr.20: 18381230-18381230 on Build GRCh38
rs569167045 Chr.20: 18381298-18381298 on Build GRCh38
rs10485580 Chr.20: 18380991-18380991 on Build GRCh38
rs759539955 Chr.20: 18381154-18381156 on Build GRCh38
rs1022836328 Chr.20: 18380968-18380968 on Build GRCh38
rs934129009 Chr.20: 18381196-18381196 on Build GRCh38
rs923301452 Chr.20: 18381229-18381229 on Build GRCh38
rs1001977412 Chr.20: 18381071-18381071 on Build GRCh38
rs764201766 Chr.20: 18380990-18380990 on Build GRCh38
rs550993136 Chr.20: 18381233-18381233 on Build GRCh38
rs753424046 Chr.20: 18381173-18381173 on Build GRCh38
rs985025690 Chr.20: 18381140-18381140 on Build GRCh38
rs566065041 Chr.20: 18380826-18380826 on Build GRCh38
rs927125425 Chr.20: 18381012-18381012 on Build GRCh38
rs903955346 Chr.20: 18381274-18381274 on Build GRCh38
rs929685893 Chr.20: 18381108-18381108 on Build GRCh38
rs969139502 Chr.20: 18381059-18381059 on Build GRCh38
rs982433107 Chr.20: 18381110-18381110 on Build GRCh38
rs936517612 Chr.20: 18381294-18381294 on Build GRCh38
rs190492412 Chr.20: 18380876-18380876 on Build GRCh38
rs1034169889 Chr.20: 18381311-18381311 on Build GRCh38
rs539885244 Chr.20: 18380952-18380952 on Build GRCh38
rs776532789 Chr.20: 18380965-18380965 on Build GRCh38
rs925111662 Chr.20: 18381279-18381279 on Build GRCh38
rs1883938 Chr.20: 18380959-18380959 on Build GRCh38
rs992734844 Chr.20: 18381089-18381089 on Build GRCh38
rs764700644 Chr.20: 18381225-18381226 on Build GRCh38
rs149171153 Chr.20: 18381168-18381169 on Build GRCh38
rs748096263 Chr.20: 18381201-18381201 on Build GRCh38
rs754491297 Chr.20: 18381174-18381174 on Build GRCh38
rs570787918 Chr.20: 18381085-18381085 on Build GRCh38
rs765239050 Chr.20: 18381304-18381304 on Build GRCh38
rs900099599 Chr.20: 18380859-18380859 on Build GRCh38
rs537746715 Chr.20: 18381116-18381116 on Build GRCh38
rs762901191 Chr.20: 18380979-18380979 on Build GRCh38
rs760379664 Chr.20: 18380861-18380861 on Build GRCh38
rs145126752 Chr.20: 18380839-18380839 on Build GRCh38
rs1009579774 Chr.20: 18381234-18381234 on Build GRCh38
rs568998921 Chr.20: 18381144-18381144 on Build GRCh38
rs776533814 Chr.20: 18381104-18381105 on Build GRCh38

View Full Product Details