Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 22561155-22561690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00261
Entrez Gene ID
140828
Gene Name
long intergenic non-protein coding RNA 261
Gene Aliases
ALIEN, C20orf56, DEANR1, HCCDR1, NCRNA00261, onco-lncRNA-17
UniGene
Hs.9842
Gene Chromosome Location
Chr.20: 22560553-22578642 on Build GRCh38
Transcripts
NR_001558.3

SNP Detail

SNP ID Chromosome Location
rs1045405701 Chr.20: 22561254-22561254 on Build GRCh38
rs184899379 Chr.20: 22561343-22561343 on Build GRCh38
rs537937617 Chr.20: 22561192-22561192 on Build GRCh38
rs918841291 Chr.20: 22561245-22561245 on Build GRCh38
rs948490220 Chr.20: 22561231-22561231 on Build GRCh38
rs997758594 Chr.20: 22561313-22561313 on Build GRCh38
rs1046668933 Chr.20: 22561229-22561229 on Build GRCh38
rs560885725 Chr.20: 22561274-22561274 on Build GRCh38
rs948437907 Chr.20: 22561573-22561573 on Build GRCh38
rs552692274 Chr.20: 22561456-22561456 on Build GRCh38
rs147584619 Chr.20: 22561309-22561309 on Build GRCh38
rs74520982 Chr.20: 22561645-22561645 on Build GRCh38
rs1203880 Chr.20: 22561385-22561385 on Build GRCh38
rs188359643 Chr.20: 22561395-22561395 on Build GRCh38
rs116705849 Chr.20: 22561362-22561362 on Build GRCh38
rs953445913 Chr.20: 22561374-22561374 on Build GRCh38
rs749771039 Chr.20: 22561187-22561187 on Build GRCh38
rs560292934 Chr.20: 22561644-22561644 on Build GRCh38
rs898416603 Chr.20: 22561326-22561326 on Build GRCh38
rs1019001490 Chr.20: 22561390-22561390 on Build GRCh38
rs916648124 Chr.20: 22561398-22561398 on Build GRCh38
rs114226276 Chr.20: 22561191-22561191 on Build GRCh38
rs941686439 Chr.20: 22561536-22561536 on Build GRCh38
rs999418163 Chr.20: 22561361-22561361 on Build GRCh38
rs930193350 Chr.20: 22561250-22561251 on Build GRCh38
rs970796104 Chr.20: 22561408-22561408 on Build GRCh38
rs924852133 Chr.20: 22561477-22561477 on Build GRCh38
rs887421416 Chr.20: 22561281-22561281 on Build GRCh38
rs768234755 Chr.20: 22561473-22561473 on Build GRCh38
rs1047400202 Chr.20: 22561255-22561255 on Build GRCh38
rs990329896 Chr.20: 22561513-22561513 on Build GRCh38
rs999470592 Chr.20: 22561613-22561613 on Build GRCh38
rs930247172 Chr.20: 22561459-22561459 on Build GRCh38
rs966627355 Chr.20: 22561356-22561356 on Build GRCh38
rs900254860 Chr.20: 22561580-22561580 on Build GRCh38
rs184520466 Chr.20: 22561639-22561639 on Build GRCh38
rs58788950 Chr.20: 22561589-22561589 on Build GRCh38
rs898410709 Chr.20: 22561636-22561636 on Build GRCh38
rs191618404 Chr.20: 22561221-22561221 on Build GRCh38
rs574585255 Chr.20: 22561273-22561273 on Build GRCh38
rs908908033 Chr.20: 22561504-22561504 on Build GRCh38
rs6048192 Chr.20: 22561479-22561479 on Build GRCh38
rs889590449 Chr.20: 22561653-22561653 on Build GRCh38
rs6106523 Chr.20: 22561214-22561214 on Build GRCh38
rs372339558 Chr.20: 22561344-22561344 on Build GRCh38
rs991290971 Chr.20: 22561220-22561220 on Build GRCh38
rs368166543 Chr.20: 22561222-22561222 on Build GRCh38
rs6048191 Chr.20: 22561282-22561282 on Build GRCh38
rs1203879 Chr.20: 22561178-22561178 on Build GRCh38
rs550725540 Chr.20: 22561355-22561355 on Build GRCh38
rs143130313 Chr.20: 22561590-22561590 on Build GRCh38
rs3171310 Chr.20: 22561468-22561468 on Build GRCh38
rs149070227 Chr.20: 22561551-22561551 on Build GRCh38
rs73902901 Chr.20: 22561295-22561295 on Build GRCh38
rs990951601 Chr.20: 22561393-22561393 on Build GRCh38
rs1038823240 Chr.20: 22561537-22561537 on Build GRCh38
rs555945301 Chr.20: 22561223-22561223 on Build GRCh38
rs982252573 Chr.20: 22561420-22561420 on Build GRCh38
rs540254527 Chr.20: 22561240-22561240 on Build GRCh38
rs554682676 Chr.20: 22561618-22561618 on Build GRCh38
rs540672851 Chr.20: 22561640-22561640 on Build GRCh38
rs192197719 Chr.20: 22561603-22561603 on Build GRCh38
rs138869855 Chr.20: 22561457-22561458 on Build GRCh38

View Full Product Details