Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10285372-10285884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRL
Entrez Gene ID
51738
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
MTLRP
UniGene
Hs.590080
Gene Chromosome Location
Chr.3: 10285750-10292947 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NR_026829.1

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10280952-10293449 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NR_026829.1

Gene Symbol
LINC00852
Entrez Gene ID
84657
Gene Name
long intergenic non-protein coding RNA 852
Gene Aliases
C3orf42, GHRL-AS2, GHRLOS2, NAG73
UniGene
Hs.302131
Gene Chromosome Location
Chr.3: 10284419-10285746 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NR_026829.1

SNP Detail

SNP ID Chromosome Location
rs748920776 Chr.3: 10285578-10285578 on Build GRCh38
rs551236832 Chr.3: 10285788-10285788 on Build GRCh38
rs377521983 Chr.3: 10285397-10285397 on Build GRCh38
rs1017156544 Chr.3: 10285457-10285457 on Build GRCh38
rs778484168 Chr.3: 10285677-10285677 on Build GRCh38
rs747672256 Chr.3: 10285770-10285770 on Build GRCh38
rs539219040 Chr.3: 10285580-10285580 on Build GRCh38
rs938977801 Chr.3: 10285445-10285448 on Build GRCh38
rs544063136 Chr.3: 10285525-10285525 on Build GRCh38
rs927661958 Chr.3: 10285445-10285446 on Build GRCh38
rs897781845 Chr.3: 10285486-10285486 on Build GRCh38
rs1012496553 Chr.3: 10285424-10285424 on Build GRCh38
rs966073309 Chr.3: 10285556-10285556 on Build GRCh38
rs1025803523 Chr.3: 10285742-10285742 on Build GRCh38
rs945955425 Chr.3: 10285864-10285864 on Build GRCh38
rs957644645 Chr.3: 10285657-10285657 on Build GRCh38
rs1045188766 Chr.3: 10285440-10285440 on Build GRCh38
rs756695811 Chr.3: 10285830-10285830 on Build GRCh38
rs1012625361 Chr.3: 10285714-10285714 on Build GRCh38
rs768160177 Chr.3: 10285858-10285858 on Build GRCh38
rs758555486 Chr.3: 10285804-10285804 on Build GRCh38
rs1056905525 Chr.3: 10285686-10285686 on Build GRCh38
rs1019412739 Chr.3: 10285542-10285542 on Build GRCh38
rs750221156 Chr.3: 10285794-10285794 on Build GRCh38
rs374865035 Chr.3: 10285511-10285511 on Build GRCh38
rs910887012 Chr.3: 10285569-10285569 on Build GRCh38
rs775181399 Chr.3: 10285844-10285844 on Build GRCh38
rs986570963 Chr.3: 10285805-10285805 on Build GRCh38
rs367697628 Chr.3: 10285524-10285524 on Build GRCh38
rs568932159 Chr.3: 10285828-10285828 on Build GRCh38
rs781154531 Chr.3: 10285827-10285827 on Build GRCh38
rs994744178 Chr.3: 10285495-10285495 on Build GRCh38
rs970599303 Chr.3: 10285694-10285694 on Build GRCh38
rs776146116 Chr.3: 10285863-10285863 on Build GRCh38
rs1023105281 Chr.3: 10285679-10285679 on Build GRCh38
rs930565885 Chr.3: 10285793-10285793 on Build GRCh38
rs201295883 Chr.3: 10285833-10285833 on Build GRCh38
rs771269632 Chr.3: 10285838-10285838 on Build GRCh38
rs1017210228 Chr.3: 10285719-10285719 on Build GRCh38
rs980551343 Chr.3: 10285697-10285697 on Build GRCh38
rs914515584 Chr.3: 10285847-10285847 on Build GRCh38
rs1041515028 Chr.3: 10285659-10285659 on Build GRCh38
rs547255898 Chr.3: 10285411-10285411 on Build GRCh38
rs994361172 Chr.3: 10285731-10285731 on Build GRCh38
rs112113740 Chr.3: 10285485-10285485 on Build GRCh38
rs766608356 Chr.3: 10285806-10285806 on Build GRCh38
rs962878623 Chr.3: 10285464-10285464 on Build GRCh38
rs898725190 Chr.3: 10285728-10285728 on Build GRCh38
rs867308216 Chr.3: 10285853-10285853 on Build GRCh38
rs1003621141 Chr.3: 10285681-10285681 on Build GRCh38
rs972867369 Chr.3: 10285523-10285523 on Build GRCh38
rs1036027367 Chr.3: 10285456-10285456 on Build GRCh38
rs754768988 Chr.3: 10285402-10285402 on Build GRCh38
rs955089350 Chr.3: 10285559-10285559 on Build GRCh38
rs368192363 Chr.3: 10285817-10285817 on Build GRCh38
rs200041403 Chr.3: 10285535-10285535 on Build GRCh38
rs868519650 Chr.3: 10285739-10285739 on Build GRCh38
rs768341252 Chr.3: 10285660-10285660 on Build GRCh38
rs1031622923 Chr.3: 10285641-10285641 on Build GRCh38
rs878990654 Chr.3: 10285859-10285859 on Build GRCh38
rs866782377 Chr.3: 10285583-10285583 on Build GRCh38
rs939108476 Chr.3: 10285722-10285722 on Build GRCh38
rs945904440 Chr.3: 10285639-10285639 on Build GRCh38
rs375772841 Chr.3: 10285849-10285849 on Build GRCh38
rs895148586 Chr.3: 10285701-10285701 on Build GRCh38
rs986906904 Chr.3: 10285567-10285567 on Build GRCh38
rs781563704 Chr.3: 10285574-10285574 on Build GRCh38
rs751795811 Chr.3: 10285807-10285807 on Build GRCh38
rs568146541 Chr.3: 10285609-10285610 on Build GRCh38
rs757282700 Chr.3: 10285846-10285849 on Build GRCh38
rs914463623 Chr.3: 10285633-10285633 on Build GRCh38

View Full Product Details