Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 86933233-86933768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506746
Entrez Gene ID
100506746
Gene Name
uncharacterized LOC100506746
Gene Aliases
-
UniGene
Hs.657766
Gene Chromosome Location
Chr.4: 86924894-86934850 on Build GRCh38
Transcripts
NR_038841.1

SNP Detail

SNP ID Chromosome Location
rs890425075 Chr.4: 86933444-86933444 on Build GRCh38
rs926294694 Chr.4: 86933553-86933553 on Build GRCh38
rs1031057685 Chr.4: 86933481-86933481 on Build GRCh38
rs192991947 Chr.4: 86933645-86933645 on Build GRCh38
rs375543762 Chr.4: 86933650-86933650 on Build GRCh38
rs963764819 Chr.4: 86933426-86933426 on Build GRCh38
rs761294867 Chr.4: 86933452-86933452 on Build GRCh38
rs982670951 Chr.4: 86933478-86933478 on Build GRCh38
rs1046770590 Chr.4: 86933302-86933302 on Build GRCh38
rs762247564 Chr.4: 86933599-86933600 on Build GRCh38
rs893880824 Chr.4: 86933482-86933482 on Build GRCh38
rs556143084 Chr.4: 86933619-86933619 on Build GRCh38
rs1007527007 Chr.4: 86933358-86933358 on Build GRCh38
rs776862620 Chr.4: 86933599-86933599 on Build GRCh38
rs890688345 Chr.4: 86933706-86933706 on Build GRCh38
rs772531064 Chr.4: 86933400-86933400 on Build GRCh38
rs951209267 Chr.4: 86933469-86933469 on Build GRCh38
rs932216697 Chr.4: 86933393-86933393 on Build GRCh38
rs113530269 Chr.4: 86933538-86933538 on Build GRCh38
rs528455821 Chr.4: 86933519-86933519 on Build GRCh38
rs150004501 Chr.4: 86933720-86933720 on Build GRCh38
rs914092265 Chr.4: 86933600-86933600 on Build GRCh38
rs994918879 Chr.4: 86933432-86933432 on Build GRCh38
rs1053539928 Chr.4: 86933592-86933592 on Build GRCh38
rs10595452 Chr.4: 86933266-86933269 on Build GRCh38
rs908052584 Chr.4: 86933322-86933322 on Build GRCh38
rs113224620 Chr.4: 86933265-86933268 on Build GRCh38
rs890471144 Chr.4: 86933325-86933325 on Build GRCh38
rs973604610 Chr.4: 86933737-86933737 on Build GRCh38
rs560200076 Chr.4: 86933434-86933434 on Build GRCh38
rs970383649 Chr.4: 86933537-86933537 on Build GRCh38
rs936442921 Chr.4: 86933590-86933590 on Build GRCh38
rs951055823 Chr.4: 86933633-86933633 on Build GRCh38
rs917793550 Chr.4: 86933507-86933507 on Build GRCh38
rs1037859119 Chr.4: 86933356-86933356 on Build GRCh38
rs941069069 Chr.4: 86933726-86933726 on Build GRCh38
rs773659500 Chr.4: 86933410-86933410 on Build GRCh38
rs757983247 Chr.4: 86933515-86933516 on Build GRCh38
rs942146422 Chr.4: 86933344-86933344 on Build GRCh38
rs994635079 Chr.4: 86933345-86933345 on Build GRCh38

View Full Product Details