Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172958904-172959397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100268168
Entrez Gene ID
100268168
Gene Name
uncharacterized LOC100268168
Gene Aliases
-
UniGene
Hs.519766
Gene Chromosome Location
Chr.5: 172954782-172959368 on Build GRCh38
Transcripts
NR_026682.1 NM_001317980.1 NM_001317981.1 XM_005265921.3 XM_011534565.2 XM_017009520.1

Gene Symbol
RPL26L1
Entrez Gene ID
51121
Gene Name
ribosomal protein L26 like 1
Gene Aliases
RPL26P1
UniGene
Hs.546390
Gene Chromosome Location
Chr.5: 172953677-172969771 on Build GRCh38
Transcripts
NR_026682.1 NM_001317980.1 NM_001317981.1 XM_005265921.3 XM_011534565.2 XM_017009520.1

SNP Detail

SNP ID Chromosome Location
rs947365765 Chr.5: 172959353-172959353 on Build GRCh38
rs112809806 Chr.5: 172958983-172958983 on Build GRCh38
rs941769246 Chr.5: 172959221-172959221 on Build GRCh38
rs58954040 Chr.5: 172958926-172958926 on Build GRCh38
rs1056579321 Chr.5: 172959372-172959372 on Build GRCh38
rs902341484 Chr.5: 172958951-172958951 on Build GRCh38
rs964899856 Chr.5: 172959074-172959074 on Build GRCh38
rs922335332 Chr.5: 172959293-172959293 on Build GRCh38
rs568600027 Chr.5: 172958950-172958950 on Build GRCh38
rs113356182 Chr.5: 172959050-172959050 on Build GRCh38
rs7732462 Chr.5: 172958924-172958924 on Build GRCh38
rs896216281 Chr.5: 172959002-172959002 on Build GRCh38
rs1013292471 Chr.5: 172959003-172959003 on Build GRCh38
rs913234598 Chr.5: 172959348-172959349 on Build GRCh38
rs908060440 Chr.5: 172959083-172959083 on Build GRCh38
rs562205625 Chr.5: 172959310-172959310 on Build GRCh38
rs1043629792 Chr.5: 172959330-172959330 on Build GRCh38
rs79066055 Chr.5: 172959326-172959326 on Build GRCh38
rs910880175 Chr.5: 172959323-172959323 on Build GRCh38
rs573217724 Chr.5: 172959223-172959223 on Build GRCh38
rs962171898 Chr.5: 172959273-172959273 on Build GRCh38
rs113436731 Chr.5: 172959217-172959217 on Build GRCh38
rs1012228823 Chr.5: 172959012-172959012 on Build GRCh38
rs541856577 Chr.5: 172959302-172959302 on Build GRCh38
rs999634085 Chr.5: 172958953-172958953 on Build GRCh38
rs151083493 Chr.5: 172959364-172959364 on Build GRCh38
rs1043646930 Chr.5: 172959362-172959362 on Build GRCh38
rs1016501847 Chr.5: 172959054-172959054 on Build GRCh38
rs1049828321 Chr.5: 172959306-172959306 on Build GRCh38
rs950380479 Chr.5: 172959027-172959027 on Build GRCh38
rs398102496 Chr.5: 172959288-172959289 on Build GRCh38
rs538816241 Chr.5: 172958987-172958987 on Build GRCh38
rs1010748642 Chr.5: 172958934-172958934 on Build GRCh38
rs933662145 Chr.5: 172959297-172959297 on Build GRCh38
rs540144680 Chr.5: 172959093-172959093 on Build GRCh38
rs980405211 Chr.5: 172959009-172959009 on Build GRCh38
rs902374925 Chr.5: 172959360-172959360 on Build GRCh38
rs972472020 Chr.5: 172959278-172959278 on Build GRCh38
rs1023890320 Chr.5: 172959038-172959038 on Build GRCh38
rs945300955 Chr.5: 172958931-172958931 on Build GRCh38
rs937879911 Chr.5: 172959368-172959368 on Build GRCh38
rs114985587 Chr.5: 172958939-172958939 on Build GRCh38
rs1033462897 Chr.5: 172958992-172958992 on Build GRCh38
rs771570084 Chr.5: 172959222-172959222 on Build GRCh38
rs112505850 Chr.5: 172959287-172959288 on Build GRCh38
rs985058221 Chr.5: 172959052-172959052 on Build GRCh38
rs974895883 Chr.5: 172959102-172959102 on Build GRCh38
rs923416747 Chr.5: 172959255-172959255 on Build GRCh38
rs896532446 Chr.5: 172959376-172959376 on Build GRCh38

View Full Product Details