Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 133096412-133096909 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00326
Entrez Gene ID
285735
Gene Name
long intergenic non-protein coding RNA 326
Gene Aliases
NCRNA00326
UniGene
Hs.407549
Gene Chromosome Location
Chr.6: 133088080-133106578 on Build GRCh38
Transcripts
NR_026969.1

SNP Detail

SNP ID Chromosome Location
rs149128117 Chr.6: 133096635-133096635 on Build GRCh38
rs568467026 Chr.6: 133096817-133096817 on Build GRCh38
rs961181055 Chr.6: 133096550-133096550 on Build GRCh38
rs1000865046 Chr.6: 133096477-133096477 on Build GRCh38
rs925523138 Chr.6: 133096809-133096809 on Build GRCh38
rs1011628420 Chr.6: 133096807-133096807 on Build GRCh38
rs560110322 Chr.6: 133096716-133096716 on Build GRCh38
rs906938037 Chr.6: 133096444-133096445 on Build GRCh38
rs1013388485 Chr.6: 133096512-133096512 on Build GRCh38
rs1024733413 Chr.6: 133096542-133096542 on Build GRCh38
rs868262702 Chr.6: 133096531-133096531 on Build GRCh38
rs945563592 Chr.6: 133096770-133096770 on Build GRCh38
rs774981259 Chr.6: 133096679-133096679 on Build GRCh38
rs1007706786 Chr.6: 133096634-133096634 on Build GRCh38
rs538310465 Chr.6: 133096755-133096755 on Build GRCh38
rs775581818 Chr.6: 133096851-133096851 on Build GRCh38
rs966149142 Chr.6: 133096664-133096664 on Build GRCh38
rs938472392 Chr.6: 133096745-133096745 on Build GRCh38
rs928462876 Chr.6: 133096720-133096720 on Build GRCh38
rs976159540 Chr.6: 133096683-133096683 on Build GRCh38
rs760700680 Chr.6: 133096862-133096862 on Build GRCh38
rs984734429 Chr.6: 133096702-133096702 on Build GRCh38
rs994514442 Chr.6: 133096569-133096569 on Build GRCh38
rs1046446040 Chr.6: 133096874-133096874 on Build GRCh38
rs771196146 Chr.6: 133096496-133096496 on Build GRCh38
rs952493526 Chr.6: 133096615-133096615 on Build GRCh38
rs1043999584 Chr.6: 133096842-133096842 on Build GRCh38
rs554838770 Chr.6: 133096883-133096883 on Build GRCh38
rs773062135 Chr.6: 133096689-133096689 on Build GRCh38
rs1026897281 Chr.6: 133096608-133096608 on Build GRCh38
rs993981851 Chr.6: 133096888-133096888 on Build GRCh38
rs1055625117 Chr.6: 133096494-133096494 on Build GRCh38
rs903890214 Chr.6: 133096451-133096451 on Build GRCh38
rs985105776 Chr.6: 133096625-133096625 on Build GRCh38
rs6914577 Chr.6: 133096842-133096842 on Build GRCh38
rs936667857 Chr.6: 133096821-133096821 on Build GRCh38
rs895195459 Chr.6: 133096510-133096510 on Build GRCh38
rs950739277 Chr.6: 133096696-133096696 on Build GRCh38
rs1009823752 Chr.6: 133096586-133096586 on Build GRCh38
rs909094343 Chr.6: 133096707-133096707 on Build GRCh38
rs181680353 Chr.6: 133096777-133096777 on Build GRCh38
rs1055107134 Chr.6: 133096841-133096841 on Build GRCh38
rs547610060 Chr.6: 133096631-133096631 on Build GRCh38
rs191054830 Chr.6: 133096765-133096765 on Build GRCh38
rs570730321 Chr.6: 133096632-133096632 on Build GRCh38

View Full Product Details