Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73710955-73711472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4284
Entrez Gene ID
100422948
Gene Name
microRNA 4284
Gene Aliases
mir-4284
UniGene
Gene Chromosome Location
Chr.7: 73711317-73711397 on Build GRCh38
Transcripts
NR_036247.1 NM_001165903.1 NM_004603.3 XM_017012567.1

Gene Symbol
STX1A
Entrez Gene ID
6804
Gene Name
syntaxin 1A
Gene Aliases
HPC-1, P35-1, STX1, SYN1A
UniGene
Hs.647024
Gene Chromosome Location
Chr.7: 73699205-73719702 on Build GRCh38
Transcripts
NR_036247.1 NM_001165903.1 NM_004603.3 XM_017012567.1

SNP Detail

SNP ID Chromosome Location
rs978125470 Chr.7: 73711036-73711036 on Build GRCh38
rs192013228 Chr.7: 73711428-73711428 on Build GRCh38
rs940585193 Chr.7: 73711305-73711305 on Build GRCh38
rs554003955 Chr.7: 73711395-73711395 on Build GRCh38
rs111645856 Chr.7: 73711070-73711070 on Build GRCh38
rs531637367 Chr.7: 73711065-73711065 on Build GRCh38
rs1034507207 Chr.7: 73711362-73711362 on Build GRCh38
rs782557210 Chr.7: 73711360-73711360 on Build GRCh38
rs976553845 Chr.7: 73711113-73711113 on Build GRCh38
rs547982737 Chr.7: 73711351-73711351 on Build GRCh38
rs11973069 Chr.7: 73711334-73711334 on Build GRCh38
rs782136021 Chr.7: 73711035-73711035 on Build GRCh38
rs782493498 Chr.7: 73711293-73711293 on Build GRCh38
rs923908921 Chr.7: 73711081-73711081 on Build GRCh38
rs1022437719 Chr.7: 73711427-73711427 on Build GRCh38
rs959515019 Chr.7: 73711019-73711019 on Build GRCh38
rs931807113 Chr.7: 73711262-73711262 on Build GRCh38
rs563498315 Chr.7: 73711370-73711371 on Build GRCh38
rs536831155 Chr.7: 73711434-73711434 on Build GRCh38
rs374282751 Chr.7: 73711202-73711202 on Build GRCh38
rs372296490 Chr.7: 73711193-73711193 on Build GRCh38
rs538948303 Chr.7: 73711082-73711082 on Build GRCh38
rs782759562 Chr.7: 73710997-73710997 on Build GRCh38
rs781857697 Chr.7: 73711335-73711335 on Build GRCh38
rs781846366 Chr.7: 73711415-73711415 on Build GRCh38
rs1022551964 Chr.7: 73711023-73711023 on Build GRCh38
rs781789847 Chr.7: 73711112-73711112 on Build GRCh38
rs991105481 Chr.7: 73711020-73711020 on Build GRCh38
rs142352692 Chr.7: 73711001-73711001 on Build GRCh38
rs895356504 Chr.7: 73711396-73711396 on Build GRCh38
rs1049003746 Chr.7: 73711273-73711273 on Build GRCh38
rs571408531 Chr.7: 73711306-73711306 on Build GRCh38
rs146471889 Chr.7: 73711005-73711005 on Build GRCh38
rs549759438 Chr.7: 73711092-73711092 on Build GRCh38
rs1012423714 Chr.7: 73711405-73711405 on Build GRCh38
rs921814961 Chr.7: 73711135-73711135 on Build GRCh38
rs781783779 Chr.7: 73711321-73711321 on Build GRCh38

View Full Product Details