Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117198497-117198998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST7
Entrez Gene ID
7982
Gene Name
suppression of tumorigenicity 7
Gene Aliases
ETS7q, FAM4A, FAM4A1, HELG, RAY1, SEN4, TSG7
UniGene
Hs.368131
Gene Chromosome Location
Chr.7: 116953327-117230023 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2 NR_002332.2

Gene Symbol
ST7-OT3
Entrez Gene ID
93655
Gene Name
ST7 overlapping transcript 3
Gene Aliases
NCRNA00026, ST7, ST7OT3
UniGene
Hs.368131
Gene Chromosome Location
Chr.7: 117182681-117209937 on Build GRCh38
Transcripts
NM_018412.3 NM_021908.2 NR_002332.2

SNP Detail

SNP ID Chromosome Location
rs924579466 Chr.7: 117198645-117198645 on Build GRCh38
rs978391642 Chr.7: 117198888-117198888 on Build GRCh38
rs868444297 Chr.7: 117198530-117198530 on Build GRCh38
rs966583778 Chr.7: 117198857-117198857 on Build GRCh38
rs989307719 Chr.7: 117198607-117198607 on Build GRCh38
rs908710867 Chr.7: 117198772-117198772 on Build GRCh38
rs575903629 Chr.7: 117198675-117198675 on Build GRCh38
rs945311393 Chr.7: 117198614-117198614 on Build GRCh38
rs994125962 Chr.7: 117198719-117198719 on Build GRCh38
rs938632603 Chr.7: 117198948-117198948 on Build GRCh38
rs964470403 Chr.7: 117198826-117198826 on Build GRCh38
rs987157794 Chr.7: 117198579-117198579 on Build GRCh38
rs1041706793 Chr.7: 117198635-117198635 on Build GRCh38
rs1026584306 Chr.7: 117198723-117198723 on Build GRCh38
rs1057201556 Chr.7: 117198664-117198664 on Build GRCh38
rs369603486 Chr.7: 117198751-117198751 on Build GRCh38
rs143607954 Chr.7: 117198701-117198701 on Build GRCh38
rs951850857 Chr.7: 117198755-117198755 on Build GRCh38
rs1008525190 Chr.7: 117198816-117198816 on Build GRCh38
rs1057191377 Chr.7: 117198966-117198966 on Build GRCh38
rs893893876 Chr.7: 117198937-117198937 on Build GRCh38
rs1013084655 Chr.7: 117198682-117198682 on Build GRCh38
rs915160426 Chr.7: 117198611-117198611 on Build GRCh38
rs749273287 Chr.7: 117198580-117198580 on Build GRCh38
rs1026427571 Chr.7: 117198558-117198558 on Build GRCh38
rs893856695 Chr.7: 117198616-117198616 on Build GRCh38
rs193217405 Chr.7: 117198710-117198710 on Build GRCh38
rs138798329 Chr.7: 117198524-117198524 on Build GRCh38
rs954339229 Chr.7: 117198747-117198747 on Build GRCh38
rs966656456 Chr.7: 117198699-117198699 on Build GRCh38
rs954414615 Chr.7: 117198578-117198578 on Build GRCh38
rs984255604 Chr.7: 117198770-117198770 on Build GRCh38
rs774236875 Chr.7: 117198730-117198730 on Build GRCh38
rs1041265537 Chr.7: 117198840-117198840 on Build GRCh38
rs1017300517 Chr.7: 117198825-117198825 on Build GRCh38
rs149390229 Chr.7: 117198592-117198592 on Build GRCh38
rs938684766 Chr.7: 117198648-117198648 on Build GRCh38
rs915088987 Chr.7: 117198852-117198852 on Build GRCh38
rs35185604 Chr.7: 117198687-117198688 on Build GRCh38
rs557745685 Chr.7: 117198735-117198735 on Build GRCh38
rs927907517 Chr.7: 117198933-117198933 on Build GRCh38
rs932683714 Chr.7: 117198891-117198891 on Build GRCh38
rs374288215 Chr.7: 117198837-117198837 on Build GRCh38
rs1020761520 Chr.7: 117198687-117198687 on Build GRCh38
rs561806663 Chr.7: 117198717-117198717 on Build GRCh38
rs940309477 Chr.7: 117198828-117198828 on Build GRCh38

View Full Product Details