Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157864009-157864516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506585
Entrez Gene ID
100506585
Gene Name
uncharacterized LOC100506585
Gene Aliases
-
UniGene
Hs.661265
Gene Chromosome Location
Chr.7: 157854585-157866092 on Build GRCh38
Transcripts
NR_038966.1 NM_001308267.1 NM_001308268.1 NM_002847.4 NM_130842.3 NM_130843.3 XM_011516446.1 XM_017012475.1 XM_017012476.1

Gene Symbol
PTPRN2
Entrez Gene ID
5799
Gene Name
protein tyrosine phosphatase, receptor type N2
Gene Aliases
IA-2beta, IAR, ICAAR, PTPRP, R-PTP-N2
UniGene
Hs.490789
Gene Chromosome Location
Chr.7: 157539052-158587802 on Build GRCh38
Transcripts
NR_038966.1 NM_001308267.1 NM_001308268.1 NM_002847.4 NM_130842.3 NM_130843.3 XM_011516446.1 XM_017012475.1 XM_017012476.1

SNP Detail

SNP ID Chromosome Location
rs900797260 Chr.7: 157864228-157864228 on Build GRCh38
rs545632944 Chr.7: 157864073-157864073 on Build GRCh38
rs116252836 Chr.7: 157864037-157864037 on Build GRCh38
rs908524017 Chr.7: 157864480-157864480 on Build GRCh38
rs769215535 Chr.7: 157864338-157864338 on Build GRCh38
rs539736969 Chr.7: 157864325-157864325 on Build GRCh38
rs947182716 Chr.7: 157864032-157864032 on Build GRCh38
rs1021970566 Chr.7: 157864326-157864326 on Build GRCh38
rs887224778 Chr.7: 157864260-157864260 on Build GRCh38
rs184867998 Chr.7: 157864196-157864196 on Build GRCh38
rs958641414 Chr.7: 157864170-157864170 on Build GRCh38
rs572729948 Chr.7: 157864335-157864335 on Build GRCh38
rs59661527 Chr.7: 157864083-157864083 on Build GRCh38
rs993058874 Chr.7: 157864295-157864295 on Build GRCh38
rs561327299 Chr.7: 157864147-157864147 on Build GRCh38
rs551322588 Chr.7: 157864399-157864399 on Build GRCh38
rs557327292 Chr.7: 157864301-157864301 on Build GRCh38
rs761046023 Chr.7: 157864386-157864386 on Build GRCh38
rs987236245 Chr.7: 157864347-157864347 on Build GRCh38
rs561488474 Chr.7: 157864340-157864340 on Build GRCh38
rs920175013 Chr.7: 157864464-157864464 on Build GRCh38
rs1052765765 Chr.7: 157864226-157864226 on Build GRCh38
rs184673103 Chr.7: 157864405-157864405 on Build GRCh38
rs552766455 Chr.7: 157864182-157864182 on Build GRCh38
rs372322767 Chr.7: 157864491-157864491 on Build GRCh38
rs529398779 Chr.7: 157864457-157864457 on Build GRCh38
rs771049873 Chr.7: 157864263-157864263 on Build GRCh38
rs756685046 Chr.7: 157864031-157864031 on Build GRCh38
rs752931364 Chr.7: 157864455-157864455 on Build GRCh38
rs535722536 Chr.7: 157864050-157864050 on Build GRCh38
rs1057161385 Chr.7: 157864395-157864395 on Build GRCh38
rs547148573 Chr.7: 157864238-157864238 on Build GRCh38
rs189710961 Chr.7: 157864288-157864288 on Build GRCh38
rs572570184 Chr.7: 157864088-157864088 on Build GRCh38
rs557502394 Chr.7: 157864328-157864328 on Build GRCh38
rs762555632 Chr.7: 157864382-157864382 on Build GRCh38
rs187310111 Chr.7: 157864115-157864115 on Build GRCh38
rs539327551 Chr.7: 157864483-157864483 on Build GRCh38
rs898461387 Chr.7: 157864235-157864235 on Build GRCh38
rs571074870 Chr.7: 157864183-157864183 on Build GRCh38
rs993555533 Chr.7: 157864411-157864411 on Build GRCh38
rs774484281 Chr.7: 157864276-157864276 on Build GRCh38
rs145403861 Chr.7: 157864474-157864474 on Build GRCh38
rs766805631 Chr.7: 157864391-157864391 on Build GRCh38
rs916092614 Chr.7: 157864198-157864199 on Build GRCh38
rs535592706 Chr.7: 157864261-157864261 on Build GRCh38
rs371069060 Chr.7: 157864243-157864243 on Build GRCh38
rs563875125 Chr.7: 157864169-157864169 on Build GRCh38
rs547111848 Chr.7: 157864437-157864437 on Build GRCh38
rs933906067 Chr.7: 157864229-157864229 on Build GRCh38
rs574912452 Chr.7: 157864042-157864042 on Build GRCh38
rs796218677 Chr.7: 157864200-157864200 on Build GRCh38
rs12668559 Chr.7: 157864385-157864385 on Build GRCh38
rs925711627 Chr.7: 157864204-157864204 on Build GRCh38
rs746938716 Chr.7: 157864246-157864246 on Build GRCh38
rs942942501 Chr.7: 157864212-157864212 on Build GRCh38
rs976018916 Chr.7: 157864458-157864458 on Build GRCh38
rs1047078221 Chr.7: 157864387-157864387 on Build GRCh38
rs527347613 Chr.7: 157864173-157864173 on Build GRCh38
rs1017773639 Chr.7: 157864456-157864456 on Build GRCh38
rs573580551 Chr.7: 157864366-157864366 on Build GRCh38
rs568414248 Chr.7: 157864438-157864438 on Build GRCh38
rs912800367 Chr.7: 157864209-157864209 on Build GRCh38
rs139348053 Chr.7: 157864148-157864148 on Build GRCh38
rs887172540 Chr.7: 157864390-157864390 on Build GRCh38
rs1014195605 Chr.7: 157864430-157864430 on Build GRCh38
rs1021540576 Chr.7: 157864414-157864414 on Build GRCh38
rs904473786 Chr.7: 157864420-157864420 on Build GRCh38
rs551041331 Chr.7: 157864089-157864089 on Build GRCh38
rs189648994 Chr.7: 157864383-157864383 on Build GRCh38
rs192362836 Chr.7: 157864367-157864367 on Build GRCh38
rs151292496 Chr.7: 157864072-157864072 on Build GRCh38
rs58539972 Chr.7: 157864339-157864339 on Build GRCh38
rs924359851 Chr.7: 157864197-157864197 on Build GRCh38
rs117326979 Chr.7: 157864479-157864479 on Build GRCh38
rs1025690388 Chr.7: 157864134-157864134 on Build GRCh38
rs55859645 Chr.7: 157864058-157864058 on Build GRCh38
rs988580423 Chr.7: 157864490-157864490 on Build GRCh38
rs1040373153 Chr.7: 157864223-157864223 on Build GRCh38
rs553372939 Chr.7: 157864041-157864041 on Build GRCh38
rs181905094 Chr.7: 157864384-157864384 on Build GRCh38
rs765285163 Chr.7: 157864422-157864422 on Build GRCh38
rs575407549 Chr.7: 157864317-157864317 on Build GRCh38
rs987183767 Chr.7: 157864443-157864443 on Build GRCh38
rs184272538 Chr.7: 157864375-157864375 on Build GRCh38
rs956165644 Chr.7: 157864484-157864484 on Build GRCh38
rs549057451 Chr.7: 157864232-157864232 on Build GRCh38
rs577241980 Chr.7: 157864082-157864082 on Build GRCh38
rs935483428 Chr.7: 157864199-157864199 on Build GRCh38
rs898508461 Chr.7: 157864105-157864105 on Build GRCh38
rs993498306 Chr.7: 157864262-157864263 on Build GRCh38
rs191805989 Chr.7: 157864153-157864153 on Build GRCh38

View Full Product Details