Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 40831716-40832257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100132831
Entrez Gene ID
100132831
Gene Name
A20-binding inhibitor of NF-kappaB activation 2 pseudogene
Gene Aliases
-
UniGene
Hs.729923
Gene Chromosome Location
Chr.X: 40831217-40833196 on Build GRCh38
Transcripts
NR_027761.1

SNP Detail

SNP ID Chromosome Location
rs182604330 Chr.X: 40831806-40831806 on Build GRCh38
rs1014439761 Chr.X: 40831991-40831991 on Build GRCh38
rs753099790 Chr.X: 40832161-40832161 on Build GRCh38
rs967864084 Chr.X: 40832205-40832205 on Build GRCh38
rs925762927 Chr.X: 40832225-40832225 on Build GRCh38
rs1005092553 Chr.X: 40832226-40832226 on Build GRCh38
rs945865895 Chr.X: 40831826-40831826 on Build GRCh38
rs767990928 Chr.X: 40831903-40831903 on Build GRCh38
rs563567276 Chr.X: 40832088-40832088 on Build GRCh38
rs771303911 Chr.X: 40832040-40832040 on Build GRCh38
rs977285240 Chr.X: 40832223-40832223 on Build GRCh38
rs765550016 Chr.X: 40832147-40832147 on Build GRCh38
rs774675818 Chr.X: 40832043-40832043 on Build GRCh38
rs775489017 Chr.X: 40832118-40832118 on Build GRCh38
rs904478942 Chr.X: 40831891-40831891 on Build GRCh38
rs760823037 Chr.X: 40832121-40832121 on Build GRCh38
rs188117767 Chr.X: 40831987-40831987 on Build GRCh38
rs190340545 Chr.X: 40832127-40832127 on Build GRCh38
rs935894736 Chr.X: 40831892-40831892 on Build GRCh38
rs911724970 Chr.X: 40831809-40831809 on Build GRCh38
rs931619295 Chr.X: 40832180-40832180 on Build GRCh38
rs760001090 Chr.X: 40832075-40832075 on Build GRCh38
rs1041658590 Chr.X: 40831849-40831849 on Build GRCh38
rs771685334 Chr.X: 40832034-40832034 on Build GRCh38
rs1004970282 Chr.X: 40832069-40832069 on Build GRCh38
rs973472649 Chr.X: 40831786-40831786 on Build GRCh38
rs768758053 Chr.X: 40831740-40831740 on Build GRCh38
rs963789907 Chr.X: 40831778-40831778 on Build GRCh38
rs66771561 Chr.X: 40831928-40831928 on Build GRCh38
rs575444938 Chr.X: 40832152-40832152 on Build GRCh38
rs990713682 Chr.X: 40831917-40831917 on Build GRCh38
rs887104153 Chr.X: 40831898-40831898 on Build GRCh38
rs761471385 Chr.X: 40831960-40831960 on Build GRCh38
rs1015045188 Chr.X: 40832074-40832074 on Build GRCh38
rs917821521 Chr.X: 40831983-40831983 on Build GRCh38
rs759894033 Chr.X: 40832091-40832091 on Build GRCh38
rs949257874 Chr.X: 40832073-40832073 on Build GRCh38
rs932081393 Chr.X: 40831752-40831752 on Build GRCh38
rs1029424936 Chr.X: 40832094-40832094 on Build GRCh38
rs371253883 Chr.X: 40832076-40832076 on Build GRCh38
rs182963606 Chr.X: 40832183-40832183 on Build GRCh38

View Full Product Details