Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 56737013-56737591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01420
Entrez Gene ID
550643
Gene Name
long intergenic non-protein coding RNA 1420
Gene Aliases
-
UniGene
Hs.355559
Gene Chromosome Location
Chr.X: 56729285-56817571 on Build GRCh38
Transcripts
NR_015367.2 NR_002308.1

Gene Symbol
UQCRBP1
Entrez Gene ID
442454
Gene Name
ubiquinol-cytochrome c reductase binding protein pseudogene 1
Gene Aliases
-
UniGene
Hs.663220
Gene Chromosome Location
Chr.X: 56736788-56737584 on Build GRCh38
Transcripts
NR_015367.2 NR_002308.1

SNP Detail

SNP ID Chromosome Location
rs941449846 Chr.X: 56737172-56737172 on Build GRCh38
rs989152991 Chr.X: 56737065-56737065 on Build GRCh38
rs192770057 Chr.X: 56737446-56737446 on Build GRCh38
rs1051808367 Chr.X: 56737277-56737277 on Build GRCh38
rs1024674641 Chr.X: 56737497-56737497 on Build GRCh38
rs758087970 Chr.X: 56737355-56737355 on Build GRCh38
rs897344038 Chr.X: 56737197-56737197 on Build GRCh38
rs1009014711 Chr.X: 56737286-56737286 on Build GRCh38
rs778405835 Chr.X: 56737149-56737149 on Build GRCh38
rs879062909 Chr.X: 56737428-56737428 on Build GRCh38
rs185346569 Chr.X: 56737134-56737134 on Build GRCh38
rs11088 Chr.X: 56737483-56737483 on Build GRCh38
rs755628194 Chr.X: 56737311-56737311 on Build GRCh38
rs878979506 Chr.X: 56737448-56737448 on Build GRCh38
rs999958872 Chr.X: 56737378-56737378 on Build GRCh38
rs768093050 Chr.X: 56737227-56737227 on Build GRCh38
rs917239208 Chr.X: 56737101-56737101 on Build GRCh38
rs879225230 Chr.X: 56737461-56737461 on Build GRCh38
rs980031072 Chr.X: 56737525-56737525 on Build GRCh38
rs2077257 Chr.X: 56737412-56737412 on Build GRCh38
rs878888475 Chr.X: 56737300-56737300 on Build GRCh38
rs780382482 Chr.X: 56737478-56737478 on Build GRCh38
rs1006644498 Chr.X: 56737494-56737494 on Build GRCh38
rs879117414 Chr.X: 56737295-56737295 on Build GRCh38
rs956090667 Chr.X: 56737415-56737415 on Build GRCh38
rs768820706 Chr.X: 56737508-56737508 on Build GRCh38
rs774134401 Chr.X: 56737559-56737559 on Build GRCh38
rs878934630 Chr.X: 56737396-56737396 on Build GRCh38
rs1032820847 Chr.X: 56737383-56737383 on Build GRCh38
rs878907589 Chr.X: 56737449-56737449 on Build GRCh38
rs879135908 Chr.X: 56737369-56737369 on Build GRCh38
rs761759639 Chr.X: 56737569-56737569 on Build GRCh38
rs879161848 Chr.X: 56737367-56737367 on Build GRCh38
rs1038469424 Chr.X: 56737188-56737188 on Build GRCh38
rs188569110 Chr.X: 56737379-56737379 on Build GRCh38
rs879247275 Chr.X: 56737292-56737292 on Build GRCh38
rs879013332 Chr.X: 56737393-56737393 on Build GRCh38
rs2009742 Chr.X: 56737504-56737504 on Build GRCh38
rs746881612 Chr.X: 56737394-56737394 on Build GRCh38
rs971418236 Chr.X: 56737509-56737510 on Build GRCh38
rs753164791 Chr.X: 56737280-56737280 on Build GRCh38
rs41306884 Chr.X: 56737360-56737360 on Build GRCh38
rs777209236 Chr.X: 56737335-56737335 on Build GRCh38
rs955952565 Chr.X: 56737063-56737063 on Build GRCh38
rs756994063 Chr.X: 56737135-56737135 on Build GRCh38

View Full Product Details