Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 69919164-69919748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC7
Entrez Gene ID
57554
Gene Name
leucine rich repeat containing 7
Gene Aliases
DENSIN
UniGene
Hs.479658
Gene Chromosome Location
Chr.1: 69567130-70152426 on Build GRCh38
Transcripts
NM_020794.2 XM_017001885.1 XM_017001886.1 XM_017001887.1 XM_017001888.1 XM_017001889.1 XM_017001890.1 XM_017001891.1 XM_017001892.1 XM_017001893.1 XM_017001894.1 XM_017001895.1 NR_023916.1

Gene Symbol
PIN1P1
Entrez Gene ID
5301
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 1 pseudogene 1
Gene Aliases
PIN1L
UniGene
Hs.654711
Gene Chromosome Location
Chr.1: 69919322-69920317 on Build GRCh38
Transcripts
NM_020794.2 XM_017001885.1 XM_017001886.1 XM_017001887.1 XM_017001888.1 XM_017001889.1 XM_017001890.1 XM_017001891.1 XM_017001892.1 XM_017001893.1 XM_017001894.1 XM_017001895.1 NR_023916.1

SNP Detail

SNP ID Chromosome Location
rs932514292 Chr.1: 69919204-69919204 on Build GRCh38
rs1004454132 Chr.1: 69919443-69919443 on Build GRCh38
rs374893471 Chr.1: 69919599-69919599 on Build GRCh38
rs192680489 Chr.1: 69919268-69919268 on Build GRCh38
rs1027793616 Chr.1: 69919585-69919585 on Build GRCh38
rs1042799542 Chr.1: 69919254-69919254 on Build GRCh38
rs1027762558 Chr.1: 69919303-69919303 on Build GRCh38
rs572541131 Chr.1: 69919352-69919352 on Build GRCh38
rs953070807 Chr.1: 69919509-69919509 on Build GRCh38
rs1004910513 Chr.1: 69919208-69919208 on Build GRCh38
rs938538374 Chr.1: 69919365-69919365 on Build GRCh38
rs922434824 Chr.1: 69919195-69919195 on Build GRCh38
rs543129303 Chr.1: 69919600-69919600 on Build GRCh38
rs961412728 Chr.1: 69919619-69919619 on Build GRCh38
rs985761620 Chr.1: 69919283-69919283 on Build GRCh38
rs968663414 Chr.1: 69919307-69919307 on Build GRCh38
rs11802766 Chr.1: 69919618-69919618 on Build GRCh38
rs959612279 Chr.1: 69919274-69919274 on Build GRCh38
rs750125100 Chr.1: 69919571-69919571 on Build GRCh38
rs902513896 Chr.1: 69919522-69919522 on Build GRCh38
rs915783398 Chr.1: 69919596-69919596 on Build GRCh38
rs932545281 Chr.1: 69919417-69919417 on Build GRCh38
rs946801804 Chr.1: 69919489-69919489 on Build GRCh38
rs1049611365 Chr.1: 69919442-69919442 on Build GRCh38
rs953959909 Chr.1: 69919669-69919669 on Build GRCh38
rs952483625 Chr.1: 69919308-69919308 on Build GRCh38
rs1037710931 Chr.1: 69919488-69919488 on Build GRCh38
rs745840859 Chr.1: 69919375-69919375 on Build GRCh38
rs61782602 Chr.1: 69919586-69919586 on Build GRCh38
rs1029316067 Chr.1: 69919662-69919662 on Build GRCh38
rs968819937 Chr.1: 69919524-69919524 on Build GRCh38
rs768159990 Chr.1: 69919655-69919655 on Build GRCh38
rs1007356902 Chr.1: 69919517-69919517 on Build GRCh38
rs532306181 Chr.1: 69919354-69919354 on Build GRCh38
rs185006918 Chr.1: 69919191-69919191 on Build GRCh38
rs745689382 Chr.1: 69919279-69919279 on Build GRCh38
rs998730154 Chr.1: 69919499-69919499 on Build GRCh38
rs1042438825 Chr.1: 69919507-69919507 on Build GRCh38
rs560592583 Chr.1: 69919264-69919264 on Build GRCh38
rs888658503 Chr.1: 69919198-69919198 on Build GRCh38
rs948808524 Chr.1: 69919386-69919386 on Build GRCh38
rs985344406 Chr.1: 69919675-69919675 on Build GRCh38
rs915053204 Chr.1: 69919463-69919463 on Build GRCh38
rs961034724 Chr.1: 69919371-69919371 on Build GRCh38
rs184822845 Chr.1: 69919344-69919344 on Build GRCh38
rs534355222 Chr.1: 69919608-69919608 on Build GRCh38
rs908213485 Chr.1: 69919353-69919353 on Build GRCh38
rs188409633 Chr.1: 69919587-69919587 on Build GRCh38
rs984196984 Chr.1: 69919603-69919603 on Build GRCh38
rs150791147 Chr.1: 69919238-69919238 on Build GRCh38
rs976474631 Chr.1: 69919630-69919630 on Build GRCh38
rs550110565 Chr.1: 69919381-69919381 on Build GRCh38
rs1048378430 Chr.1: 69919393-69919393 on Build GRCh38
rs564950734 Chr.1: 69919215-69919215 on Build GRCh38
rs1010971273 Chr.1: 69919239-69919239 on Build GRCh38
rs879797543 Chr.1: 69919530-69919530 on Build GRCh38
rs570322947 Chr.1: 69919414-69919414 on Build GRCh38
rs567697555 Chr.1: 69919269-69919269 on Build GRCh38
rs924456561 Chr.1: 69919362-69919362 on Build GRCh38
rs1035238958 Chr.1: 69919273-69919273 on Build GRCh38
rs187488570 Chr.1: 69919197-69919197 on Build GRCh38
rs1037046389 Chr.1: 69919706-69919706 on Build GRCh38
rs879226414 Chr.1: 69919693-69919693 on Build GRCh38
rs539357005 Chr.1: 69919523-69919523 on Build GRCh38
rs1056382341 Chr.1: 69919539-69919539 on Build GRCh38
rs750775784 Chr.1: 69919715-69919715 on Build GRCh38
rs755734303 Chr.1: 69919577-69919577 on Build GRCh38
rs948797099 Chr.1: 69919598-69919598 on Build GRCh38
rs770684530 Chr.1: 69919242-69919242 on Build GRCh38

View Full Product Details