Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21351118-21351673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIPEPP3
Entrez Gene ID
650794
Gene Name
mitochondrial intermediate peptidase pseudogene 3
Gene Aliases
-
UniGene
Hs.147563
Gene Chromosome Location
Chr.13: 21298125-21392923 on Build GRCh38
Transcripts
NR_046461.1

SNP Detail

SNP ID Chromosome Location
rs912462188 Chr.13: 21351463-21351463 on Build GRCh38
rs1010515027 Chr.13: 21351291-21351291 on Build GRCh38
rs1038002500 Chr.13: 21351148-21351148 on Build GRCh38
rs918280073 Chr.13: 21351355-21351355 on Build GRCh38
rs552831363 Chr.13: 21351493-21351493 on Build GRCh38
rs768625285 Chr.13: 21351407-21351407 on Build GRCh38
rs962837111 Chr.13: 21351605-21351605 on Build GRCh38
rs538700606 Chr.13: 21351329-21351329 on Build GRCh38
rs973763547 Chr.13: 21351319-21351319 on Build GRCh38
rs185819412 Chr.13: 21351633-21351633 on Build GRCh38
rs971621260 Chr.13: 21351295-21351295 on Build GRCh38
rs548474547 Chr.13: 21351189-21351189 on Build GRCh38
rs1000296165 Chr.13: 21351514-21351514 on Build GRCh38
rs778457430 Chr.13: 21351314-21351314 on Build GRCh38
rs763447579 Chr.13: 21351594-21351594 on Build GRCh38
rs114540479 Chr.13: 21351181-21351181 on Build GRCh38
rs535745228 Chr.13: 21351545-21351545 on Build GRCh38
rs372522615 Chr.13: 21351356-21351356 on Build GRCh38
rs929627080 Chr.13: 21351437-21351437 on Build GRCh38
rs745425094 Chr.13: 21351373-21351373 on Build GRCh38
rs142400727 Chr.13: 21351531-21351531 on Build GRCh38
rs756684088 Chr.13: 21351308-21351308 on Build GRCh38
rs973817666 Chr.13: 21351625-21351625 on Build GRCh38
rs59012420 Chr.13: 21351570-21351570 on Build GRCh38
rs1058533 Chr.13: 21351223-21351223 on Build GRCh38
rs761976986 Chr.13: 21351525-21351525 on Build GRCh38
rs1963728 Chr.13: 21351299-21351299 on Build GRCh38
rs376089033 Chr.13: 21351489-21351489 on Build GRCh38
rs1039154016 Chr.13: 21351201-21351201 on Build GRCh38
rs116989024 Chr.13: 21351206-21351206 on Build GRCh38
rs377294027 Chr.13: 21351526-21351526 on Build GRCh38
rs553859362 Chr.13: 21351451-21351451 on Build GRCh38
rs373153129 Chr.13: 21351506-21351506 on Build GRCh38
rs536464054 Chr.13: 21351188-21351188 on Build GRCh38
rs1964148 Chr.13: 21351534-21351534 on Build GRCh38
rs572271960 Chr.13: 21351456-21351456 on Build GRCh38
rs1033029945 Chr.13: 21351254-21351254 on Build GRCh38
rs1024974108 Chr.13: 21351294-21351294 on Build GRCh38
rs369342187 Chr.13: 21351499-21351499 on Build GRCh38
rs148322827 Chr.13: 21351579-21351580 on Build GRCh38
rs776881573 Chr.13: 21351515-21351515 on Build GRCh38
rs918333986 Chr.13: 21351639-21351639 on Build GRCh38
rs900593667 Chr.13: 21351216-21351216 on Build GRCh38
rs9580089 Chr.13: 21351258-21351258 on Build GRCh38
rs1034943340 Chr.13: 21351590-21351590 on Build GRCh38
rs891840028 Chr.13: 21351530-21351530 on Build GRCh38
rs980682786 Chr.13: 21351298-21351298 on Build GRCh38
rs962453207 Chr.13: 21351300-21351300 on Build GRCh38
rs942599450 Chr.13: 21351492-21351492 on Build GRCh38
rs778313983 Chr.13: 21351180-21351180 on Build GRCh38
rs148797913 Chr.13: 21351174-21351174 on Build GRCh38

View Full Product Details