Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77915542-77916089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC645752
Entrez Gene ID
645752
Gene Name
golgin A6 family member A pseudogene
Gene Aliases
-
UniGene
Hs.546408
Gene Chromosome Location
Chr.15: 77914217-77926846 on Build GRCh38
Transcripts
NR_027024.1

SNP Detail

SNP ID Chromosome Location
rs755465024 Chr.15: 77915856-77915856 on Build GRCh38
rs760077534 Chr.15: 77915910-77915910 on Build GRCh38
rs916587649 Chr.15: 77915868-77915868 on Build GRCh38
rs1035501112 Chr.15: 77916035-77916035 on Build GRCh38
rs775459051 Chr.15: 77915747-77915747 on Build GRCh38
rs778051216 Chr.15: 77915599-77915599 on Build GRCh38
rs147455906 Chr.15: 77915566-77915566 on Build GRCh38
rs960756137 Chr.15: 77916049-77916049 on Build GRCh38
rs999249220 Chr.15: 77915767-77915767 on Build GRCh38
rs754995714 Chr.15: 77915945-77915945 on Build GRCh38
rs749619980 Chr.15: 77915723-77915723 on Build GRCh38
rs758076944 Chr.15: 77915729-77915729 on Build GRCh38
rs185285135 Chr.15: 77915613-77915613 on Build GRCh38
rs761987415 Chr.15: 77915838-77915838 on Build GRCh38
rs548360119 Chr.15: 77915756-77915756 on Build GRCh38
rs76380402 Chr.15: 77915894-77915894 on Build GRCh38
rs781499011 Chr.15: 77915608-77915608 on Build GRCh38
rs746699403 Chr.15: 77915800-77915800 on Build GRCh38
rs774272771 Chr.15: 77915834-77915834 on Build GRCh38
rs746252853 Chr.15: 77915611-77915611 on Build GRCh38
rs769743279 Chr.15: 77915746-77915746 on Build GRCh38
rs552076226 Chr.15: 77916014-77916014 on Build GRCh38
rs749918857 Chr.15: 77915888-77915888 on Build GRCh38
rs558164823 Chr.15: 77915783-77915783 on Build GRCh38
rs77773002 Chr.15: 77915944-77915944 on Build GRCh38
rs777985803 Chr.15: 77915742-77915743 on Build GRCh38
rs774881633 Chr.15: 77915602-77915603 on Build GRCh38
rs971859817 Chr.15: 77916022-77916022 on Build GRCh38
rs187640529 Chr.15: 77915987-77915987 on Build GRCh38
rs74587726 Chr.15: 77915897-77915897 on Build GRCh38
rs775514164 Chr.15: 77915821-77915821 on Build GRCh38
rs67793790 Chr.15: 77915572-77915572 on Build GRCh38
rs768045619 Chr.15: 77915592-77915593 on Build GRCh38
rs552794838 Chr.15: 77915820-77915820 on Build GRCh38
rs753029024 Chr.15: 77915924-77915924 on Build GRCh38
rs569664786 Chr.15: 77915757-77915757 on Build GRCh38
rs566121347 Chr.15: 77915696-77915696 on Build GRCh38
rs138428146 Chr.15: 77915596-77915596 on Build GRCh38
rs756959736 Chr.15: 77915698-77915699 on Build GRCh38
rs544110699 Chr.15: 77915898-77915898 on Build GRCh38
rs757874732 Chr.15: 77915606-77915606 on Build GRCh38
rs752521626 Chr.15: 77915667-77915667 on Build GRCh38
rs777354432 Chr.15: 77915730-77915730 on Build GRCh38
rs762790778 Chr.15: 77915748-77915748 on Build GRCh38
rs776001730 Chr.15: 77915967-77915967 on Build GRCh38
rs765192630 Chr.15: 77915737-77915738 on Build GRCh38
rs965822118 Chr.15: 77915787-77915787 on Build GRCh38
rs776193716 Chr.15: 77915739-77915739 on Build GRCh38
rs146394787 Chr.15: 77915989-77915989 on Build GRCh38
rs751663536 Chr.15: 77915600-77915600 on Build GRCh38
rs750205804 Chr.15: 77915739-77915740 on Build GRCh38
rs746193558 Chr.15: 77915949-77915950 on Build GRCh38
rs777597114 Chr.15: 77915891-77915891 on Build GRCh38
rs180719506 Chr.15: 77915595-77915595 on Build GRCh38
rs74027203 Chr.15: 77915919-77915919 on Build GRCh38
rs756121360 Chr.15: 77915741-77915742 on Build GRCh38
rs553077610 Chr.15: 77915889-77915889 on Build GRCh38
rs767085153 Chr.15: 77915933-77915933 on Build GRCh38
rs368760820 Chr.15: 77916016-77916016 on Build GRCh38
rs751714838 Chr.15: 77915682-77915682 on Build GRCh38
rs547588963 Chr.15: 77915633-77915633 on Build GRCh38
rs772205860 Chr.15: 77915899-77915899 on Build GRCh38
rs547730640 Chr.15: 77915955-77915955 on Build GRCh38
rs992092219 Chr.15: 77915866-77915866 on Build GRCh38
rs770916407 Chr.15: 77915626-77915626 on Build GRCh38
rs58158543 Chr.15: 77915679-77915679 on Build GRCh38
rs745345571 Chr.15: 77915810-77915810 on Build GRCh38
rs755811450 Chr.15: 77915939-77915939 on Build GRCh38
rs765001281 Chr.15: 77915636-77915636 on Build GRCh38
rs760759405 Chr.15: 77915847-77915847 on Build GRCh38
rs939354702 Chr.15: 77915714-77915714 on Build GRCh38
rs778860021 Chr.15: 77915947-77915947 on Build GRCh38
rs536069423 Chr.15: 77915706-77915706 on Build GRCh38
rs756627224 Chr.15: 77915704-77915704 on Build GRCh38
rs772749888 Chr.15: 77915619-77915619 on Build GRCh38
rs191225724 Chr.15: 77915618-77915618 on Build GRCh38
rs71400903 Chr.15: 77915632-77915632 on Build GRCh38
rs779757262 Chr.15: 77915942-77915942 on Build GRCh38
rs536926627 Chr.15: 77915758-77915758 on Build GRCh38
rs58331113 Chr.15: 77915799-77915799 on Build GRCh38
rs534330332 Chr.15: 77916050-77916050 on Build GRCh38
rs766194699 Chr.15: 77915580-77915580 on Build GRCh38
rs542892043 Chr.15: 77915872-77915872 on Build GRCh38
rs534584583 Chr.15: 77915806-77915806 on Build GRCh38
rs749577220 Chr.15: 77915743-77915744 on Build GRCh38
rs763309413 Chr.15: 77915673-77915673 on Build GRCh38
rs771111936 Chr.15: 77915959-77915959 on Build GRCh38
rs748207897 Chr.15: 77915740-77915741 on Build GRCh38
rs756913991 Chr.15: 77915805-77915805 on Build GRCh38
rs757222027 Chr.15: 77915695-77915695 on Build GRCh38
rs750955607 Chr.15: 77915697-77915697 on Build GRCh38
rs77389582 Chr.15: 77915958-77915958 on Build GRCh38
rs746527196 Chr.15: 77915731-77915731 on Build GRCh38
rs750152663 Chr.15: 77915935-77915935 on Build GRCh38
rs753479936 Chr.15: 77915646-77915648 on Build GRCh38
rs1022914899 Chr.15: 77915586-77915586 on Build GRCh38
rs760803575 Chr.15: 77915903-77915903 on Build GRCh38
rs190786809 Chr.15: 77915590-77915590 on Build GRCh38
rs565542005 Chr.15: 77915909-77915909 on Build GRCh38
rs763841141 Chr.15: 77915615-77915620 on Build GRCh38
rs772694830 Chr.15: 77915574-77915574 on Build GRCh38
rs543158638 Chr.15: 77915589-77915589 on Build GRCh38
rs769983671 Chr.15: 77915612-77915612 on Build GRCh38
rs760653452 Chr.15: 77915576-77915576 on Build GRCh38
rs773425478 Chr.15: 77915900-77915900 on Build GRCh38
rs750492877 Chr.15: 77915780-77915780 on Build GRCh38
rs1006101837 Chr.15: 77916000-77916000 on Build GRCh38
rs763322724 Chr.15: 77915927-77915927 on Build GRCh38
rs766831974 Chr.15: 77915786-77915786 on Build GRCh38
rs770415220 Chr.15: 77915732-77915732 on Build GRCh38
rs999700434 Chr.15: 77915592-77915592 on Build GRCh38
rs753697793 Chr.15: 77915582-77915582 on Build GRCh38
rs183300573 Chr.15: 77915871-77915871 on Build GRCh38
rs779193506 Chr.15: 77915796-77915796 on Build GRCh38
rs755268960 Chr.15: 77915790-77915790 on Build GRCh38
rs749096331 Chr.15: 77915829-77915829 on Build GRCh38
rs771109529 Chr.15: 77915757-77915757 on Build GRCh38
rs768635422 Chr.15: 77915832-77915832 on Build GRCh38
rs867756072 Chr.15: 77915630-77915630 on Build GRCh38
rs530211294 Chr.15: 77915980-77915980 on Build GRCh38
rs541087776 Chr.15: 77915611-77915612 on Build GRCh38
rs763369971 Chr.15: 77915976-77915976 on Build GRCh38
rs759999157 Chr.15: 77915580-77915584 on Build GRCh38
rs745843412 Chr.15: 77915742-77915742 on Build GRCh38
rs559635202 Chr.15: 77915961-77915961 on Build GRCh38
rs909050137 Chr.15: 77915678-77915678 on Build GRCh38
rs768688875 Chr.15: 77915895-77915895 on Build GRCh38
rs548112599 Chr.15: 77915984-77915984 on Build GRCh38
rs770219150 Chr.15: 77915963-77915963 on Build GRCh38
rs532555992 Chr.15: 77915953-77915953 on Build GRCh38
rs765641116 Chr.15: 77915857-77915857 on Build GRCh38
rs753076532 Chr.15: 77915858-77915858 on Build GRCh38
rs766885176 Chr.15: 77915851-77915851 on Build GRCh38
rs74809134 Chr.15: 77915979-77915979 on Build GRCh38

View Full Product Details