Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18669858-18670409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXO3B
Entrez Gene ID
2310
Gene Name
forkhead box O3B pseudogene
Gene Aliases
FKHRL1P1, ZNF286C
UniGene
Hs.592126
Gene Chromosome Location
Chr.17: 18665923-18673181 on Build GRCh38
Transcripts
NR_026718.1 NM_001145045.1

Gene Symbol
ZNF286B
Entrez Gene ID
729288
Gene Name
zinc finger protein 286B
Gene Aliases
ZNF286C, ZNF286L, ZNF590
UniGene
Hs.534279
Gene Chromosome Location
Chr.17: 18658429-18682259 on Build GRCh38
Transcripts
NR_026718.1 NM_001145045.1

SNP Detail

SNP ID Chromosome Location
rs11657302 Chr.17: 18670029-18670029 on Build GRCh38
rs900019577 Chr.17: 18670320-18670320 on Build GRCh38
rs535134966 Chr.17: 18669943-18669943 on Build GRCh38
rs997016793 Chr.17: 18670329-18670329 on Build GRCh38
rs908300003 Chr.17: 18670142-18670142 on Build GRCh38
rs9905534 Chr.17: 18670033-18670033 on Build GRCh38
rs200279988 Chr.17: 18670262-18670262 on Build GRCh38
rs1011086990 Chr.17: 18669950-18669950 on Build GRCh38
rs1052531351 Chr.17: 18669896-18669896 on Build GRCh38
rs927913234 Chr.17: 18670209-18670209 on Build GRCh38
rs772259665 Chr.17: 18669974-18669976 on Build GRCh38
rs183254959 Chr.17: 18670114-18670114 on Build GRCh38
rs748269737 Chr.17: 18670157-18670157 on Build GRCh38
rs1015701912 Chr.17: 18670358-18670358 on Build GRCh38
rs577100020 Chr.17: 18669902-18669902 on Build GRCh38
rs1011430816 Chr.17: 18670267-18670267 on Build GRCh38
rs948612571 Chr.17: 18670189-18670189 on Build GRCh38
rs1052917991 Chr.17: 18670238-18670238 on Build GRCh38
rs964245163 Chr.17: 18670040-18670040 on Build GRCh38
rs1043869628 Chr.17: 18670278-18670278 on Build GRCh38
rs955071177 Chr.17: 18670139-18670139 on Build GRCh38
rs566229377 Chr.17: 18670348-18670348 on Build GRCh38
rs189666271 Chr.17: 18670241-18670241 on Build GRCh38
rs113183486 Chr.17: 18669973-18669975 on Build GRCh38
rs563779106 Chr.17: 18669899-18669899 on Build GRCh38
rs866020588 Chr.17: 18669942-18669942 on Build GRCh38
rs775560411 Chr.17: 18669975-18669977 on Build GRCh38
rs997094028 Chr.17: 18670100-18670100 on Build GRCh38
rs868091525 Chr.17: 18669944-18669944 on Build GRCh38
rs973885653 Chr.17: 18670158-18670158 on Build GRCh38
rs865861989 Chr.17: 18669976-18669976 on Build GRCh38
rs762760360 Chr.17: 18670127-18670127 on Build GRCh38
rs147429843 Chr.17: 18670331-18670331 on Build GRCh38
rs573457760 Chr.17: 18669928-18669929 on Build GRCh38
rs550221219 Chr.17: 18670233-18670233 on Build GRCh38
rs760938049 Chr.17: 18669984-18669984 on Build GRCh38
rs9907392 Chr.17: 18670031-18670031 on Build GRCh38
rs955186682 Chr.17: 18670346-18670346 on Build GRCh38
rs772136614 Chr.17: 18670188-18670188 on Build GRCh38
rs868548370 Chr.17: 18669948-18669948 on Build GRCh38
rs539693069 Chr.17: 18670359-18670359 on Build GRCh38
rs546024613 Chr.17: 18669906-18669906 on Build GRCh38
rs538856982 Chr.17: 18670322-18670322 on Build GRCh38
rs200414780 Chr.17: 18669970-18669970 on Build GRCh38
rs796509107 Chr.17: 18669973-18669976 on Build GRCh38
rs892603565 Chr.17: 18669924-18669924 on Build GRCh38
rs187724219 Chr.17: 18670202-18670202 on Build GRCh38
rs939383441 Chr.17: 18670232-18670232 on Build GRCh38

View Full Product Details