Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75590104-75590648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO15B
Entrez Gene ID
80022
Gene Name
myosin XVB
Gene Aliases
MYO15BP
UniGene
Hs.390817
Gene Chromosome Location
Chr.17: 75587545-75626850 on Build GRCh38
Transcripts
NM_001309242.1 XM_017025120.1 XM_017025121.1 XM_017025122.1 XM_017025123.1 XM_017025124.1 XM_017025125.1 XM_017025126.1 XM_017025127.1 XM_017025128.1 XM_017025129.1 XM_017025131.1 XM_017025132.1 XM_017025134.1 XM_017025141.1 XM_017025142.1 XM_017025144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84415 c.606G>A p.K202K Chr.17: 75590190-75590190 on Build GRCh38
7235448 c.662T>G p.L221* Chr.17: 75590246-75590246 on Build GRCh38
3680556 c.716C>T p.P239L Chr.17: 75590300-75590300 on Build GRCh38
6314388 c.668G>T p.G223V Chr.17: 75590252-75590252 on Build GRCh38
5619494 c.660G>A p.R220R Chr.17: 75590244-75590244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751890508 Chr.17: 75590140-75590140 on Build GRCh38
rs550553618 Chr.17: 75590527-75590527 on Build GRCh38
rs558139065 Chr.17: 75590325-75590325 on Build GRCh38
rs942679689 Chr.17: 75590206-75590206 on Build GRCh38
rs992906371 Chr.17: 75590136-75590136 on Build GRCh38
rs915851491 Chr.17: 75590381-75590381 on Build GRCh38
rs567117658 Chr.17: 75590166-75590166 on Build GRCh38
rs1016108552 Chr.17: 75590581-75590581 on Build GRCh38
rs79794615 Chr.17: 75590332-75590332 on Build GRCh38
rs996820671 Chr.17: 75590333-75590333 on Build GRCh38
rs989652179 Chr.17: 75590369-75590369 on Build GRCh38
rs117993948 Chr.17: 75590439-75590439 on Build GRCh38
rs779724912 Chr.17: 75590255-75590255 on Build GRCh38
rs964146678 Chr.17: 75590599-75590599 on Build GRCh38
rs774011016 Chr.17: 75590281-75590281 on Build GRCh38
rs555463736 Chr.17: 75590289-75590289 on Build GRCh38
rs540370172 Chr.17: 75590419-75590419 on Build GRCh38
rs933792962 Chr.17: 75590125-75590125 on Build GRCh38
rs1004268781 Chr.17: 75590561-75590561 on Build GRCh38
rs971420627 Chr.17: 75590147-75590147 on Build GRCh38
rs73998360 Chr.17: 75590244-75590244 on Build GRCh38
rs183131280 Chr.17: 75590467-75590467 on Build GRCh38
rs1011074522 Chr.17: 75590559-75590559 on Build GRCh38
rs1038298821 Chr.17: 75590170-75590170 on Build GRCh38
rs753553809 Chr.17: 75590432-75590432 on Build GRCh38
rs902820824 Chr.17: 75590470-75590470 on Build GRCh38
rs139813328 Chr.17: 75590300-75590300 on Build GRCh38
rs1022510213 Chr.17: 75590330-75590330 on Build GRCh38
rs191359022 Chr.17: 75590328-75590328 on Build GRCh38
rs911299159 Chr.17: 75590370-75590370 on Build GRCh38
rs1032438113 Chr.17: 75590577-75590577 on Build GRCh38
rs924130556 Chr.17: 75590157-75590157 on Build GRCh38
rs762969783 Chr.17: 75590402-75590402 on Build GRCh38
rs4789218 Chr.17: 75590443-75590443 on Build GRCh38
rs977210849 Chr.17: 75590627-75590627 on Build GRCh38
rs200802537 Chr.17: 75590366-75590366 on Build GRCh38
rs766234990 Chr.17: 75590376-75590376 on Build GRCh38
rs772974096 Chr.17: 75590351-75590351 on Build GRCh38
rs998552576 Chr.17: 75590478-75590478 on Build GRCh38
rs1024430882 Chr.17: 75590306-75590306 on Build GRCh38
rs116689597 Chr.17: 75590450-75590450 on Build GRCh38
rs820242 Chr.17: 75590277-75590277 on Build GRCh38
rs750349372 Chr.17: 75590279-75590279 on Build GRCh38
rs1046940588 Chr.17: 75590404-75590404 on Build GRCh38
rs781380175 Chr.17: 75590161-75590161 on Build GRCh38
rs918142438 Chr.17: 75590536-75590548 on Build GRCh38
rs748743776 Chr.17: 75590256-75590256 on Build GRCh38
rs894113899 Chr.17: 75590219-75590219 on Build GRCh38
rs62089225 Chr.17: 75590152-75590152 on Build GRCh38
rs1041475176 Chr.17: 75590209-75590209 on Build GRCh38
rs4789219 Chr.17: 75590479-75590479 on Build GRCh38
rs568490070 Chr.17: 75590348-75590348 on Build GRCh38
rs1031162932 Chr.17: 75590261-75590261 on Build GRCh38
rs62089226 Chr.17: 75590246-75590246 on Build GRCh38
rs79229026 Chr.17: 75590178-75590178 on Build GRCh38
rs955596867 Chr.17: 75590554-75590554 on Build GRCh38
rs575528560 Chr.17: 75590386-75590386 on Build GRCh38
rs911330248 Chr.17: 75590167-75590167 on Build GRCh38
rs1046878473 Chr.17: 75590250-75590250 on Build GRCh38
rs973959244 Chr.17: 75590377-75590377 on Build GRCh38
rs867165287 Chr.17: 75590406-75590406 on Build GRCh38
rs1025191937 Chr.17: 75590142-75590142 on Build GRCh38
rs958286382 Chr.17: 75590552-75590552 on Build GRCh38
rs752803255 Chr.17: 75590397-75590397 on Build GRCh38
rs536940116 Chr.17: 75590365-75590365 on Build GRCh38
rs373906828 Chr.17: 75590549-75590549 on Build GRCh38
rs928324277 Chr.17: 75590420-75590420 on Build GRCh38
rs1018475798 Chr.17: 75590572-75590572 on Build GRCh38
rs948384186 Chr.17: 75590242-75590242 on Build GRCh38

View Full Product Details