Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73672191-73672755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALMS1P1
Entrez Gene ID
200420
Gene Name
ALMS1, centrosome and basal body associated protein pseudogene 1
Gene Aliases
ALMS1L, ALMS1P
UniGene
Hs.739204
Gene Chromosome Location
Chr.2: 73644919-73685572 on Build GRCh38
Transcripts
NR_003683.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426234 c.3T>G p.I1M Chr.2: 73672486-73672486 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1035717079 Chr.2: 73672329-73672329 on Build GRCh38
rs565229657 Chr.2: 73672727-73672727 on Build GRCh38
rs561049722 Chr.2: 73672694-73672694 on Build GRCh38
rs759876439 Chr.2: 73672224-73672224 on Build GRCh38
rs924047932 Chr.2: 73672731-73672731 on Build GRCh38
rs543060844 Chr.2: 73672707-73672707 on Build GRCh38
rs376258897 Chr.2: 73672551-73672551 on Build GRCh38
rs915292420 Chr.2: 73672495-73672495 on Build GRCh38
rs555893690 Chr.2: 73672708-73672708 on Build GRCh38
rs985478471 Chr.2: 73672489-73672489 on Build GRCh38
rs1056703859 Chr.2: 73672562-73672562 on Build GRCh38
rs768529846 Chr.2: 73672638-73672638 on Build GRCh38
rs565598885 Chr.2: 73672347-73672347 on Build GRCh38
rs976334331 Chr.2: 73672684-73672684 on Build GRCh38
rs188406874 Chr.2: 73672547-73672547 on Build GRCh38
rs966304837 Chr.2: 73672406-73672406 on Build GRCh38
rs7574950 Chr.2: 73672400-73672400 on Build GRCh38
rs1049828916 Chr.2: 73672268-73672268 on Build GRCh38
rs537090988 Chr.2: 73672629-73672629 on Build GRCh38
rs538821237 Chr.2: 73672621-73672621 on Build GRCh38
rs887493788 Chr.2: 73672600-73672600 on Build GRCh38
rs922595995 Chr.2: 73672220-73672220 on Build GRCh38
rs2421586 Chr.2: 73672592-73672592 on Build GRCh38
rs187736652 Chr.2: 73672308-73672308 on Build GRCh38
rs1042902804 Chr.2: 73672518-73672518 on Build GRCh38
rs1003792807 Chr.2: 73672328-73672328 on Build GRCh38
rs902732650 Chr.2: 73672315-73672315 on Build GRCh38
rs753186404 Chr.2: 73672335-73672335 on Build GRCh38
rs777451079 Chr.2: 73672360-73672360 on Build GRCh38
rs575901646 Chr.2: 73672695-73672695 on Build GRCh38
rs566152123 Chr.2: 73672444-73672444 on Build GRCh38
rs145807705 Chr.2: 73672504-73672504 on Build GRCh38
rs193201865 Chr.2: 73672339-73672339 on Build GRCh38
rs369992040 Chr.2: 73672448-73672448 on Build GRCh38
rs572227303 Chr.2: 73672664-73672664 on Build GRCh38
rs115275930 Chr.2: 73672628-73672628 on Build GRCh38
rs2421585 Chr.2: 73672486-73672486 on Build GRCh38
rs773292537 Chr.2: 73672714-73672714 on Build GRCh38
rs532564954 Chr.2: 73672387-73672387 on Build GRCh38
rs185084950 Chr.2: 73672456-73672456 on Build GRCh38
rs116640029 Chr.2: 73672334-73672334 on Build GRCh38
rs560692923 Chr.2: 73672274-73672274 on Build GRCh38
rs192587238 Chr.2: 73672669-73672669 on Build GRCh38
rs932663135 Chr.2: 73672235-73672235 on Build GRCh38
rs893909470 Chr.2: 73672271-73672271 on Build GRCh38
rs1011843521 Chr.2: 73672590-73672590 on Build GRCh38
rs756978167 Chr.2: 73672407-73672407 on Build GRCh38
rs968495391 Chr.2: 73672710-73672710 on Build GRCh38
rs769522237 Chr.2: 73672555-73672555 on Build GRCh38
rs375660179 Chr.2: 73672348-73672348 on Build GRCh38
rs111840340 Chr.2: 73672303-73672303 on Build GRCh38
rs779864387 Chr.2: 73672573-73672573 on Build GRCh38

View Full Product Details