Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238233339-238233906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC643387
Entrez Gene ID
643387
Gene Name
TAR DNA binding protein pseudogene
Gene Aliases
-
UniGene
Hs.598359
Gene Chromosome Location
Chr.2: 238231686-238234345 on Build GRCh38
Transcripts
NR_026923.1

SNP Detail

SNP ID Chromosome Location
rs181474257 Chr.2: 238233693-238233693 on Build GRCh38
rs533246802 Chr.2: 238233565-238233565 on Build GRCh38
rs758367245 Chr.2: 238233604-238233604 on Build GRCh38
rs56200201 Chr.2: 238233815-238233815 on Build GRCh38
rs115877723 Chr.2: 238233601-238233601 on Build GRCh38
rs573858829 Chr.2: 238233883-238233883 on Build GRCh38
rs1018079403 Chr.2: 238233873-238233873 on Build GRCh38
rs550572404 Chr.2: 238233642-238233642 on Build GRCh38
rs969499503 Chr.2: 238233520-238233520 on Build GRCh38
rs900802906 Chr.2: 238233415-238233415 on Build GRCh38
rs147992554 Chr.2: 238233603-238233603 on Build GRCh38
rs148911096 Chr.2: 238233464-238233464 on Build GRCh38
rs557975675 Chr.2: 238233421-238233421 on Build GRCh38
rs573920381 Chr.2: 238233497-238233497 on Build GRCh38
rs993493524 Chr.2: 238233819-238233819 on Build GRCh38
rs370697687 Chr.2: 238233631-238233631 on Build GRCh38
rs942442217 Chr.2: 238233366-238233366 on Build GRCh38
rs534008898 Chr.2: 238233827-238233827 on Build GRCh38
rs185454690 Chr.2: 238233613-238233613 on Build GRCh38
rs141681464 Chr.2: 238233609-238233609 on Build GRCh38
rs11696058 Chr.2: 238233538-238233538 on Build GRCh38
rs190706466 Chr.2: 238233617-238233617 on Build GRCh38
rs894099139 Chr.2: 238233684-238233684 on Build GRCh38
rs935019277 Chr.2: 238233678-238233678 on Build GRCh38
rs1052091296 Chr.2: 238233683-238233683 on Build GRCh38
rs370978719 Chr.2: 238233491-238233491 on Build GRCh38
rs138733187 Chr.2: 238233692-238233692 on Build GRCh38
rs1045265531 Chr.2: 238233706-238233706 on Build GRCh38
rs1011112136 Chr.2: 238233475-238233475 on Build GRCh38
rs539567843 Chr.2: 238233370-238233370 on Build GRCh38
rs1023804065 Chr.2: 238233484-238233484 on Build GRCh38
rs962329981 Chr.2: 238233553-238233553 on Build GRCh38
rs1003648671 Chr.2: 238233769-238233769 on Build GRCh38
rs555310676 Chr.2: 238233839-238233839 on Build GRCh38
rs143438655 Chr.2: 238233524-238233524 on Build GRCh38
rs573003700 Chr.2: 238233444-238233444 on Build GRCh38
rs983971104 Chr.2: 238233840-238233840 on Build GRCh38
rs55808641 Chr.2: 238233639-238233639 on Build GRCh38
rs1035536289 Chr.2: 238233814-238233814 on Build GRCh38
rs780316242 Chr.2: 238233633-238233633 on Build GRCh38
rs143175112 Chr.2: 238233729-238233729 on Build GRCh38
rs11692007 Chr.2: 238233447-238233447 on Build GRCh38
rs751243422 Chr.2: 238233668-238233668 on Build GRCh38
rs1027610373 Chr.2: 238233826-238233826 on Build GRCh38
rs1052666513 Chr.2: 238233453-238233453 on Build GRCh38

View Full Product Details