Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16671374-16671755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD62P1-PARP4P3
Entrez Gene ID
23783
Gene Name
ANKRD62P1-PARP4P3 readthrough, transcribed pseudogene
Gene Aliases
VWFP1-ANKRD62P1-PARP4P3
UniGene
Hs.627452
Gene Chromosome Location
Chr.22: 16653708-16675540 on Build GRCh38
Transcripts
NR_040115.1

SNP Detail

SNP ID Chromosome Location
rs745377838 Chr.22: 16671517-16671517 on Build GRCh38
rs1036059520 Chr.22: 16671727-16671727 on Build GRCh38
rs956844575 Chr.22: 16671589-16671589 on Build GRCh38
rs553541822 Chr.22: 16671658-16671658 on Build GRCh38
rs906248797 Chr.22: 16671469-16671469 on Build GRCh38
rs578011155 Chr.22: 16671573-16671573 on Build GRCh38
rs924076702 Chr.22: 16671578-16671578 on Build GRCh38
rs1006883059 Chr.22: 16671566-16671566 on Build GRCh38
rs4819849 Chr.22: 16671721-16671721 on Build GRCh38
rs1003251238 Chr.22: 16671483-16671483 on Build GRCh38
rs571736920 Chr.22: 16671695-16671695 on Build GRCh38
rs552386962 Chr.22: 16671484-16671484 on Build GRCh38
rs558091708 Chr.22: 16671505-16671505 on Build GRCh38
rs781211309 Chr.22: 16671468-16671469 on Build GRCh38
rs989686549 Chr.22: 16671632-16671632 on Build GRCh38
rs994661274 Chr.22: 16671504-16671504 on Build GRCh38
rs758468834 Chr.22: 16671706-16671706 on Build GRCh38
rs368477011 Chr.22: 16671567-16671567 on Build GRCh38
rs897678519 Chr.22: 16671491-16671491 on Build GRCh38
rs117963034 Chr.22: 16671572-16671572 on Build GRCh38
rs754279395 Chr.22: 16671453-16671453 on Build GRCh38
rs1027512453 Chr.22: 16671548-16671548 on Build GRCh38
rs953456636 Chr.22: 16671553-16671554 on Build GRCh38
rs948313276 Chr.22: 16671655-16671655 on Build GRCh38
rs915487851 Chr.22: 16671644-16671644 on Build GRCh38
rs530757220 Chr.22: 16671461-16671461 on Build GRCh38

View Full Product Details