Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 75664658-75665205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRG2C
Entrez Gene ID
100288801
Gene Name
FSHD region gene 2 family member C
Gene Aliases
-
UniGene
Hs.274541
Gene Chromosome Location
Chr.3: 75646761-75667217 on Build GRCh38
Transcripts
NM_001124759.1 XM_011533254.2 XM_011533256.2 XM_017005489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4158331 c.291G>A p.R97R Chr.3: 75665160-75665160 on Build GRCh38
1618045 c.179-7C>T p.? Chr.3: 75664812-75664812 on Build GRCh38
4158330 c.219A>G p.E73E Chr.3: 75664859-75664859 on Build GRCh38
1666704 c.250G>A p.G84R Chr.3: 75664890-75664890 on Build GRCh38
4158332 c.325G>T p.D109Y Chr.3: 75665194-75665194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71244678 Chr.3: 75664890-75664890 on Build GRCh38
rs12107205 Chr.3: 75664716-75664716 on Build GRCh38
rs77905349 Chr.3: 75664826-75664826 on Build GRCh38
rs376221321 Chr.3: 75664762-75664762 on Build GRCh38
rs377681226 Chr.3: 75664825-75664825 on Build GRCh38
rs76698561 Chr.3: 75665112-75665112 on Build GRCh38
rs59911589 Chr.3: 75665035-75665035 on Build GRCh38
rs763120246 Chr.3: 75664873-75664873 on Build GRCh38
rs74528535 Chr.3: 75664758-75664758 on Build GRCh38
rs79560234 Chr.3: 75664752-75664752 on Build GRCh38
rs75914959 Chr.3: 75664988-75664988 on Build GRCh38
rs73840321 Chr.3: 75664944-75664944 on Build GRCh38
rs76139301 Chr.3: 75665091-75665091 on Build GRCh38
rs267599932 Chr.3: 75665149-75665149 on Build GRCh38
rs374393526 Chr.3: 75665006-75665006 on Build GRCh38
rs77194044 Chr.3: 75664728-75664728 on Build GRCh38
rs78181421 Chr.3: 75664807-75664807 on Build GRCh38
rs79850029 Chr.3: 75665160-75665160 on Build GRCh38
rs75415205 Chr.3: 75665100-75665100 on Build GRCh38
rs76161015 Chr.3: 75664746-75664746 on Build GRCh38
rs12106841 Chr.3: 75664707-75664707 on Build GRCh38
rs10451970 Chr.3: 75665080-75665080 on Build GRCh38
rs866929828 Chr.3: 75665174-75665174 on Build GRCh38
rs74714110 Chr.3: 75665147-75665147 on Build GRCh38
rs74458795 Chr.3: 75664804-75664804 on Build GRCh38
rs78097733 Chr.3: 75664829-75664829 on Build GRCh38
rs72503534 Chr.3: 75664951-75664951 on Build GRCh38
rs77258640 Chr.3: 75665051-75665051 on Build GRCh38
rs73840319 Chr.3: 75664679-75664679 on Build GRCh38
rs80018434 Chr.3: 75664976-75664976 on Build GRCh38
rs6781177 Chr.3: 75664797-75664797 on Build GRCh38
rs76950474 Chr.3: 75665113-75665113 on Build GRCh38
rs73840322 Chr.3: 75665034-75665034 on Build GRCh38
rs75913622 Chr.3: 75664967-75664967 on Build GRCh38
rs373472815 Chr.3: 75664859-75664859 on Build GRCh38
rs71315304 Chr.3: 75664995-75664995 on Build GRCh38
rs75456211 Chr.3: 75664883-75664883 on Build GRCh38
rs374465172 Chr.3: 75664753-75664753 on Build GRCh38
rs75017289 Chr.3: 75664812-75664812 on Build GRCh38
rs571779240 Chr.3: 75664978-75664978 on Build GRCh38

View Full Product Details