Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70078647-70079200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN1S2AP
Entrez Gene ID
286828
Gene Name
casein alpha s2-like A, pseudogene
Gene Aliases
CSN1S2A
UniGene
Hs.631945
Gene Chromosome Location
Chr.4: 70067386-70085273 on Build GRCh38
Transcripts
NR_003720.1

SNP Detail

SNP ID Chromosome Location
rs769168120 Chr.4: 70078952-70078952 on Build GRCh38
rs944938194 Chr.4: 70078730-70078730 on Build GRCh38
rs557697368 Chr.4: 70078820-70078820 on Build GRCh38
rs761895123 Chr.4: 70079019-70079019 on Build GRCh38
rs758639852 Chr.4: 70078770-70078770 on Build GRCh38
rs1056166420 Chr.4: 70079165-70079165 on Build GRCh38
rs372624671 Chr.4: 70079154-70079154 on Build GRCh38
rs912768100 Chr.4: 70078863-70078863 on Build GRCh38
rs550183446 Chr.4: 70078991-70078992 on Build GRCh38
rs903783792 Chr.4: 70079023-70079023 on Build GRCh38
rs570427075 Chr.4: 70078915-70078915 on Build GRCh38
rs190077176 Chr.4: 70079107-70079107 on Build GRCh38
rs117168764 Chr.4: 70079164-70079164 on Build GRCh38
rs183852940 Chr.4: 70079146-70079146 on Build GRCh38
rs72651140 Chr.4: 70078992-70078992 on Build GRCh38
rs972908400 Chr.4: 70079077-70079077 on Build GRCh38
rs1048737873 Chr.4: 70078948-70078948 on Build GRCh38
rs1399812 Chr.4: 70078735-70078735 on Build GRCh38
rs961564726 Chr.4: 70079059-70079059 on Build GRCh38
rs1019090433 Chr.4: 70078950-70078950 on Build GRCh38
rs1028330253 Chr.4: 70079028-70079028 on Build GRCh38
rs565959563 Chr.4: 70078728-70078728 on Build GRCh38
rs532104425 Chr.4: 70078689-70078689 on Build GRCh38
rs146851805 Chr.4: 70078803-70078803 on Build GRCh38
rs1020826277 Chr.4: 70078780-70078780 on Build GRCh38
rs755888448 Chr.4: 70078928-70078928 on Build GRCh38
rs568527899 Chr.4: 70079035-70079035 on Build GRCh38
rs973003078 Chr.4: 70078893-70078893 on Build GRCh38
rs937770412 Chr.4: 70078958-70078958 on Build GRCh38
rs181222045 Chr.4: 70078914-70078914 on Build GRCh38
rs749598854 Chr.4: 70078930-70078930 on Build GRCh38
rs755180707 Chr.4: 70078718-70078718 on Build GRCh38
rs947865151 Chr.4: 70079114-70079114 on Build GRCh38
rs868389269 Chr.4: 70079136-70079136 on Build GRCh38
rs10030699 Chr.4: 70079100-70079100 on Build GRCh38
rs776543189 Chr.4: 70079024-70079027 on Build GRCh38
rs185316752 Chr.4: 70079037-70079037 on Build GRCh38
rs986845904 Chr.4: 70078824-70078824 on Build GRCh38
rs912056022 Chr.4: 70078729-70078729 on Build GRCh38
rs921585197 Chr.4: 70078760-70078760 on Build GRCh38
rs527486396 Chr.4: 70078879-70078879 on Build GRCh38
rs571268473 Chr.4: 70078880-70078880 on Build GRCh38
rs1033040902 Chr.4: 70078696-70078696 on Build GRCh38
rs746712477 Chr.4: 70078789-70078790 on Build GRCh38
rs1020858949 Chr.4: 70079012-70079012 on Build GRCh38
rs116132032 Chr.4: 70078783-70078783 on Build GRCh38
rs1001169763 Chr.4: 70079025-70079025 on Build GRCh38
rs896228266 Chr.4: 70079176-70079176 on Build GRCh38
rs1023958396 Chr.4: 70078888-70078888 on Build GRCh38
rs1019690476 Chr.4: 70079049-70079049 on Build GRCh38
rs979444753 Chr.4: 70078786-70078786 on Build GRCh38
rs7695886 Chr.4: 70079095-70079095 on Build GRCh38
rs940166841 Chr.4: 70078877-70078877 on Build GRCh38
rs556309349 Chr.4: 70079005-70079005 on Build GRCh38
rs558456138 Chr.4: 70079115-70079115 on Build GRCh38
rs896363690 Chr.4: 70078998-70078998 on Build GRCh38
rs903795183 Chr.4: 70078889-70078889 on Build GRCh38
rs1044946469 Chr.4: 70079149-70079149 on Build GRCh38
rs907539513 Chr.4: 70079065-70079065 on Build GRCh38

View Full Product Details