Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111046914-111047462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTM2P1
Entrez Gene ID
442245
Gene Name
glutathione S-transferase mu 2 pseudogene 1
Gene Aliases
-
UniGene
Hs.551627
Gene Chromosome Location
Chr.6: 111046419-111047554 on Build GRCh38
Transcripts
NR_002932.2

SNP Detail

SNP ID Chromosome Location
rs923831613 Chr.6: 111047293-111047293 on Build GRCh38
rs886910355 Chr.6: 111047400-111047400 on Build GRCh38
rs879036231 Chr.6: 111046988-111046988 on Build GRCh38
rs749267644 Chr.6: 111047342-111047342 on Build GRCh38
rs765907530 Chr.6: 111047310-111047310 on Build GRCh38
rs761755876 Chr.6: 111047011-111047011 on Build GRCh38
rs1140964 Chr.6: 111047137-111047137 on Build GRCh38
rs191697463 Chr.6: 111047334-111047334 on Build GRCh38
rs745936149 Chr.6: 111047171-111047171 on Build GRCh38
rs1002756098 Chr.6: 111047164-111047164 on Build GRCh38
rs750003815 Chr.6: 111047217-111047217 on Build GRCh38
rs776440836 Chr.6: 111047002-111047002 on Build GRCh38
rs779679014 Chr.6: 111047101-111047101 on Build GRCh38
rs186868520 Chr.6: 111047253-111047253 on Build GRCh38
rs780893065 Chr.6: 111047246-111047246 on Build GRCh38
rs139244562 Chr.6: 111046979-111046979 on Build GRCh38
rs777381095 Chr.6: 111047339-111047339 on Build GRCh38
rs879049145 Chr.6: 111047438-111047438 on Build GRCh38
rs746600476 Chr.6: 111047119-111047119 on Build GRCh38
rs754640690 Chr.6: 111047321-111047321 on Build GRCh38
rs182113736 Chr.6: 111047366-111047366 on Build GRCh38
rs1022117179 Chr.6: 111047240-111047240 on Build GRCh38
rs1039836926 Chr.6: 111047028-111047028 on Build GRCh38
rs754302952 Chr.6: 111046963-111046963 on Build GRCh38
rs963201046 Chr.6: 111047158-111047158 on Build GRCh38
rs1056403852 Chr.6: 111047082-111047082 on Build GRCh38
rs776919194 Chr.6: 111047280-111047280 on Build GRCh38
rs201657335 Chr.6: 111046954-111046954 on Build GRCh38
rs773745182 Chr.6: 111047378-111047378 on Build GRCh38
rs2229048 Chr.6: 111046994-111046994 on Build GRCh38
rs752232283 Chr.6: 111047410-111047410 on Build GRCh38
rs878914286 Chr.6: 111047421-111047421 on Build GRCh38
rs555520208 Chr.6: 111047367-111047367 on Build GRCh38
rs754589008 Chr.6: 111047241-111047241 on Build GRCh38
rs1056795 Chr.6: 111047133-111047133 on Build GRCh38
rs770309737 Chr.6: 111047376-111047376 on Build GRCh38
rs762885088 Chr.6: 111047419-111047427 on Build GRCh38
rs770942540 Chr.6: 111047013-111047013 on Build GRCh38
rs752221656 Chr.6: 111047329-111047329 on Build GRCh38
rs765762467 Chr.6: 111047216-111047216 on Build GRCh38
rs774454083 Chr.6: 111047016-111047016 on Build GRCh38
rs1140963 Chr.6: 111047140-111047140 on Build GRCh38
rs140098562 Chr.6: 111047416-111047416 on Build GRCh38
rs749927625 Chr.6: 111047089-111047089 on Build GRCh38
rs879185936 Chr.6: 111047286-111047286 on Build GRCh38
rs763403543 Chr.6: 111047309-111047309 on Build GRCh38
rs556123247 Chr.6: 111047322-111047322 on Build GRCh38
rs77870449 Chr.6: 111047268-111047268 on Build GRCh38
rs1140961 Chr.6: 111047165-111047165 on Build GRCh38
rs73763501 Chr.6: 111047257-111047257 on Build GRCh38
rs946606403 Chr.6: 111047024-111047024 on Build GRCh38
rs569100272 Chr.6: 111047127-111047127 on Build GRCh38
rs777767700 Chr.6: 111047260-111047260 on Build GRCh38
rs919666868 Chr.6: 111047346-111047346 on Build GRCh38
rs753849415 Chr.6: 111047426-111047426 on Build GRCh38
rs746789380 Chr.6: 111047198-111047198 on Build GRCh38
rs541933986 Chr.6: 111046962-111046962 on Build GRCh38
rs2227958 Chr.6: 111046961-111046961 on Build GRCh38
rs539678053 Chr.6: 111047316-111047316 on Build GRCh38
rs771303772 Chr.6: 111047385-111047385 on Build GRCh38
rs757909325 Chr.6: 111047221-111047221 on Build GRCh38
rs1140962 Chr.6: 111047150-111047150 on Build GRCh38
rs763054073 Chr.6: 111047344-111047344 on Build GRCh38
rs756729427 Chr.6: 111046971-111046971 on Build GRCh38
rs763261113 Chr.6: 111047379-111047379 on Build GRCh38
rs751238330 Chr.6: 111047231-111047231 on Build GRCh38
rs767136283 Chr.6: 111047407-111047407 on Build GRCh38
rs772209048 Chr.6: 111047181-111047181 on Build GRCh38
rs560643831 Chr.6: 111047042-111047042 on Build GRCh38
rs113020139 Chr.6: 111047041-111047041 on Build GRCh38
rs537996384 Chr.6: 111047187-111047187 on Build GRCh38
rs759225982 Chr.6: 111047396-111047396 on Build GRCh38
rs773724487 Chr.6: 111047284-111047284 on Build GRCh38
rs968867831 Chr.6: 111047244-111047244 on Build GRCh38
rs765455868 Chr.6: 111047283-111047283 on Build GRCh38
rs112652573 Chr.6: 111047055-111047055 on Build GRCh38
rs758105341 Chr.6: 111047094-111047094 on Build GRCh38
rs572576013 Chr.6: 111047096-111047096 on Build GRCh38
rs772298894 Chr.6: 111047262-111047262 on Build GRCh38
rs553974306 Chr.6: 111046999-111046999 on Build GRCh38
rs753175834 Chr.6: 111047044-111047044 on Build GRCh38
rs548253499 Chr.6: 111047197-111047197 on Build GRCh38
rs577063905 Chr.6: 111047281-111047281 on Build GRCh38
rs746794954 Chr.6: 111046995-111046995 on Build GRCh38
rs745538612 Chr.6: 111046984-111046984 on Build GRCh38
rs766613209 Chr.6: 111047411-111047411 on Build GRCh38
rs769630718 Chr.6: 111046997-111047028 on Build GRCh38
rs775482298 Chr.6: 111047239-111047241 on Build GRCh38
rs768900344 Chr.6: 111047270-111047270 on Build GRCh38
rs765820699 Chr.6: 111047229-111047229 on Build GRCh38
rs994347537 Chr.6: 111047194-111047194 on Build GRCh38
rs775865827 Chr.6: 111047188-111047188 on Build GRCh38
rs954883873 Chr.6: 111047311-111047311 on Build GRCh38
rs879068013 Chr.6: 111047300-111047300 on Build GRCh38
rs757741179 Chr.6: 111047083-111047083 on Build GRCh38
rs770039810 Chr.6: 111047233-111047233 on Build GRCh38
rs879109980 Chr.6: 111047290-111047290 on Build GRCh38
rs933614686 Chr.6: 111046972-111046972 on Build GRCh38
rs532794922 Chr.6: 111047045-111047045 on Build GRCh38
rs757228949 Chr.6: 111047343-111047343 on Build GRCh38
rs754322029 Chr.6: 111047061-111047061 on Build GRCh38
rs773930963 Chr.6: 111047386-111047386 on Build GRCh38
rs745857166 Chr.6: 111046993-111046993 on Build GRCh38
rs768455708 Chr.6: 111047124-111047124 on Build GRCh38
rs757061241 Chr.6: 111047429-111047429 on Build GRCh38
rs775669671 Chr.6: 111047267-111047267 on Build GRCh38
rs774349037 Chr.6: 111047153-111047153 on Build GRCh38
rs748232367 Chr.6: 111047369-111047369 on Build GRCh38
rs778326217 Chr.6: 111047085-111047085 on Build GRCh38
rs2229049 Chr.6: 111046982-111046982 on Build GRCh38
rs767834935 Chr.6: 111047043-111047043 on Build GRCh38
rs879203229 Chr.6: 111047402-111047402 on Build GRCh38
rs942353085 Chr.6: 111047330-111047330 on Build GRCh38

View Full Product Details