Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15068134-15068611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDXDC1
Entrez Gene ID
23042
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Gene Aliases
LP8165
UniGene
Hs.370781
Gene Chromosome Location
Chr.16: 14974591-15139410 on Build GRCh38
Transcripts
NM_001285449.1 NM_001324020.1 NM_001324021.1 XM_017023065.1 NM_001301064.1 NM_018427.4 XM_005255376.1 XM_005255377.4 XM_011522540.2 XM_011522541.1

Gene Symbol
RRN3
Entrez Gene ID
54700
Gene Name
RRN3 homolog, RNA polymerase I transcription factor
Gene Aliases
A-270G1.2, TIFIA
UniGene
Hs.460078
Gene Chromosome Location
Chr.16: 15060022-15094335 on Build GRCh38
Transcripts
NM_001285449.1 NM_001324020.1 NM_001324021.1 XM_017023065.1 NM_001301064.1 NM_018427.4 XM_005255376.1 XM_005255377.4 XM_011522540.2 XM_011522541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5794199 c.1553+7T>C p.? Chr.16: 15068162-15068162 on Build GRCh38
3817249 c.1524A>G p.S508S Chr.16: 15068198-15068198 on Build GRCh38
4256044 c.1457T>C p.L486P Chr.16: 15068265-15068265 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766067058 Chr.16: 15068161-15068161 on Build GRCh38
rs762773216 Chr.16: 15068273-15068273 on Build GRCh38
rs752190884 Chr.16: 15068176-15068176 on Build GRCh38
rs942312881 Chr.16: 15068551-15068552 on Build GRCh38
rs780175717 Chr.16: 15068229-15068229 on Build GRCh38
rs368582942 Chr.16: 15068286-15068287 on Build GRCh38
rs759233703 Chr.16: 15068299-15068299 on Build GRCh38
rs1048048814 Chr.16: 15068301-15068301 on Build GRCh38
rs774336480 Chr.16: 15068293-15068293 on Build GRCh38
rs1030591474 Chr.16: 15068368-15068368 on Build GRCh38
rs888220624 Chr.16: 15068318-15068318 on Build GRCh38
rs757649459 Chr.16: 15068200-15068200 on Build GRCh38
rs759068836 Chr.16: 15068165-15068165 on Build GRCh38
rs997652698 Chr.16: 15068367-15068368 on Build GRCh38
rs184986413 Chr.16: 15068339-15068339 on Build GRCh38
rs999969068 Chr.16: 15068424-15068424 on Build GRCh38
rs949199841 Chr.16: 15068205-15068205 on Build GRCh38
rs11075254 Chr.16: 15068494-15068494 on Build GRCh38
rs769899106 Chr.16: 15068287-15068287 on Build GRCh38
rs181406540 Chr.16: 15068245-15068245 on Build GRCh38
rs770876202 Chr.16: 15068281-15068281 on Build GRCh38
rs1048559448 Chr.16: 15068535-15068535 on Build GRCh38
rs564073678 Chr.16: 15068270-15068270 on Build GRCh38
rs150321506 Chr.16: 15068365-15068365 on Build GRCh38
rs774788503 Chr.16: 15068301-15068302 on Build GRCh38
rs549801559 Chr.16: 15068554-15068554 on Build GRCh38
rs773031919 Chr.16: 15068265-15068265 on Build GRCh38
rs759988993 Chr.16: 15068314-15068315 on Build GRCh38
rs537591201 Chr.16: 15068475-15068475 on Build GRCh38
rs754163798 Chr.16: 15068199-15068199 on Build GRCh38
rs772317742 Chr.16: 15068216-15068216 on Build GRCh38
rs752606451 Chr.16: 15068482-15068482 on Build GRCh38
rs767114725 Chr.16: 15068166-15068166 on Build GRCh38
rs1046304979 Chr.16: 15068253-15068253 on Build GRCh38
rs966985240 Chr.16: 15068379-15068379 on Build GRCh38
rs918531057 Chr.16: 15068510-15068510 on Build GRCh38
rs376236304 Chr.16: 15068502-15068502 on Build GRCh38
rs762673471 Chr.16: 15068157-15068157 on Build GRCh38
rs929894665 Chr.16: 15068517-15068517 on Build GRCh38
rs573574650 Chr.16: 15068473-15068473 on Build GRCh38
rs778154313 Chr.16: 15068198-15068198 on Build GRCh38
rs916636938 Chr.16: 15068490-15068490 on Build GRCh38
rs746870174 Chr.16: 15068287-15068288 on Build GRCh38
rs751293013 Chr.16: 15068164-15068164 on Build GRCh38
rs909567294 Chr.16: 15068549-15068549 on Build GRCh38
rs759754499 Chr.16: 15068331-15068332 on Build GRCh38
rs769716727 Chr.16: 15068260-15068260 on Build GRCh38
rs756641855 Chr.16: 15068183-15068183 on Build GRCh38
rs1039481659 Chr.16: 15068551-15068551 on Build GRCh38
rs779057571 Chr.16: 15068201-15068201 on Build GRCh38
rs771013931 Chr.16: 15068287-15068289 on Build GRCh38
rs747114200 Chr.16: 15068244-15068244 on Build GRCh38
rs377467199 Chr.16: 15068203-15068203 on Build GRCh38
rs1039448473 Chr.16: 15068357-15068357 on Build GRCh38
rs9924644 Chr.16: 15068189-15068189 on Build GRCh38
rs1032333686 Chr.16: 15068428-15068429 on Build GRCh38
rs555106796 Chr.16: 15068437-15068437 on Build GRCh38
rs990884105 Chr.16: 15068486-15068486 on Build GRCh38

View Full Product Details