Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19464080-19464655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPTE2
Entrez Gene ID
93492
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
TPIP
UniGene
Hs.377488
Gene Chromosome Location
Chr.13: 19422877-19561574 on Build GRCh38
Transcripts
NM_001141968.1 NM_130785.3 NM_199254.2 NR_073485.1 NR_073486.1 NR_073487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4213374 c.677-3C>T p.? Chr.13: 19464523-19464523 on Build GRCh38
7253134 c.497G>T p.R166I Chr.13: 19464469-19464469 on Build GRCh38
3467559 c.502C>T p.P168S Chr.13: 19464464-19464464 on Build GRCh38
4506962 c.734C>T p.P245L Chr.13: 19464463-19464463 on Build GRCh38
1365688 c.734C>A p.P245Q Chr.13: 19464463-19464463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145515595 Chr.13: 19464480-19464480 on Build GRCh38
rs573484696 Chr.13: 19464615-19464615 on Build GRCh38
rs563616541 Chr.13: 19464436-19464436 on Build GRCh38
rs991478950 Chr.13: 19464208-19464208 on Build GRCh38
rs575918607 Chr.13: 19464574-19464574 on Build GRCh38
rs757568707 Chr.13: 19464493-19464493 on Build GRCh38
rs997895030 Chr.13: 19464333-19464334 on Build GRCh38
rs565069440 Chr.13: 19464572-19464572 on Build GRCh38
rs775039828 Chr.13: 19464567-19464567 on Build GRCh38
rs74575548 Chr.13: 19464337-19464337 on Build GRCh38
rs542762555 Chr.13: 19464396-19464396 on Build GRCh38
rs778149817 Chr.13: 19464409-19464409 on Build GRCh38
rs774436949 Chr.13: 19464558-19464558 on Build GRCh38
rs74983036 Chr.13: 19464341-19464341 on Build GRCh38
rs1010672605 Chr.13: 19464399-19464399 on Build GRCh38
rs540134036 Chr.13: 19464583-19464583 on Build GRCh38
rs751893847 Chr.13: 19464484-19464484 on Build GRCh38
rs939759009 Chr.13: 19464317-19464317 on Build GRCh38
rs34800894 Chr.13: 19464575-19464575 on Build GRCh38
rs190202540 Chr.13: 19464331-19464331 on Build GRCh38
rs182752354 Chr.13: 19464433-19464433 on Build GRCh38
rs546451930 Chr.13: 19464324-19464324 on Build GRCh38
rs530741541 Chr.13: 19464616-19464617 on Build GRCh38
rs747467707 Chr.13: 19464412-19464412 on Build GRCh38
rs770652243 Chr.13: 19464560-19464560 on Build GRCh38
rs756821483 Chr.13: 19464544-19464545 on Build GRCh38
rs557059633 Chr.13: 19464561-19464561 on Build GRCh38
rs771525966 Chr.13: 19464413-19464413 on Build GRCh38
rs759035513 Chr.13: 19464461-19464461 on Build GRCh38
rs398021738 Chr.13: 19464582-19464582 on Build GRCh38
rs916866232 Chr.13: 19464222-19464222 on Build GRCh38
rs759508913 Chr.13: 19464564-19464564 on Build GRCh38
rs762465046 Chr.13: 19464569-19464569 on Build GRCh38
rs879581283 Chr.13: 19464629-19464629 on Build GRCh38
rs551982045 Chr.13: 19464472-19464472 on Build GRCh38
rs370979800 Chr.13: 19464441-19464441 on Build GRCh38
rs1022319378 Chr.13: 19464129-19464129 on Build GRCh38
rs753748085 Chr.13: 19464460-19464460 on Build GRCh38
rs889673617 Chr.13: 19464612-19464612 on Build GRCh38
rs201125226 Chr.13: 19464474-19464474 on Build GRCh38
rs766269272 Chr.13: 19464454-19464454 on Build GRCh38
rs758738499 Chr.13: 19464405-19464405 on Build GRCh38
rs1049274318 Chr.13: 19464278-19464278 on Build GRCh38
rs764721330 Chr.13: 19464566-19464566 on Build GRCh38
rs139121187 Chr.13: 19464482-19464482 on Build GRCh38
rs574769623 Chr.13: 19464195-19464195 on Build GRCh38
rs1022098392 Chr.13: 19464410-19464410 on Build GRCh38
rs537955930 Chr.13: 19464138-19464138 on Build GRCh38
rs760397535 Chr.13: 19464453-19464453 on Build GRCh38
rs548128494 Chr.13: 19464575-19464576 on Build GRCh38
rs113739829 Chr.13: 19464429-19464429 on Build GRCh38
rs557654438 Chr.13: 19464231-19464231 on Build GRCh38
rs7339103 Chr.13: 19464444-19464444 on Build GRCh38
rs558429136 Chr.13: 19464585-19464585 on Build GRCh38
rs909602141 Chr.13: 19464304-19464304 on Build GRCh38
rs148045725 Chr.13: 19464518-19464518 on Build GRCh38
rs1050094924 Chr.13: 19464376-19464376 on Build GRCh38
rs1049424180 Chr.13: 19464584-19464584 on Build GRCh38
rs946643748 Chr.13: 19464617-19464617 on Build GRCh38
rs34940161 Chr.13: 19464289-19464290 on Build GRCh38
rs760616751 Chr.13: 19464463-19464463 on Build GRCh38
rs374344080 Chr.13: 19464422-19464422 on Build GRCh38
rs780324300 Chr.13: 19464535-19464535 on Build GRCh38
rs764774519 Chr.13: 19464465-19464465 on Build GRCh38
rs2497247 Chr.13: 19464147-19464147 on Build GRCh38
rs111718378 Chr.13: 19464438-19464438 on Build GRCh38
rs115226626 Chr.13: 19464190-19464190 on Build GRCh38
rs564644944 Chr.13: 19464329-19464329 on Build GRCh38
rs568416507 Chr.13: 19464517-19464517 on Build GRCh38
rs749521463 Chr.13: 19464536-19464536 on Build GRCh38
rs559143693 Chr.13: 19464355-19464355 on Build GRCh38
rs143491494 Chr.13: 19464506-19464506 on Build GRCh38
rs781564574 Chr.13: 19464414-19464414 on Build GRCh38
rs560389507 Chr.13: 19464276-19464276 on Build GRCh38
rs768937901 Chr.13: 19464552-19464552 on Build GRCh38
rs76728921 Chr.13: 19464228-19464228 on Build GRCh38
rs927979560 Chr.13: 19464183-19464183 on Build GRCh38

View Full Product Details