Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169837367-169837673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRIQ4
Entrez Gene ID
344657
Gene Name
leucine rich repeats and IQ motif containing 4
Gene Aliases
LRRC64
UniGene
Hs.319484
Gene Chromosome Location
Chr.3: 169812947-169838007 on Build GRCh38
Transcripts
NM_001080460.1 XM_005247421.2 XM_006713613.3 XM_011512748.1 XM_011512749.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3203893 c.1576A>G p.K526E Chr.3: 169837524-169837524 on Build GRCh38
3589966 c.1645G>A p.D549N Chr.3: 169837593-169837593 on Build GRCh38
261853 c.1605G>A p.K535K Chr.3: 169837553-169837553 on Build GRCh38
3589965 c.1537G>A p.A513T Chr.3: 169837485-169837485 on Build GRCh38
3203892 c.1546C>T p.R516C Chr.3: 169837494-169837494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755826280 Chr.3: 169837533-169837533 on Build GRCh38
rs952950350 Chr.3: 169837404-169837404 on Build GRCh38
rs751270052 Chr.3: 169837502-169837502 on Build GRCh38
rs746160133 Chr.3: 169837567-169837567 on Build GRCh38
rs549292022 Chr.3: 169837447-169837447 on Build GRCh38
rs369016431 Chr.3: 169837529-169837529 on Build GRCh38
rs775206967 Chr.3: 169837510-169837510 on Build GRCh38
rs766529627 Chr.3: 169837505-169837506 on Build GRCh38
rs752990886 Chr.3: 169837628-169837628 on Build GRCh38
rs751681194 Chr.3: 169837564-169837564 on Build GRCh38
rs375837621 Chr.3: 169837501-169837501 on Build GRCh38
rs757666982 Chr.3: 169837563-169837563 on Build GRCh38
rs1045511330 Chr.3: 169837413-169837413 on Build GRCh38
rs750712735 Chr.3: 169837500-169837501 on Build GRCh38
rs531424559 Chr.3: 169837622-169837622 on Build GRCh38
rs534963520 Chr.3: 169837495-169837495 on Build GRCh38
rs112011297 Chr.3: 169837487-169837487 on Build GRCh38
rs200900869 Chr.3: 169837485-169837485 on Build GRCh38
rs772569175 Chr.3: 169837579-169837579 on Build GRCh38
rs781740274 Chr.3: 169837618-169837619 on Build GRCh38
rs779498004 Chr.3: 169837564-169837564 on Build GRCh38
rs754972775 Chr.3: 169837603-169837604 on Build GRCh38
rs768852074 Chr.3: 169837494-169837494 on Build GRCh38
rs753428359 Chr.3: 169837543-169837543 on Build GRCh38
rs771517619 Chr.3: 169837435-169837435 on Build GRCh38
rs770719650 Chr.3: 169837647-169837647 on Build GRCh38
rs369857553 Chr.3: 169837633-169837633 on Build GRCh38
rs774352137 Chr.3: 169837649-169837649 on Build GRCh38
rs201862263 Chr.3: 169837624-169837624 on Build GRCh38
rs746277055 Chr.3: 169837456-169837456 on Build GRCh38
rs763567273 Chr.3: 169837407-169837407 on Build GRCh38
rs375552536 Chr.3: 169837448-169837448 on Build GRCh38
rs1013943861 Chr.3: 169837641-169837641 on Build GRCh38
rs202034195 Chr.3: 169837530-169837530 on Build GRCh38
rs926849283 Chr.3: 169837391-169837391 on Build GRCh38
rs780630856 Chr.3: 169837585-169837585 on Build GRCh38
rs747217051 Chr.3: 169837606-169837606 on Build GRCh38
rs758622006 Chr.3: 169837503-169837504 on Build GRCh38
rs759343405 Chr.3: 169837509-169837509 on Build GRCh38
rs78013694 Chr.3: 169837608-169837608 on Build GRCh38
rs764104763 Chr.3: 169837443-169837443 on Build GRCh38

View Full Product Details