Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24533002-24533331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCAN3
Entrez Gene ID
11123
Gene Name
RCAN family member 3
Gene Aliases
DSCR1L2, MCIP3, RCN3, hRCN3
UniGene
Hs.656799
Gene Chromosome Location
Chr.1: 24502351-24537020 on Build GRCh38
Transcripts
NM_001251977.1 NM_001251978.1 NM_001251979.1 NM_001251980.1 NM_001251981.1 NM_001251982.1 NM_001251983.1 NM_001251984.1 NM_001251985.1 NM_013441.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746902 c.414G>A p.P138P Chr.1: 24533127-24533127 on Build GRCh38
2154120 c.461C>A p.P154Q Chr.1: 24533174-24533174 on Build GRCh38
6054425 c.477G>C p.Q159H Chr.1: 24533190-24533190 on Build GRCh38
3746903 c.462G>A p.P154P Chr.1: 24533175-24533175 on Build GRCh38
907321 c.488C>T p.A163V Chr.1: 24533201-24533201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768289364 Chr.1: 24533290-24533290 on Build GRCh38
rs781611544 Chr.1: 24533160-24533160 on Build GRCh38
rs773604708 Chr.1: 24533094-24533094 on Build GRCh38
rs941050209 Chr.1: 24533156-24533156 on Build GRCh38
rs114568126 Chr.1: 24533104-24533104 on Build GRCh38
rs200577938 Chr.1: 24533250-24533250 on Build GRCh38
rs763997085 Chr.1: 24533101-24533101 on Build GRCh38
rs748206064 Chr.1: 24533167-24533167 on Build GRCh38
rs760454730 Chr.1: 24533176-24533176 on Build GRCh38
rs141883360 Chr.1: 24533275-24533277 on Build GRCh38
rs1040287237 Chr.1: 24533154-24533154 on Build GRCh38
rs773447065 Chr.1: 24533173-24533173 on Build GRCh38
rs751382988 Chr.1: 24533032-24533032 on Build GRCh38
rs764063484 Chr.1: 24533178-24533178 on Build GRCh38
rs985460353 Chr.1: 24533124-24533124 on Build GRCh38
rs1022634604 Chr.1: 24533027-24533027 on Build GRCh38
rs12096648 Chr.1: 24533197-24533197 on Build GRCh38
rs565033977 Chr.1: 24533071-24533071 on Build GRCh38
rs752728860 Chr.1: 24533049-24533049 on Build GRCh38
rs992332238 Chr.1: 24533119-24533119 on Build GRCh38
rs578111021 Chr.1: 24533221-24533221 on Build GRCh38
rs750032379 Chr.1: 24533106-24533106 on Build GRCh38
rs974614763 Chr.1: 24533083-24533083 on Build GRCh38
rs375739733 Chr.1: 24533195-24533195 on Build GRCh38
rs758080794 Chr.1: 24533117-24533117 on Build GRCh38
rs766001734 Chr.1: 24533206-24533206 on Build GRCh38
rs770041022 Chr.1: 24533172-24533172 on Build GRCh38
rs368507829 Chr.1: 24533143-24533143 on Build GRCh38
rs137921271 Chr.1: 24533098-24533098 on Build GRCh38
rs12061734 Chr.1: 24533277-24533277 on Build GRCh38
rs756718630 Chr.1: 24533223-24533223 on Build GRCh38
rs776599668 Chr.1: 24533193-24533193 on Build GRCh38
rs781758386 Chr.1: 24533213-24533213 on Build GRCh38
rs769051810 Chr.1: 24533085-24533085 on Build GRCh38
rs199595334 Chr.1: 24533067-24533067 on Build GRCh38
rs756027986 Chr.1: 24533055-24533055 on Build GRCh38
rs189531936 Chr.1: 24533037-24533037 on Build GRCh38
rs776444541 Chr.1: 24533303-24533303 on Build GRCh38
rs749944522 Chr.1: 24533202-24533202 on Build GRCh38
rs982418193 Chr.1: 24533129-24533129 on Build GRCh38
rs376206966 Chr.1: 24533066-24533066 on Build GRCh38
rs758015034 Chr.1: 24533205-24533205 on Build GRCh38
rs764690503 Chr.1: 24533201-24533201 on Build GRCh38
rs196430 Chr.1: 24533175-24533175 on Build GRCh38
rs755493321 Chr.1: 24533152-24533152 on Build GRCh38
rs747514066 Chr.1: 24533082-24533082 on Build GRCh38
rs771180301 Chr.1: 24533246-24533246 on Build GRCh38
rs906847689 Chr.1: 24533271-24533271 on Build GRCh38
rs537312929 Chr.1: 24533090-24533090 on Build GRCh38
rs200941489 Chr.1: 24533174-24533174 on Build GRCh38
rs1001314048 Chr.1: 24533302-24533302 on Build GRCh38
rs200334877 Chr.1: 24533095-24533095 on Build GRCh38
rs747351378 Chr.1: 24533140-24533140 on Build GRCh38
rs112653974 Chr.1: 24533072-24533072 on Build GRCh38
rs368201107 Chr.1: 24533229-24533229 on Build GRCh38
rs777933417 Chr.1: 24533226-24533226 on Build GRCh38
rs934518179 Chr.1: 24533168-24533168 on Build GRCh38
rs146349634 Chr.1: 24533045-24533045 on Build GRCh38
rs751944876 Chr.1: 24533209-24533209 on Build GRCh38
rs536861438 Chr.1: 24533181-24533181 on Build GRCh38
rs769120431 Chr.1: 24533263-24533265 on Build GRCh38
rs918141676 Chr.1: 24533126-24533126 on Build GRCh38
rs748292030 Chr.1: 24533089-24533089 on Build GRCh38
rs375061134 Chr.1: 24533088-24533088 on Build GRCh38
rs196429 Chr.1: 24533127-24533127 on Build GRCh38
rs376349376 Chr.1: 24533105-24533105 on Build GRCh38
rs34116411 Chr.1: 24533097-24533097 on Build GRCh38

View Full Product Details