Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45032495-45032802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RELB
Entrez Gene ID
5971
Gene Name
RELB proto-oncogene, NF-kB subunit
Gene Aliases
I-REL, IREL, REL-B
UniGene
Hs.654402
Gene Chromosome Location
Chr.19: 45001449-45038198 on Build GRCh38
Transcripts
NM_006509.3 XM_005259127.3 XM_005259128.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086284 c.1179G>T p.L393F Chr.19: 45032721-45032721 on Build GRCh38
5650272 c.1002A>G p.S334S Chr.19: 45032544-45032544 on Build GRCh38
163982 c.1198C>T p.R400C Chr.19: 45032740-45032740 on Build GRCh38
7082216 c.1137C>T p.V379V Chr.19: 45032679-45032679 on Build GRCh38
6764827 c.1170C>T p.S390S Chr.19: 45032712-45032712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753160999 Chr.19: 45032595-45032595 on Build GRCh38
rs777161803 Chr.19: 45032744-45032744 on Build GRCh38
rs778309243 Chr.19: 45032719-45032719 on Build GRCh38
rs759228803 Chr.19: 45032659-45032659 on Build GRCh38
rs377464997 Chr.19: 45032722-45032722 on Build GRCh38
rs769788233 Chr.19: 45032646-45032646 on Build GRCh38
rs373225536 Chr.19: 45032697-45032697 on Build GRCh38
rs370561511 Chr.19: 45032740-45032740 on Build GRCh38
rs373675698 Chr.19: 45032630-45032630 on Build GRCh38
rs774057292 Chr.19: 45032551-45032551 on Build GRCh38
rs940958016 Chr.19: 45032665-45032665 on Build GRCh38
rs769438391 Chr.19: 45032525-45032525 on Build GRCh38
rs770622333 Chr.19: 45032548-45032548 on Build GRCh38
rs780229030 Chr.19: 45032710-45032710 on Build GRCh38
rs765733684 Chr.19: 45032679-45032679 on Build GRCh38
rs772333691 Chr.19: 45032519-45032519 on Build GRCh38
rs776684431 Chr.19: 45032521-45032521 on Build GRCh38
rs202054363 Chr.19: 45032598-45032598 on Build GRCh38
rs767065056 Chr.19: 45032583-45032583 on Build GRCh38
rs989604806 Chr.19: 45032520-45032520 on Build GRCh38
rs775189147 Chr.19: 45032741-45032741 on Build GRCh38
rs751787862 Chr.19: 45032781-45032781 on Build GRCh38
rs768095546 Chr.19: 45032742-45032742 on Build GRCh38
rs749105163 Chr.19: 45032656-45032656 on Build GRCh38
rs779259433 Chr.19: 45032616-45032616 on Build GRCh38
rs374270251 Chr.19: 45032631-45032631 on Build GRCh38
rs10424046 Chr.19: 45032778-45032778 on Build GRCh38
rs759035059 Chr.19: 45032576-45032576 on Build GRCh38
rs375363104 Chr.19: 45032764-45032764 on Build GRCh38
rs762459475 Chr.19: 45032539-45032539 on Build GRCh38
rs185766096 Chr.19: 45032763-45032763 on Build GRCh38
rs750636612 Chr.19: 45032685-45032685 on Build GRCh38
rs375120718 Chr.19: 45032518-45032518 on Build GRCh38
rs761265324 Chr.19: 45032596-45032596 on Build GRCh38
rs906425059 Chr.19: 45032745-45032745 on Build GRCh38
rs771520600 Chr.19: 45032664-45032664 on Build GRCh38
rs370104310 Chr.19: 45032676-45032676 on Build GRCh38
rs745463649 Chr.19: 45032733-45032733 on Build GRCh38
rs773085923 Chr.19: 45032535-45032535 on Build GRCh38
rs765651779 Chr.19: 45032766-45032766 on Build GRCh38
rs774992822 Chr.19: 45032667-45032667 on Build GRCh38
rs770682728 Chr.19: 45032729-45032729 on Build GRCh38
rs147289748 Chr.19: 45032640-45032640 on Build GRCh38
rs757625998 Chr.19: 45032601-45032601 on Build GRCh38
rs750720456 Chr.19: 45032622-45032622 on Build GRCh38
rs761020541 Chr.19: 45032674-45032674 on Build GRCh38
rs777753328 Chr.19: 45032713-45032713 on Build GRCh38
rs908357641 Chr.19: 45032621-45032621 on Build GRCh38
rs369508361 Chr.19: 45032522-45032522 on Build GRCh38
rs570938316 Chr.19: 45032765-45032765 on Build GRCh38
rs762213313 Chr.19: 45032677-45032677 on Build GRCh38
rs953440455 Chr.19: 45032634-45032634 on Build GRCh38
rs62117229 Chr.19: 45032605-45032605 on Build GRCh38
rs182578708 Chr.19: 45032730-45032730 on Build GRCh38
rs777367214 Chr.19: 45032647-45032647 on Build GRCh38
rs773728920 Chr.19: 45032777-45032777 on Build GRCh38
rs758834064 Chr.19: 45032658-45032658 on Build GRCh38
rs920803568 Chr.19: 45032694-45032694 on Build GRCh38
rs139280106 Chr.19: 45032712-45032712 on Build GRCh38

View Full Product Details