Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29636728-29636900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NF2
Entrez Gene ID
4771
Gene Name
neurofibromin 2
Gene Aliases
ACN, BANF, SCH
UniGene
Hs.187898
Gene Chromosome Location
Chr.22: 29603556-29698600 on Build GRCh38
Transcripts
NM_000268.3 NM_016418.5 NM_181825.2 NM_181828.2 NM_181829.2 NM_181830.2 NM_181831.2 NM_181832.2 NM_181833.2 XM_017028809.1 XM_017028810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6281357 c.192G>A p.L64L Chr.22: 29636828-29636828 on Build GRCh38
6910239 c.240+1G>T p.? Chr.22: 29636877-29636877 on Build GRCh38
1592917 c.172G>T p.E58* Chr.22: 29636808-29636808 on Build GRCh38
3424105 c.175A>G p.T59A Chr.22: 29636811-29636811 on Build GRCh38
6910240 c.240+1G>T p.? Chr.22: 29636877-29636877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370147621 Chr.22: 29636769-29636769 on Build GRCh38
rs541636141 Chr.22: 29636830-29636830 on Build GRCh38
rs373178836 Chr.22: 29636822-29636822 on Build GRCh38
rs752220604 Chr.22: 29636796-29636796 on Build GRCh38
rs764089682 Chr.22: 29636799-29636799 on Build GRCh38
rs779218238 Chr.22: 29636826-29636826 on Build GRCh38
rs759129920 Chr.22: 29636781-29636781 on Build GRCh38
rs587776562 Chr.22: 29636877-29636877 on Build GRCh38
rs780872661 Chr.22: 29636816-29636816 on Build GRCh38
rs998713939 Chr.22: 29636867-29636867 on Build GRCh38
rs121434261 Chr.22: 29636821-29636821 on Build GRCh38
rs557924117 Chr.22: 29636837-29636837 on Build GRCh38
rs778460074 Chr.22: 29636838-29636838 on Build GRCh38
rs752664255 Chr.22: 29636771-29636771 on Build GRCh38
rs368773485 Chr.22: 29636806-29636806 on Build GRCh38
rs121434259 Chr.22: 29636805-29636805 on Build GRCh38
rs757125309 Chr.22: 29636813-29636813 on Build GRCh38
rs1018147647 Chr.22: 29636780-29636780 on Build GRCh38
rs764901064 Chr.22: 29636790-29636790 on Build GRCh38
rs772274240 Chr.22: 29636832-29636832 on Build GRCh38
rs559558673 Chr.22: 29636844-29636844 on Build GRCh38
rs762440698 Chr.22: 29636797-29636797 on Build GRCh38
rs17851784 Chr.22: 29636868-29636868 on Build GRCh38
rs587776564 Chr.22: 29636761-29636762 on Build GRCh38
rs747447569 Chr.22: 29636840-29636840 on Build GRCh38
rs587776565 Chr.22: 29636841-29636847 on Build GRCh38

View Full Product Details