Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4095328-4095470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K2
Entrez Gene ID
5605
Gene Name
mitogen-activated protein kinase kinase 2
Gene Aliases
CFC4, MAPKK2, MEK2, MKK2, PRKMK2
UniGene
Hs.465627
Gene Chromosome Location
Chr.19: 4090321-4124184 on Build GRCh38
Transcripts
NM_030662.3 XM_006722799.2 XM_017026989.1 XM_017026990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6366469 c.1021G>A p.D341N Chr.19: 4095413-4095413 on Build GRCh38
3960073 c.1021G>C p.D341H Chr.19: 4095413-4095413 on Build GRCh38
6557268 c.1013T>A p.F338Y Chr.19: 4095421-4095421 on Build GRCh38
5782190 c.999G>A p.L333L Chr.19: 4095435-4095435 on Build GRCh38
6557266 c.1014C>A p.F338L Chr.19: 4095420-4095420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747043902 Chr.19: 4095363-4095363 on Build GRCh38
rs587781029 Chr.19: 4095375-4095375 on Build GRCh38
rs755029740 Chr.19: 4095376-4095376 on Build GRCh38
rs564067943 Chr.19: 4095413-4095413 on Build GRCh38
rs541565088 Chr.19: 4095350-4095350 on Build GRCh38
rs779065052 Chr.19: 4095427-4095427 on Build GRCh38
rs906888337 Chr.19: 4095372-4095372 on Build GRCh38
rs963596101 Chr.19: 4095403-4095403 on Build GRCh38
rs1015139062 Chr.19: 4095398-4095398 on Build GRCh38
rs541304144 Chr.19: 4095377-4095377 on Build GRCh38
rs587781028 Chr.19: 4095429-4095429 on Build GRCh38
rs970893474 Chr.19: 4095361-4095361 on Build GRCh38
rs727504353 Chr.19: 4095449-4095449 on Build GRCh38
rs1024494837 Chr.19: 4095357-4095357 on Build GRCh38
rs561722470 Chr.19: 4095354-4095354 on Build GRCh38
rs201128231 Chr.19: 4095355-4095355 on Build GRCh38
rs1057518088 Chr.19: 4095395-4095395 on Build GRCh38
rs1005472404 Chr.19: 4095381-4095381 on Build GRCh38
rs1011864039 Chr.19: 4095384-4095384 on Build GRCh38
rs1047318082 Chr.19: 4095367-4095367 on Build GRCh38
rs192389729 Chr.19: 4095414-4095414 on Build GRCh38
rs1057517900 Chr.19: 4095398-4095398 on Build GRCh38
rs35737722 Chr.19: 4095405-4095405 on Build GRCh38

View Full Product Details