Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 135190218-135190392 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYB
Entrez Gene ID
4602
Gene Name
MYB proto-oncogene, transcription factor
Gene Aliases
Cmyb, c-myb, c-myb_CDS, efg
UniGene
Hs.606320
Gene Chromosome Location
Chr.6: 135180981-135219173 on Build GRCh38
Transcripts
NM_001130172.1 NM_001130173.1 NM_001161656.1 NM_001161657.1 NM_001161658.1 NM_001161659.1 NM_001161660.1 NM_005375.3 NR_134958.1 NR_134959.1 NR_134960.1 NR_134961.1 NR_134962.1 NR_134963.1 NR_134964.1 NR_134965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7090827 c.421G>C p.E141Q Chr.6: 135190241-135190241 on Build GRCh38
5959597 c.498G>A p.W166* Chr.6: 135190318-135190318 on Build GRCh38
5564748 c.527G>A p.R176Q Chr.6: 135190347-135190347 on Build GRCh38
4704275 c.465T>G p.I155M Chr.6: 135190285-135190285 on Build GRCh38
1698038 c.437C>T p.S146F Chr.6: 135190257-135190257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012919506 Chr.6: 135190327-135190327 on Build GRCh38
rs770089949 Chr.6: 135190306-135190306 on Build GRCh38
rs778119027 Chr.6: 135190328-135190328 on Build GRCh38
rs749730179 Chr.6: 135190346-135190346 on Build GRCh38
rs772220664 Chr.6: 135190369-135190369 on Build GRCh38
rs771042810 Chr.6: 135190356-135190356 on Build GRCh38
rs748681326 Chr.6: 135190264-135190264 on Build GRCh38
rs774680496 Chr.6: 135190357-135190357 on Build GRCh38
rs781635664 Chr.6: 135190246-135190246 on Build GRCh38
rs41287046 Chr.6: 135190366-135190366 on Build GRCh38

View Full Product Details