Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177231849-177231997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFE2L2
Entrez Gene ID
4780
Gene Name
nuclear factor, erythroid 2 like 2
Gene Aliases
HEBP1, NRF2
UniGene
Hs.744006
Gene Chromosome Location
Chr.2: 177230303-177265131 on Build GRCh38
Transcripts
NM_001145412.3 NM_001145413.3 NM_001313900.1 NM_001313901.1 NM_001313902.1 NM_001313903.1 NM_001313904.1 NM_006164.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6946092 c.739C>A p.L247I Chr.2: 177231864-177231864 on Build GRCh38
6981439 c.614delA p.D205fs*16 Chr.2: 177231989-177231989 on Build GRCh38
5785377 c.670G>C p.D224H Chr.2: 177231933-177231933 on Build GRCh38
6201621 c.634A>T p.M212L Chr.2: 177231969-177231969 on Build GRCh38
3961570 c.689C>T p.S230L Chr.2: 177231914-177231914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771141150 Chr.2: 177231957-177231957 on Build GRCh38
rs754094243 Chr.2: 177231898-177231898 on Build GRCh38
rs746497256 Chr.2: 177231925-177231925 on Build GRCh38
rs369394957 Chr.2: 177231966-177231966 on Build GRCh38
rs369687249 Chr.2: 177231909-177231909 on Build GRCh38
rs372577933 Chr.2: 177231936-177231936 on Build GRCh38
rs778786297 Chr.2: 177231903-177231903 on Build GRCh38
rs756983196 Chr.2: 177231900-177231900 on Build GRCh38
rs779897588 Chr.2: 177231911-177231911 on Build GRCh38
rs774762514 Chr.2: 177231959-177231959 on Build GRCh38
rs60132461 Chr.2: 177231947-177231947 on Build GRCh38
rs1035583151 Chr.2: 177231922-177231924 on Build GRCh38
rs760780542 Chr.2: 177231885-177231885 on Build GRCh38
rs764274303 Chr.2: 177231894-177231894 on Build GRCh38
rs905676824 Chr.2: 177231892-177231892 on Build GRCh38
rs576075280 Chr.2: 177231941-177231941 on Build GRCh38
rs35557421 Chr.2: 177231896-177231896 on Build GRCh38
rs909198253 Chr.2: 177231890-177231890 on Build GRCh38
rs139187151 Chr.2: 177231906-177231906 on Build GRCh38
rs776021543 Chr.2: 177231967-177231967 on Build GRCh38

View Full Product Details