Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44249707-44249866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSP90AB1
Entrez Gene ID
3326
Gene Name
heat shock protein 90 alpha family class B member 1
Gene Aliases
D6S182, HSP84, HSP90B, HSPC2, HSPCB
UniGene
Hs.509736
Gene Chromosome Location
Chr.6: 44246166-44253888 on Build GRCh38
Transcripts
NM_001271969.1 NM_001271970.1 NM_001271971.1 NM_001271972.1 NM_007355.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371565 c.451G>A p.D151N Chr.6: 44249771-44249771 on Build GRCh38
6682510 c.503G>A p.R168H Chr.6: 44249823-44249823 on Build GRCh38
3684323 c.504T>C p.R168R Chr.6: 44249824-44249824 on Build GRCh38
5710145 c.445C>T p.H149Y Chr.6: 44249765-44249765 on Build GRCh38
5880955 c.502C>G p.R168G Chr.6: 44249822-44249822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113937534 Chr.6: 44249743-44249743 on Build GRCh38
rs755971334 Chr.6: 44249776-44249776 on Build GRCh38
rs777161405 Chr.6: 44249798-44249798 on Build GRCh38
rs200278311 Chr.6: 44249827-44249827 on Build GRCh38
rs11538981 Chr.6: 44249812-44249812 on Build GRCh38
rs111881934 Chr.6: 44249824-44249824 on Build GRCh38
rs528456416 Chr.6: 44249792-44249792 on Build GRCh38
rs201866283 Chr.6: 44249834-44249834 on Build GRCh38
rs150511888 Chr.6: 44249778-44249778 on Build GRCh38
rs764170093 Chr.6: 44249734-44249734 on Build GRCh38
rs11538970 Chr.6: 44249787-44249787 on Build GRCh38
rs757172689 Chr.6: 44249765-44249765 on Build GRCh38
rs11538976 Chr.6: 44249823-44249823 on Build GRCh38
rs201355139 Chr.6: 44249839-44249839 on Build GRCh38
rs774316367 Chr.6: 44249833-44249833 on Build GRCh38
rs751590992 Chr.6: 44249752-44249752 on Build GRCh38
rs370988754 Chr.6: 44249822-44249822 on Build GRCh38
rs560978430 Chr.6: 44249770-44249770 on Build GRCh38
rs746296192 Chr.6: 44249806-44249806 on Build GRCh38
rs768359852 Chr.6: 44249781-44249781 on Build GRCh38
rs750226490 Chr.6: 44249771-44249771 on Build GRCh38
rs775772305 Chr.6: 44249815-44249815 on Build GRCh38
rs778322003 Chr.6: 44249785-44249785 on Build GRCh38

View Full Product Details