Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17214916-17215076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLCN
Entrez Gene ID
201163
Gene Name
folliculin
Gene Aliases
BHD, FLCL
UniGene
Hs.31652
Gene Chromosome Location
Chr.17: 17206945-17237188 on Build GRCh38
Transcripts
NM_144997.5 XM_011523714.2 XM_011523716.1 XM_011523718.2 XM_011523719.2 XM_011523721.2 XM_017024304.1 XM_017024305.1 XM_017024306.1 XM_017024307.1 XM_017024308.1 XM_017024309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976079 c.1537A>C p.N513H Chr.17: 17214986-17214986 on Build GRCh38
6940451 c.1463C>T p.A488V Chr.17: 17215060-17215060 on Build GRCh38
6227827 c.1466C>A p.A489D Chr.17: 17215057-17215057 on Build GRCh38
6947846 c.1492_1498delGATGTGG p.D498fs*13 Chr.17: 17215025-17215031 on Build GRCh38
6949777 c.1492G>T p.D498Y Chr.17: 17215031-17215031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs398124530 Chr.17: 17214990-17214990 on Build GRCh38
rs879255679 Chr.17: 17215032-17215033 on Build GRCh38
rs879255680 Chr.17: 17214995-17214995 on Build GRCh38
rs199643834 Chr.17: 17215000-17215000 on Build GRCh38
rs398124529 Chr.17: 17214999-17215001 on Build GRCh38
rs530572650 Chr.17: 17214959-17214959 on Build GRCh38
rs12451312 Chr.17: 17214975-17214975 on Build GRCh38
rs531459106 Chr.17: 17214993-17214993 on Build GRCh38
rs758516602 Chr.17: 17215044-17215044 on Build GRCh38
rs772863587 Chr.17: 17214963-17214963 on Build GRCh38
rs879255681 Chr.17: 17214993-17214995 on Build GRCh38
rs886042033 Chr.17: 17215019-17215019 on Build GRCh38
rs886037608 Chr.17: 17214987-17214990 on Build GRCh38
rs112111994 Chr.17: 17214971-17214971 on Build GRCh38
rs779837933 Chr.17: 17215047-17215047 on Build GRCh38
rs879255682 Chr.17: 17214991-17214991 on Build GRCh38
rs750535468 Chr.17: 17215036-17215036 on Build GRCh38
rs778904029 Chr.17: 17215015-17215015 on Build GRCh38
rs768168088 Chr.17: 17214974-17214974 on Build GRCh38
rs939272152 Chr.17: 17215026-17215026 on Build GRCh38
rs757471403 Chr.17: 17215011-17215011 on Build GRCh38
rs1036053975 Chr.17: 17214950-17214950 on Build GRCh38
rs776135068 Chr.17: 17214945-17214945 on Build GRCh38
rs769365049 Chr.17: 17214961-17214961 on Build GRCh38
rs927844338 Chr.17: 17215014-17215014 on Build GRCh38
rs1045022900 Chr.17: 17215013-17215013 on Build GRCh38
rs371523217 Chr.17: 17214949-17214949 on Build GRCh38
rs376715412 Chr.17: 17215010-17215010 on Build GRCh38
rs763899011 Chr.17: 17214970-17214971 on Build GRCh38

View Full Product Details