Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22625192-22625362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
UniGene
Hs.632151
Gene Chromosome Location
Chr.11: 22622533-22625841 on Build GRCh38
Transcripts
NM_022725.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
26271 c.554C>T p.P185L Chr.11: 22625257-22625257 on Build GRCh38
428847 c.559A>T p.R187W Chr.11: 22625252-22625252 on Build GRCh38
6391218 c.583C>T p.R195C Chr.11: 22625228-22625228 on Build GRCh38
5667384 c.547G>C p.E183Q Chr.11: 22625264-22625264 on Build GRCh38
6025139 c.574C>A p.L192M Chr.11: 22625237-22625237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540547787 Chr.11: 22625252-22625252 on Build GRCh38
rs950789710 Chr.11: 22625305-22625305 on Build GRCh38
rs113910234 Chr.11: 22625254-22625254 on Build GRCh38
rs772589448 Chr.11: 22625274-22625274 on Build GRCh38
rs764066836 Chr.11: 22625293-22625293 on Build GRCh38
rs761801623 Chr.11: 22625311-22625311 on Build GRCh38
rs761018953 Chr.11: 22625291-22625291 on Build GRCh38
rs893075597 Chr.11: 22625231-22625231 on Build GRCh38
rs754014970 Chr.11: 22625309-22625309 on Build GRCh38
rs587778340 Chr.11: 22625326-22625327 on Build GRCh38
rs992569572 Chr.11: 22625287-22625287 on Build GRCh38
rs146219377 Chr.11: 22625238-22625238 on Build GRCh38
rs775816396 Chr.11: 22625282-22625282 on Build GRCh38
rs746753493 Chr.11: 22625218-22625218 on Build GRCh38
rs751518324 Chr.11: 22625325-22625325 on Build GRCh38
rs1000985563 Chr.11: 22625217-22625217 on Build GRCh38
rs759924821 Chr.11: 22625272-22625272 on Build GRCh38
rs766279442 Chr.11: 22625315-22625315 on Build GRCh38
rs772831530 Chr.11: 22625251-22625251 on Build GRCh38
rs865886264 Chr.11: 22625237-22625237 on Build GRCh38
rs917900309 Chr.11: 22625292-22625292 on Build GRCh38
rs769259296 Chr.11: 22625246-22625246 on Build GRCh38
rs774728053 Chr.11: 22625264-22625264 on Build GRCh38
rs754783383 Chr.11: 22625236-22625236 on Build GRCh38

View Full Product Details